Assay Details
Gene Symbol: PDHX-FGFR2 fusion | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | OM_PDHX-FGFR2_P1F7.0 | - | 67 | 83 |

Gene Symbol: PDHX-FGFR2 fusion | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | OM_PDHX-FGFR2_P1F7.0 | - | 67 | 83 |