This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05061418_ft
See other BCR-FGFR1 FUSION GE Assays ›
Gene Symbol:
BCR-FGFR1 fusion BCR-FGFR1 fusion
Gene Name:
FUSION
FUSION
Gene Aliases:
-
BCR, FGFR1
Chromosome Location:
-
Chromosome Location:
-
Species:
Human
Species Specific ID (Flybase ID):
-
-
Assay ID Hs05061418_ft
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: BCR-FGFR1 fusion
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA OM_BCR-FGFR1_B4F10.0 - 45 119
Gene Symbol: BCR-FGFR1 fusion
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AJ298916.1 - 4 602 119

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Fusion Homo sapiens

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0