This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05060768_s1
See other IL1RL2 GE Assays ›
Gene Symbol:
IL1RL2
Gene Name:
interleukin 1 receptor like 2
Gene Aliases:
IL-1Rrp2, IL-36R, IL1R-rp2, IL1RRP2
Chromosome Location:
Chr.2: 102186973 - 102242906 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05060768_s1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: IL1RL2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005264035.1 XP_005264092.1 12 2470 79
XM_005264036.1 XP_005264093.1 12 2392 79
XM_017005175.1 XP_016860664.1 10 2178 79
XM_017005176.1 XP_016860665.1 9 2106 79
XM_017005178.1 XP_016860667.1 11 2223 79
XM_017005179.1 XP_016860668.1 11 2476 79

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

transmembrane signal receptor

Gene Ontology Categories:

Function(s) Process(es)

inflammatory response
cellular defense response
signal transduction
positive regulation of interleukin-6 production
innate immune response
positive regulation of T cell differentiation
regulation of inflammatory response
interleukin-1-mediated signaling pathway
interleukin-1 receptor activity
interleukin-1, Type I, activating receptor activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0