This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05058752_s1
See other SLC5A1 GE Assays ›
Gene Symbol:
SLC5A1
Gene Name:
solute carrier family 5 member 1
Gene Aliases:
D22S675, NAGT, SGLT1
Chromosome Location:
Chr.22: 32043032 - 32113029 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05058752_s1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SLC5A1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_000343.3 NP_000334.1 1 120 84
XM_011530331.1 XP_011528633.1 1 102 84
GenBank mRNA DA429018.1 - 1 102 84
DQ656053.2 - 1 120 84

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the NM_000343.3, XM_011530331.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene SLC5A1

Panther Classification:

Molecular Function -

secondary carrier transporter

Gene Ontology Categories:

Function(s) Process(es)

intestinal D-glucose absorption
sodium ion transport
hexose transport
glucose transport
glucose transmembrane transport
glucose:sodium symporter activity
protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC2200007120.hg.1 ENST00000266088.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007120.hg.1 ENST00000266088.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007120.hg.1 ENST00000266088.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007120.hg.1 ENST00000266088.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007120.hg.1 ENST00000266088.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007120.hg.1 ENST00000266088.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007120.hg.1 ENST00000266088.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007120.hg.1 ENST00000266088.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007120.hg.1 ENST00000266088.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007120.hg.1 ENST00000266088.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007120.hg.1 ENST00000266088.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007120.hg.1 ENST00000266088.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007120.hg.1 ENST00000266088.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007120.hg.1 ENST00000266088.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007120.hg.1 ENST00000266088.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007120.hg.1 ENST00000266088.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007120.hg.1 ENST00000266088.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007120.hg.1 ENST00000266088.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007120.hg.1 ENST00000477969.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007120.hg.1 ENST00000477969.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007120.hg.1 ENST00000477969.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007120.hg.1 ENST00000477969.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007120.hg.1 ENST00000477969.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007120.hg.1 ENST00000477969.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007120.hg.1 ENST00000486394.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007120.hg.1 ENST00000486394.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007120.hg.1 ENST00000486394.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007120.hg.1 ENST00000486394.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007120.hg.1 ENST00000543737.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007120.hg.1 ENST00000543737.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007120.hg.1 ENST00000543737.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007120.hg.1 ENST00000543737.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007120.hg.1 ENST00000543737.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007120.hg.1 ENST00000543737.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007120.hg.1 ENST00000543737.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007120.hg.1 ENST00000543737.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007120.hg.1 ENST00000543737.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007120.hg.1 ENST00000543737.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007120.hg.1 ENST00000543737.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007120.hg.1 ENST00000543737.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007120.hg.1 ENST00000543737.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007120.hg.1 ENST00000543737.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007120.hg.1 ENST00000543737.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007120.hg.1 ENST00000543737.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200007120.hg.1 NM_000343.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200007120.hg.1 NM_000343.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200007120.hg.1 NM_000343.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200007120.hg.1 NM_000343.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200007120.hg.1 NM_000343.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200007120.hg.1 NM_000343.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200007120.hg.1 NM_000343.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200007120.hg.1 NM_000343.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200007120.hg.1 NM_000343.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200007120.hg.1 NM_000343.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200007120.hg.1 NM_000343.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200007120.hg.1 NM_000343.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200007120.hg.1 NM_000343.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200007120.hg.1 NM_000343.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200007120.hg.1 NM_000343.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200007120.hg.1 NM_000343.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200007120.hg.1 NM_000343.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200007120.hg.1 NM_000343.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200007120.hg.1 NM_001256314.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200007120.hg.1 NM_001256314.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200007120.hg.1 NM_001256314.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200007120.hg.1 NM_001256314.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200007120.hg.1 NM_001256314.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200007120.hg.1 NM_001256314.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200007120.hg.1 NM_001256314.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200007120.hg.1 NM_001256314.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200007120.hg.1 NM_001256314.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200007120.hg.1 NM_001256314.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200007120.hg.1 NM_001256314.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200007120.hg.1 NM_001256314.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200007120.hg.1 NM_001256314.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200007120.hg.1 NM_001256314.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200007120.hg.1 NM_001256314.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200007120.hg.1 NM_001256314.1 Refseq
Affymetrix Human Clariom D Assay TC2200007120.hg.1 ENST00000266088.8 Ensembl
Affymetrix Human Clariom D Assay TC2200007120.hg.1 ENST00000266088.8 Ensembl
Affymetrix Human Clariom D Assay TC2200007120.hg.1 ENST00000266088.8 Ensembl
Affymetrix Human Clariom D Assay TC2200007120.hg.1 ENST00000266088.8 Ensembl
Affymetrix Human Clariom D Assay TC2200007120.hg.1 ENST00000266088.8 Ensembl
Affymetrix Human Clariom D Assay TC2200007120.hg.1 ENST00000266088.8 Ensembl
Affymetrix Human Clariom D Assay TC2200007120.hg.1 ENST00000266088.8 Ensembl
Affymetrix Human Clariom D Assay TC2200007120.hg.1 ENST00000266088.8 Ensembl
Affymetrix Human Clariom D Assay TC2200007120.hg.1 ENST00000266088.8 Ensembl
Affymetrix Human Clariom D Assay TC2200007120.hg.1 ENST00000266088.8 Ensembl
Affymetrix Human Clariom D Assay TC2200007120.hg.1 ENST00000266088.8 Ensembl
Affymetrix Human Clariom D Assay TC2200007120.hg.1 ENST00000266088.8 Ensembl
Affymetrix Human Clariom D Assay TC2200007120.hg.1 ENST00000266088.8 Ensembl
Affymetrix Human Clariom D Assay TC2200007120.hg.1 ENST00000266088.8 Ensembl
Affymetrix Human Clariom D Assay TC2200007120.hg.1 ENST00000266088.8 Ensembl
Affymetrix Human Clariom D Assay TC2200007120.hg.1 ENST00000266088.8 Ensembl
Affymetrix Human Clariom D Assay TC2200007120.hg.1 ENST00000266088.8 Ensembl
Affymetrix Human Clariom D Assay TC2200007120.hg.1 ENST00000266088.8 Ensembl
Affymetrix Human Clariom D Assay TC2200007120.hg.1 ENST00000477969.1 Ensembl
Affymetrix Human Clariom D Assay TC2200007120.hg.1 ENST00000477969.1 Ensembl
Affymetrix Human Clariom D Assay TC2200007120.hg.1 ENST00000477969.1 Ensembl
Affymetrix Human Clariom D Assay TC2200007120.hg.1 ENST00000477969.1 Ensembl
Affymetrix Human Clariom D Assay TC2200007120.hg.1 ENST00000477969.1 Ensembl
Affymetrix Human Clariom D Assay TC2200007120.hg.1 ENST00000477969.1 Ensembl
Affymetrix Human Clariom D Assay TC2200007120.hg.1 ENST00000486394.1 Ensembl
Affymetrix Human Clariom D Assay TC2200007120.hg.1 ENST00000486394.1 Ensembl
Affymetrix Human Clariom D Assay TC2200007120.hg.1 ENST00000486394.1 Ensembl
Affymetrix Human Clariom D Assay TC2200007120.hg.1 ENST00000486394.1 Ensembl
Affymetrix Human Clariom D Assay TC2200007120.hg.1 ENST00000543737.2 Ensembl
Affymetrix Human Clariom D Assay TC2200007120.hg.1 ENST00000543737.2 Ensembl
Affymetrix Human Clariom D Assay TC2200007120.hg.1 ENST00000543737.2 Ensembl
Affymetrix Human Clariom D Assay TC2200007120.hg.1 ENST00000543737.2 Ensembl
Affymetrix Human Clariom D Assay TC2200007120.hg.1 ENST00000543737.2 Ensembl
Affymetrix Human Clariom D Assay TC2200007120.hg.1 ENST00000543737.2 Ensembl
Affymetrix Human Clariom D Assay TC2200007120.hg.1 ENST00000543737.2 Ensembl
Affymetrix Human Clariom D Assay TC2200007120.hg.1 ENST00000543737.2 Ensembl
Affymetrix Human Clariom D Assay TC2200007120.hg.1 ENST00000543737.2 Ensembl
Affymetrix Human Clariom D Assay TC2200007120.hg.1 ENST00000543737.2 Ensembl
Affymetrix Human Clariom D Assay TC2200007120.hg.1 ENST00000543737.2 Ensembl
Affymetrix Human Clariom D Assay TC2200007120.hg.1 ENST00000543737.2 Ensembl
Affymetrix Human Clariom D Assay TC2200007120.hg.1 ENST00000543737.2 Ensembl
Affymetrix Human Clariom D Assay TC2200007120.hg.1 ENST00000543737.2 Ensembl
Affymetrix Human Clariom D Assay TC2200007120.hg.1 ENST00000543737.2 Ensembl
Affymetrix Human Clariom D Assay TC2200007120.hg.1 ENST00000543737.2 Ensembl
Affymetrix Human Clariom D Assay TC2200007120.hg.1 NM_000343.3 Refseq
Affymetrix Human Clariom D Assay TC2200007120.hg.1 NM_000343.3 Refseq
Affymetrix Human Clariom D Assay TC2200007120.hg.1 NM_000343.3 Refseq
Affymetrix Human Clariom D Assay TC2200007120.hg.1 NM_000343.3 Refseq
Affymetrix Human Clariom D Assay TC2200007120.hg.1 NM_000343.3 Refseq
Affymetrix Human Clariom D Assay TC2200007120.hg.1 NM_000343.3 Refseq
Affymetrix Human Clariom D Assay TC2200007120.hg.1 NM_000343.3 Refseq
Affymetrix Human Clariom D Assay TC2200007120.hg.1 NM_000343.3 Refseq
Affymetrix Human Clariom D Assay TC2200007120.hg.1 NM_000343.3 Refseq
Affymetrix Human Clariom D Assay TC2200007120.hg.1 NM_000343.3 Refseq
Affymetrix Human Clariom D Assay TC2200007120.hg.1 NM_000343.3 Refseq
Affymetrix Human Clariom D Assay TC2200007120.hg.1 NM_000343.3 Refseq
Affymetrix Human Clariom D Assay TC2200007120.hg.1 NM_000343.3 Refseq
Affymetrix Human Clariom D Assay TC2200007120.hg.1 NM_000343.3 Refseq
Affymetrix Human Clariom D Assay TC2200007120.hg.1 NM_000343.3 Refseq
Affymetrix Human Clariom D Assay TC2200007120.hg.1 NM_000343.3 Refseq
Affymetrix Human Clariom D Assay TC2200007120.hg.1 NM_000343.3 Refseq
Affymetrix Human Clariom D Assay TC2200007120.hg.1 NM_000343.3 Refseq
Affymetrix Human Clariom D Assay TC2200007120.hg.1 NM_001256314.1 Refseq
Affymetrix Human Clariom D Assay TC2200007120.hg.1 NM_001256314.1 Refseq
Affymetrix Human Clariom D Assay TC2200007120.hg.1 NM_001256314.1 Refseq
Affymetrix Human Clariom D Assay TC2200007120.hg.1 NM_001256314.1 Refseq
Affymetrix Human Clariom D Assay TC2200007120.hg.1 NM_001256314.1 Refseq
Affymetrix Human Clariom D Assay TC2200007120.hg.1 NM_001256314.1 Refseq
Affymetrix Human Clariom D Assay TC2200007120.hg.1 NM_001256314.1 Refseq
Affymetrix Human Clariom D Assay TC2200007120.hg.1 NM_001256314.1 Refseq
Affymetrix Human Clariom D Assay TC2200007120.hg.1 NM_001256314.1 Refseq
Affymetrix Human Clariom D Assay TC2200007120.hg.1 NM_001256314.1 Refseq
Affymetrix Human Clariom D Assay TC2200007120.hg.1 NM_001256314.1 Refseq
Affymetrix Human Clariom D Assay TC2200007120.hg.1 NM_001256314.1 Refseq
Affymetrix Human Clariom D Assay TC2200007120.hg.1 NM_001256314.1 Refseq
Affymetrix Human Clariom D Assay TC2200007120.hg.1 NM_001256314.1 Refseq
Affymetrix Human Clariom D Assay TC2200007120.hg.1 NM_001256314.1 Refseq
Affymetrix Human Clariom D Assay TC2200007120.hg.1 NM_001256314.1 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0