Assay Details
Gene Symbol: LRP8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK122887.1 | - | 1 | 146 | 131 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |

Gene Symbol: LRP8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK122887.1 | - | 1 | 146 | 131 |
Set Membership: |
![]() ![]() ![]() ![]() |