This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05054479_s1
See other GATA6 GE Assays ›
Gene Symbol:
GATA6
Gene Name:
GATA binding protein 6
Gene Aliases:
-
Chromosome Location:
Chr.18: 22169437 - 22202528 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05054479_s1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GATA6
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_005257.5 NP_005248.2 1 80 93
GenBank mRNA BC027891.1 - 1 62 93
CN280495.1 - 1 66 93
HY052912.1 - 1 80 93
X95701.1 - 1 74 93

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the BC027891.1, NM_005257.5, X95701.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene GATA6

Panther Classification:

Molecular Function -

DNA-binding transcription factor

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
in utero embryonic development
liver development
outflow tract septum morphogenesis
type B pancreatic cell differentiation
pancreatic A cell differentiation
transcription from RNA polymerase II promoter
phospholipid metabolic process
endodermal cell fate determination
blood coagulation
male gonad development
cardiac muscle hypertrophy in response to stress
positive regulation of BMP signaling pathway
negative regulation of transforming growth factor beta1 production
negative regulation of transforming growth factor beta2 production
tube morphogenesis
response to drug
negative regulation of apoptotic process
response to estrogen
cellular protein metabolic process
cell fate commitment
positive regulation of angiogenesis
negative regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
cell development
organ formation
smooth muscle cell differentiation
positive regulation of cardioblast differentiation
cardiac muscle cell differentiation
positive regulation of cardiac muscle cell proliferation
lung saccule development
Clara cell differentiation
Type II pneumocyte differentiation
intestinal epithelial cell differentiation
cardiac vascular smooth muscle cell differentiation
response to growth factor
positive regulation of cell cycle arrest
cellular response to gonadotropin stimulus
cellular response to hypoxia
cellular response to BMP stimulus
RNA polymerase II regulatory region sequence-specific DNA binding
RNA polymerase II core promoter sequence-specific DNA binding
RNA polymerase II transcription factor activity, sequence-specific DNA binding
RNA polymerase II transcription factor binding
RNA polymerase II repressing transcription factor binding
transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding
chromatin binding
transcription factor activity, sequence-specific DNA binding
transcription factor activity, RNA polymerase II distal enhancer sequence-specific binding
protein binding
transcription factor binding
zinc ion binding
protein kinase binding
transcription regulatory region DNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1800006852.hg.1 ENST00000269216.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800006852.hg.1 ENST00000269216.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800006852.hg.1 ENST00000269216.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800006852.hg.1 ENST00000269216.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800006852.hg.1 ENST00000269216.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800006852.hg.1 ENST00000269216.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800006852.hg.1 ENST00000269216.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800006852.hg.1 ENST00000269216.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800006852.hg.1 ENST00000269216.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800006852.hg.1 ENST00000269216.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800006852.hg.1 ENST00000581694.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800006852.hg.1 ENST00000581694.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800006852.hg.1 ENST00000581694.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800006852.hg.1 ENST00000581694.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800006852.hg.1 ENST00000581694.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800006852.hg.1 ENST00000581694.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800006852.hg.1 ENST00000581694.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800006852.hg.1 ENST00000581694.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800006852.hg.1 ENST00000581694.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800006852.hg.1 NM_005257.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800006852.hg.1 NM_005257.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800006852.hg.1 NM_005257.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800006852.hg.1 NM_005257.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800006852.hg.1 NM_005257.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800006852.hg.1 NM_005257.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800006852.hg.1 NM_005257.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800006852.hg.1 NM_005257.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800006852.hg.1 NM_005257.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800006852.hg.1 NM_005257.5 Refseq
Affymetrix Human Clariom D Assay TC1800006852.hg.1 ENST00000269216.7 Ensembl
Affymetrix Human Clariom D Assay TC1800006852.hg.1 ENST00000269216.7 Ensembl
Affymetrix Human Clariom D Assay TC1800006852.hg.1 ENST00000269216.7 Ensembl
Affymetrix Human Clariom D Assay TC1800006852.hg.1 ENST00000269216.7 Ensembl
Affymetrix Human Clariom D Assay TC1800006852.hg.1 ENST00000269216.7 Ensembl
Affymetrix Human Clariom D Assay TC1800006852.hg.1 ENST00000269216.7 Ensembl
Affymetrix Human Clariom D Assay TC1800006852.hg.1 ENST00000269216.7 Ensembl
Affymetrix Human Clariom D Assay TC1800006852.hg.1 ENST00000269216.7 Ensembl
Affymetrix Human Clariom D Assay TC1800006852.hg.1 ENST00000269216.7 Ensembl
Affymetrix Human Clariom D Assay TC1800006852.hg.1 ENST00000269216.7 Ensembl
Affymetrix Human Clariom D Assay TC1800006852.hg.1 ENST00000581694.1 Ensembl
Affymetrix Human Clariom D Assay TC1800006852.hg.1 ENST00000581694.1 Ensembl
Affymetrix Human Clariom D Assay TC1800006852.hg.1 ENST00000581694.1 Ensembl
Affymetrix Human Clariom D Assay TC1800006852.hg.1 ENST00000581694.1 Ensembl
Affymetrix Human Clariom D Assay TC1800006852.hg.1 ENST00000581694.1 Ensembl
Affymetrix Human Clariom D Assay TC1800006852.hg.1 ENST00000581694.1 Ensembl
Affymetrix Human Clariom D Assay TC1800006852.hg.1 ENST00000581694.1 Ensembl
Affymetrix Human Clariom D Assay TC1800006852.hg.1 ENST00000581694.1 Ensembl
Affymetrix Human Clariom D Assay TC1800006852.hg.1 ENST00000581694.1 Ensembl
Affymetrix Human Clariom D Assay TC1800006852.hg.1 NM_005257.5 Refseq
Affymetrix Human Clariom D Assay TC1800006852.hg.1 NM_005257.5 Refseq
Affymetrix Human Clariom D Assay TC1800006852.hg.1 NM_005257.5 Refseq
Affymetrix Human Clariom D Assay TC1800006852.hg.1 NM_005257.5 Refseq
Affymetrix Human Clariom D Assay TC1800006852.hg.1 NM_005257.5 Refseq
Affymetrix Human Clariom D Assay TC1800006852.hg.1 NM_005257.5 Refseq
Affymetrix Human Clariom D Assay TC1800006852.hg.1 NM_005257.5 Refseq
Affymetrix Human Clariom D Assay TC1800006852.hg.1 NM_005257.5 Refseq
Affymetrix Human Clariom D Assay TC1800006852.hg.1 NM_005257.5 Refseq
Affymetrix Human Clariom D Assay TC1800006852.hg.1 NM_005257.5 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-56ffd697dc-88d4p:80/100.66.130.94:80.
git-commit: 082a6d415be982b1f92c35e55dd71609d4576d3e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0