This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05052713_s1
See other UBR1 GE Assays ›
Gene Symbol:
UBR1
Gene Name:
ubiquitin protein ligase E3 component n-recognin 1
Gene Aliases:
JBS
Chromosome Location:
Chr.15: 42942897 - 43106088 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05052713_s1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: UBR1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_174916.2 NP_777576.1 25 2696 74
GenBank mRNA AF061556.1 - 7 527 74
AF525401.1 - 25 2579 74
AK027803.1 - 22 2137 74
AK056441.1 - 25 2646 74
AK292516.1 - 25 2643 74
AK302489.1 - 25 2723 74
AY061886.1 - 25 2630 74
BC113505.1 - 25 2708 8322557 74
BC113507.1 - 25 2708 8322559 74
CR749326.1 - 7 1750 74

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene UBR1

Panther Classification:

Molecular Function -

ubiquitin-protein ligase

Gene Ontology Categories:

Function(s) Process(es)

protein ubiquitination
negative regulation of TOR signaling
cellular response to leucine
ubiquitin-dependent protein catabolic process via the N-end rule pathway
zinc ion binding
ligase activity
ubiquitin protein ligase activity
leucine binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0