This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05050520_s1
See other GPR132 GE Assays ›
Gene Symbol:
GPR132
Gene Name:
G protein-coupled receptor 132
Gene Aliases:
G2A
Chromosome Location:
Chr.14: 105049389 - 105065550 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05050520_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GPR132
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001278694.1 NP_001265623.1 5 1419 102
NM_001278695.1 NP_001265624.1 3 549 102
NM_001278696.1 NP_001265625.1 2 435 102
NM_013345.3 NP_037477.1 4 1329 102
GenBank mRNA AB465599.1 - 4 1209 102
AB465600.1 - 3 429 102
AF083955.1 - 4 1197 102
AK292142.1 - 5 1292 102
AK303654.1 - 3 429 102
AX775841.1 - 3 444 102
AX775935.1 - 3 429 102
AX775939.1 - 3 907 102
BC084546.1 - 2 884 5455247 102
DB137292.1 - 2 315 102
EU432121.1 - 2 297 102
EU543705.1 - 4 418 102

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AX775939.1, NM_001278696.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene GPR132

Panther Classification:

Molecular Function -

G-protein coupled receptor

Gene Ontology Categories:

Function(s) Process(es)

G1/S transition of mitotic cell cycle
G-protein coupled receptor signaling pathway
negative regulation of G2/M transition of mitotic cell cycle
G-protein coupled receptor activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1400010398.hg.1 ENST00000329797.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010398.hg.1 ENST00000329797.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010398.hg.1 ENST00000329797.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010398.hg.1 ENST00000329797.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010398.hg.1 ENST00000329797.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010398.hg.1 ENST00000329797.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010398.hg.1 ENST00000329797.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010398.hg.1 ENST00000329797.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010398.hg.1 ENST00000329797.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010398.hg.1 ENST00000329797.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010398.hg.1 ENST00000392585.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010398.hg.1 ENST00000392585.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010398.hg.1 ENST00000392585.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010398.hg.1 ENST00000392585.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010398.hg.1 ENST00000392585.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010398.hg.1 ENST00000392585.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010398.hg.1 ENST00000539291.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010398.hg.1 ENST00000539291.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010398.hg.1 ENST00000539291.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010398.hg.1 ENST00000539291.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010398.hg.1 ENST00000539291.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010398.hg.1 ENST00000539291.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010398.hg.1 ENST00000539291.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010398.hg.1 ENST00000539291.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010398.hg.1 ENST00000539291.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010398.hg.1 ENST00000539291.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010398.hg.1 ENST00000546679.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010398.hg.1 ENST00000546679.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010398.hg.1 ENST00000549990.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010398.hg.1 ENST00000549990.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010398.hg.1 ENST00000549990.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010398.hg.1 ENST00000549990.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010398.hg.1 ENST00000549990.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010398.hg.1 ENST00000549990.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010398.hg.1 ENST00000551869.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010398.hg.1 ENST00000551869.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010398.hg.1 ENST00000551869.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010398.hg.1 ENST00000551869.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010398.hg.1 ENST00000551869.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010398.hg.1 ENST00000551869.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010398.hg.1 ENST00000551869.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010398.hg.1 NM_001278694.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400010398.hg.1 NM_001278694.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400010398.hg.1 NM_001278694.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400010398.hg.1 NM_001278694.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400010398.hg.1 NM_001278694.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400010398.hg.1 NM_001278694.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400010398.hg.1 NM_001278694.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400010398.hg.1 NM_001278694.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400010398.hg.1 NM_001278694.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400010398.hg.1 NM_001278694.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400010398.hg.1 NM_001278694.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400010398.hg.1 NM_001278694.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400010398.hg.1 NM_001278695.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400010398.hg.1 NM_001278695.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400010398.hg.1 NM_001278695.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400010398.hg.1 NM_001278695.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400010398.hg.1 NM_001278695.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400010398.hg.1 NM_001278695.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400010398.hg.1 NM_001278695.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400010398.hg.1 NM_001278696.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400010398.hg.1 NM_001278696.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400010398.hg.1 NM_001278696.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400010398.hg.1 NM_001278696.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400010398.hg.1 NM_001278696.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400010398.hg.1 NM_001278696.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400010398.hg.1 NM_013345.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400010398.hg.1 NM_013345.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400010398.hg.1 NM_013345.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400010398.hg.1 NM_013345.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400010398.hg.1 NM_013345.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400010398.hg.1 NM_013345.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400010398.hg.1 NM_013345.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400010398.hg.1 NM_013345.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400010398.hg.1 NM_013345.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400010398.hg.1 NM_013345.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400010398.hg.1 NM_013345.3 Refseq
Affymetrix Human Clariom D Assay TC1400010398.hg.1 ENST00000329797.7 Ensembl
Affymetrix Human Clariom D Assay TC1400010398.hg.1 ENST00000329797.7 Ensembl
Affymetrix Human Clariom D Assay TC1400010398.hg.1 ENST00000329797.7 Ensembl
Affymetrix Human Clariom D Assay TC1400010398.hg.1 ENST00000329797.7 Ensembl
Affymetrix Human Clariom D Assay TC1400010398.hg.1 ENST00000329797.7 Ensembl
Affymetrix Human Clariom D Assay TC1400010398.hg.1 ENST00000329797.7 Ensembl
Affymetrix Human Clariom D Assay TC1400010398.hg.1 ENST00000329797.7 Ensembl
Affymetrix Human Clariom D Assay TC1400010398.hg.1 ENST00000329797.7 Ensembl
Affymetrix Human Clariom D Assay TC1400010398.hg.1 ENST00000329797.7 Ensembl
Affymetrix Human Clariom D Assay TC1400010398.hg.1 ENST00000329797.7 Ensembl
Affymetrix Human Clariom D Assay TC1400010398.hg.1 ENST00000392585.2 Ensembl
Affymetrix Human Clariom D Assay TC1400010398.hg.1 ENST00000392585.2 Ensembl
Affymetrix Human Clariom D Assay TC1400010398.hg.1 ENST00000392585.2 Ensembl
Affymetrix Human Clariom D Assay TC1400010398.hg.1 ENST00000392585.2 Ensembl
Affymetrix Human Clariom D Assay TC1400010398.hg.1 ENST00000392585.2 Ensembl
Affymetrix Human Clariom D Assay TC1400010398.hg.1 ENST00000392585.2 Ensembl
Affymetrix Human Clariom D Assay TC1400010398.hg.1 ENST00000539291.6 Ensembl
Affymetrix Human Clariom D Assay TC1400010398.hg.1 ENST00000539291.6 Ensembl
Affymetrix Human Clariom D Assay TC1400010398.hg.1 ENST00000539291.6 Ensembl
Affymetrix Human Clariom D Assay TC1400010398.hg.1 ENST00000539291.6 Ensembl
Affymetrix Human Clariom D Assay TC1400010398.hg.1 ENST00000539291.6 Ensembl
Affymetrix Human Clariom D Assay TC1400010398.hg.1 ENST00000539291.6 Ensembl
Affymetrix Human Clariom D Assay TC1400010398.hg.1 ENST00000539291.6 Ensembl
Affymetrix Human Clariom D Assay TC1400010398.hg.1 ENST00000539291.6 Ensembl
Affymetrix Human Clariom D Assay TC1400010398.hg.1 ENST00000539291.6 Ensembl
Affymetrix Human Clariom D Assay TC1400010398.hg.1 ENST00000539291.6 Ensembl
Affymetrix Human Clariom D Assay TC1400010398.hg.1 ENST00000546679.1 Ensembl
Affymetrix Human Clariom D Assay TC1400010398.hg.1 ENST00000546679.1 Ensembl
Affymetrix Human Clariom D Assay TC1400010398.hg.1 ENST00000549990.1 Ensembl
Affymetrix Human Clariom D Assay TC1400010398.hg.1 ENST00000549990.1 Ensembl
Affymetrix Human Clariom D Assay TC1400010398.hg.1 ENST00000549990.1 Ensembl
Affymetrix Human Clariom D Assay TC1400010398.hg.1 ENST00000549990.1 Ensembl
Affymetrix Human Clariom D Assay TC1400010398.hg.1 ENST00000549990.1 Ensembl
Affymetrix Human Clariom D Assay TC1400010398.hg.1 ENST00000549990.1 Ensembl
Affymetrix Human Clariom D Assay TC1400010398.hg.1 ENST00000551869.1 Ensembl
Affymetrix Human Clariom D Assay TC1400010398.hg.1 ENST00000551869.1 Ensembl
Affymetrix Human Clariom D Assay TC1400010398.hg.1 ENST00000551869.1 Ensembl
Affymetrix Human Clariom D Assay TC1400010398.hg.1 ENST00000551869.1 Ensembl
Affymetrix Human Clariom D Assay TC1400010398.hg.1 ENST00000551869.1 Ensembl
Affymetrix Human Clariom D Assay TC1400010398.hg.1 ENST00000551869.1 Ensembl
Affymetrix Human Clariom D Assay TC1400010398.hg.1 ENST00000551869.1 Ensembl
Affymetrix Human Clariom D Assay TC1400010398.hg.1 NM_001278694.1 Refseq
Affymetrix Human Clariom D Assay TC1400010398.hg.1 NM_001278694.1 Refseq
Affymetrix Human Clariom D Assay TC1400010398.hg.1 NM_001278694.1 Refseq
Affymetrix Human Clariom D Assay TC1400010398.hg.1 NM_001278694.1 Refseq
Affymetrix Human Clariom D Assay TC1400010398.hg.1 NM_001278694.1 Refseq
Affymetrix Human Clariom D Assay TC1400010398.hg.1 NM_001278694.1 Refseq
Affymetrix Human Clariom D Assay TC1400010398.hg.1 NM_001278694.1 Refseq
Affymetrix Human Clariom D Assay TC1400010398.hg.1 NM_001278694.1 Refseq
Affymetrix Human Clariom D Assay TC1400010398.hg.1 NM_001278694.1 Refseq
Affymetrix Human Clariom D Assay TC1400010398.hg.1 NM_001278694.1 Refseq
Affymetrix Human Clariom D Assay TC1400010398.hg.1 NM_001278694.1 Refseq
Affymetrix Human Clariom D Assay TC1400010398.hg.1 NM_001278694.1 Refseq
Affymetrix Human Clariom D Assay TC1400010398.hg.1 NM_001278695.1 Refseq
Affymetrix Human Clariom D Assay TC1400010398.hg.1 NM_001278695.1 Refseq
Affymetrix Human Clariom D Assay TC1400010398.hg.1 NM_001278695.1 Refseq
Affymetrix Human Clariom D Assay TC1400010398.hg.1 NM_001278695.1 Refseq
Affymetrix Human Clariom D Assay TC1400010398.hg.1 NM_001278695.1 Refseq
Affymetrix Human Clariom D Assay TC1400010398.hg.1 NM_001278695.1 Refseq
Affymetrix Human Clariom D Assay TC1400010398.hg.1 NM_001278695.1 Refseq
Affymetrix Human Clariom D Assay TC1400010398.hg.1 NM_001278696.1 Refseq
Affymetrix Human Clariom D Assay TC1400010398.hg.1 NM_001278696.1 Refseq
Affymetrix Human Clariom D Assay TC1400010398.hg.1 NM_001278696.1 Refseq
Affymetrix Human Clariom D Assay TC1400010398.hg.1 NM_001278696.1 Refseq
Affymetrix Human Clariom D Assay TC1400010398.hg.1 NM_001278696.1 Refseq
Affymetrix Human Clariom D Assay TC1400010398.hg.1 NM_001278696.1 Refseq
Affymetrix Human Clariom D Assay TC1400010398.hg.1 NM_013345.3 Refseq
Affymetrix Human Clariom D Assay TC1400010398.hg.1 NM_013345.3 Refseq
Affymetrix Human Clariom D Assay TC1400010398.hg.1 NM_013345.3 Refseq
Affymetrix Human Clariom D Assay TC1400010398.hg.1 NM_013345.3 Refseq
Affymetrix Human Clariom D Assay TC1400010398.hg.1 NM_013345.3 Refseq
Affymetrix Human Clariom D Assay TC1400010398.hg.1 NM_013345.3 Refseq
Affymetrix Human Clariom D Assay TC1400010398.hg.1 NM_013345.3 Refseq
Affymetrix Human Clariom D Assay TC1400010398.hg.1 NM_013345.3 Refseq
Affymetrix Human Clariom D Assay TC1400010398.hg.1 NM_013345.3 Refseq
Affymetrix Human Clariom D Assay TC1400010398.hg.1 NM_013345.3 Refseq
Affymetrix Human Clariom D Assay TC1400010398.hg.1 NM_013345.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0