This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05050519_g1
See other GPR68 GE Assays ›
Gene Symbol:
GPR68
Gene Name:
G protein-coupled receptor 68
Gene Aliases:
GPR12A, OGR1
Chromosome Location:
Chr.14: 91232532 - 91264581 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05050519_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GPR68
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_003485.3 NP_003476.3 1 125 62
GenBank mRNA BC098567.1 - 1 68 30925589 62
BM723705.1 - 1 170 62
CN287036.1 - 1 125 62

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the BC098567.1, NM_003485.3 transcripts.

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 G-protein coupled receptor Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene GPR68

Panther Classification:

Molecular Function -

G-protein coupled receptor

Gene Ontology Categories:

Function(s) Process(es)

inflammatory response
G-protein coupled receptor signaling pathway
positive regulation of insulin secretion involved in cellular response to glucose stimulus
negative regulation of monocyte differentiation
cellular response to pH
positive regulation of osteoclast development
G-protein coupled receptor activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1400009978.hg.1 ENST00000529102.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009978.hg.1 ENST00000529102.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009978.hg.1 ENST00000529102.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009978.hg.1 ENST00000529102.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009978.hg.1 ENST00000529300.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009978.hg.1 ENST00000529300.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009978.hg.1 ENST00000529300.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009978.hg.1 ENST00000529300.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009978.hg.1 ENST00000531499.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009978.hg.1 ENST00000531499.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009978.hg.1 ENST00000531499.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009978.hg.1 ENST00000531499.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009978.hg.1 ENST00000531499.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009978.hg.1 ENST00000531499.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009978.hg.1 ENST00000531499.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009978.hg.1 ENST00000531499.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009978.hg.1 ENST00000531499.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009978.hg.1 ENST00000535815.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009978.hg.1 ENST00000535815.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009978.hg.1 ENST00000535815.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009978.hg.1 ENST00000535815.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009978.hg.1 ENST00000535815.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009978.hg.1 ENST00000535815.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009978.hg.1 ENST00000535815.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400009978.hg.1 NM_001177676.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009978.hg.1 NM_001177676.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009978.hg.1 NM_001177676.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009978.hg.1 NM_001177676.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009978.hg.1 NM_001177676.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009978.hg.1 NM_001177676.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009978.hg.1 NM_001177676.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009978.hg.1 NM_003485.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009978.hg.1 NM_003485.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009978.hg.1 NM_003485.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009978.hg.1 NM_003485.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009978.hg.1 NM_003485.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009978.hg.1 NM_003485.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009978.hg.1 NM_003485.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009978.hg.1 NM_003485.3 Refseq
Affymetrix Human Clariom D Assay TC1400009978.hg.1 ENST00000529102.1 Ensembl
Affymetrix Human Clariom D Assay TC1400009978.hg.1 ENST00000529102.1 Ensembl
Affymetrix Human Clariom D Assay TC1400009978.hg.1 ENST00000529102.1 Ensembl
Affymetrix Human Clariom D Assay TC1400009978.hg.1 ENST00000529102.1 Ensembl
Affymetrix Human Clariom D Assay TC1400009978.hg.1 ENST00000529300.1 Ensembl
Affymetrix Human Clariom D Assay TC1400009978.hg.1 ENST00000529300.1 Ensembl
Affymetrix Human Clariom D Assay TC1400009978.hg.1 ENST00000529300.1 Ensembl
Affymetrix Human Clariom D Assay TC1400009978.hg.1 ENST00000529300.1 Ensembl
Affymetrix Human Clariom D Assay TC1400009978.hg.1 ENST00000531499.2 Ensembl
Affymetrix Human Clariom D Assay TC1400009978.hg.1 ENST00000531499.2 Ensembl
Affymetrix Human Clariom D Assay TC1400009978.hg.1 ENST00000531499.2 Ensembl
Affymetrix Human Clariom D Assay TC1400009978.hg.1 ENST00000531499.2 Ensembl
Affymetrix Human Clariom D Assay TC1400009978.hg.1 ENST00000531499.2 Ensembl
Affymetrix Human Clariom D Assay TC1400009978.hg.1 ENST00000531499.2 Ensembl
Affymetrix Human Clariom D Assay TC1400009978.hg.1 ENST00000531499.2 Ensembl
Affymetrix Human Clariom D Assay TC1400009978.hg.1 ENST00000531499.2 Ensembl
Affymetrix Human Clariom D Assay TC1400009978.hg.1 ENST00000531499.2 Ensembl
Affymetrix Human Clariom D Assay TC1400009978.hg.1 ENST00000535815.5 Ensembl
Affymetrix Human Clariom D Assay TC1400009978.hg.1 ENST00000535815.5 Ensembl
Affymetrix Human Clariom D Assay TC1400009978.hg.1 ENST00000535815.5 Ensembl
Affymetrix Human Clariom D Assay TC1400009978.hg.1 ENST00000535815.5 Ensembl
Affymetrix Human Clariom D Assay TC1400009978.hg.1 ENST00000535815.5 Ensembl
Affymetrix Human Clariom D Assay TC1400009978.hg.1 ENST00000535815.5 Ensembl
Affymetrix Human Clariom D Assay TC1400009978.hg.1 ENST00000535815.5 Ensembl
Affymetrix Human Clariom D Assay TC1400009978.hg.1 NM_001177676.1 Refseq
Affymetrix Human Clariom D Assay TC1400009978.hg.1 NM_001177676.1 Refseq
Affymetrix Human Clariom D Assay TC1400009978.hg.1 NM_001177676.1 Refseq
Affymetrix Human Clariom D Assay TC1400009978.hg.1 NM_001177676.1 Refseq
Affymetrix Human Clariom D Assay TC1400009978.hg.1 NM_001177676.1 Refseq
Affymetrix Human Clariom D Assay TC1400009978.hg.1 NM_001177676.1 Refseq
Affymetrix Human Clariom D Assay TC1400009978.hg.1 NM_001177676.1 Refseq
Affymetrix Human Clariom D Assay TC1400009978.hg.1 NM_003485.3 Refseq
Affymetrix Human Clariom D Assay TC1400009978.hg.1 NM_003485.3 Refseq
Affymetrix Human Clariom D Assay TC1400009978.hg.1 NM_003485.3 Refseq
Affymetrix Human Clariom D Assay TC1400009978.hg.1 NM_003485.3 Refseq
Affymetrix Human Clariom D Assay TC1400009978.hg.1 NM_003485.3 Refseq
Affymetrix Human Clariom D Assay TC1400009978.hg.1 NM_003485.3 Refseq
Affymetrix Human Clariom D Assay TC1400009978.hg.1 NM_003485.3 Refseq
Affymetrix Human Clariom D Assay TC1400009978.hg.1 NM_003485.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0