This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05050189_s1
See other CDC42BPB GE Assays ›
Gene Symbol:
CDC42BPB
Gene Name:
CDC42 binding protein kinase beta
Gene Aliases:
MRCKB
Chromosome Location:
Chr.14: 102932379 - 103057405 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05050189_s1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CDC42BPB
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK126684.1 - 1 3447 93

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1400010691.hg.1 ENST00000560931.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400010691.hg.1 ENST00000560931.1 Ensembl
Affymetrix Human Clariom D Assay TC1400010691.hg.1 ENST00000560931.1 Ensembl
Affymetrix Human Clariom D Assay TC1400010691.hg.1 ENST00000560931.1 Ensembl

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-56ffd697dc-88d4p:80/100.66.130.94:80.
git-commit: 082a6d415be982b1f92c35e55dd71609d4576d3e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0