This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05049938_s1
See other LATS2 GE Assays ›
Gene Symbol:
LATS2
Gene Name:
large tumor suppressor kinase 2
Gene Aliases:
KPM
Chromosome Location:
Chr.13: 20973036 - 21061583 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05049938_s1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LATS2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_014572.2 NP_055387.2 2 724 82
XM_005266342.1 XP_005266399.1 2 762 82
XM_017020542.1 XP_016876031.1 2 690 82
GenBank mRNA AB028019.1 - 1 161 82
AB209897.1 - 2 667 82
AF207547.1 - 2 656 82
AK314235.1 - 2 647 82
BC071572.1 - 2 678 82
BC127671.1 - 2 650 82

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Inventoried Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene LATS2

Panther Classification:

Molecular Function -

non-receptor serine/threonine protein kinase

Gene Ontology Categories:

Function(s) Process(es)

G1/S transition of mitotic cell cycle
inner cell mass cell fate commitment
inner cell mass cellular morphogenesis
protein phosphorylation
mitotic nuclear division
hormone-mediated signaling pathway
peptidyl-serine phosphorylation
keratinocyte differentiation
cellular protein localization
hippo signaling
intracellular signal transduction
positive regulation of apoptotic process
negative regulation of cyclin-dependent protein serine/threonine kinase activity
regulation of organ growth
cell division
negative regulation of canonical Wnt signaling pathway
protein serine/threonine kinase activity
protein binding
ATP binding
metal ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-655dcf6cc8-78hlp:80/100.66.128.142:80.
git-commit: 0d764ad91c70359c551b4d51a80efc6847a0ee54
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0