This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05048862_s1
See other NAB2 GE Assays ›
Gene Symbol:
NAB2
Gene Name:
NGFI-A binding protein 2
Gene Aliases:
MADER
Chromosome Location:
Chr.12: 57088894 - 57095476 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05048862_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NAB2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_005967.3 NP_005958.1 2 984 87
XM_005268894.2 XP_005268951.1 2 984 87
GenBank mRNA AK314229.1 - 2 764 87
BC065931.1 - 2 771 6157017 87
U48361.1 - 2 763 87
X70991.1 - 1 459 87

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene NAB2

Gene Ontology Categories:

Function(s) Process(es)

endochondral ossification
transcription, DNA-templated
nervous system development
cell proliferation
Schwann cell differentiation
negative regulation of transcription from RNA polymerase III promoter
myelination
regulation of epidermis development
transcription corepressor activity
protein binding
transcription factor binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1200007859.hg.1 ENST00000300131.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007859.hg.1 ENST00000300131.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007859.hg.1 ENST00000300131.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007859.hg.1 ENST00000300131.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007859.hg.1 ENST00000300131.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007859.hg.1 ENST00000300131.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007859.hg.1 ENST00000300131.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007859.hg.1 ENST00000300131.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007859.hg.1 ENST00000300131.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007859.hg.1 ENST00000300131.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007859.hg.1 ENST00000300131.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007859.hg.1 ENST00000300131.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007859.hg.1 ENST00000300131.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007859.hg.1 ENST00000300131.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007859.hg.1 ENST00000300131.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007859.hg.1 ENST00000300131.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007859.hg.1 ENST00000300131.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007859.hg.1 ENST00000342556.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007859.hg.1 ENST00000342556.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007859.hg.1 ENST00000342556.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007859.hg.1 ENST00000342556.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007859.hg.1 ENST00000342556.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007859.hg.1 ENST00000342556.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007859.hg.1 ENST00000342556.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007859.hg.1 ENST00000342556.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007859.hg.1 ENST00000342556.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007859.hg.1 ENST00000342556.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007859.hg.1 ENST00000342556.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007859.hg.1 ENST00000342556.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007859.hg.1 ENST00000342556.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007859.hg.1 ENST00000342556.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007859.hg.1 ENST00000554718.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007859.hg.1 ENST00000554718.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007859.hg.1 ENST00000554718.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007859.hg.1 ENST00000554718.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007859.hg.1 ENST00000554839.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007859.hg.1 ENST00000554839.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007859.hg.1 ENST00000554839.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007859.hg.1 ENST00000554839.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007859.hg.1 ENST00000555857.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007859.hg.1 ENST00000555857.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007859.hg.1 ENST00000555857.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007859.hg.1 ENST00000555857.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007859.hg.1 ENST00000555857.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007859.hg.1 NM_005967.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200007859.hg.1 NM_005967.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200007859.hg.1 NM_005967.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200007859.hg.1 NM_005967.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200007859.hg.1 NM_005967.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200007859.hg.1 NM_005967.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200007859.hg.1 NM_005967.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200007859.hg.1 NM_005967.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200007859.hg.1 NM_005967.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200007859.hg.1 NM_005967.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200007859.hg.1 NM_005967.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200007859.hg.1 NM_005967.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200007859.hg.1 NM_005967.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200007859.hg.1 NM_005967.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200007859.hg.1 NM_005967.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200007859.hg.1 NM_005967.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200007859.hg.1 NM_005967.3 Refseq
Affymetrix Human Clariom D Assay TC1200007859.hg.1 ENST00000300131.7 Ensembl
Affymetrix Human Clariom D Assay TC1200007859.hg.1 ENST00000300131.7 Ensembl
Affymetrix Human Clariom D Assay TC1200007859.hg.1 ENST00000300131.7 Ensembl
Affymetrix Human Clariom D Assay TC1200007859.hg.1 ENST00000300131.7 Ensembl
Affymetrix Human Clariom D Assay TC1200007859.hg.1 ENST00000300131.7 Ensembl
Affymetrix Human Clariom D Assay TC1200007859.hg.1 ENST00000300131.7 Ensembl
Affymetrix Human Clariom D Assay TC1200007859.hg.1 ENST00000300131.7 Ensembl
Affymetrix Human Clariom D Assay TC1200007859.hg.1 ENST00000300131.7 Ensembl
Affymetrix Human Clariom D Assay TC1200007859.hg.1 ENST00000300131.7 Ensembl
Affymetrix Human Clariom D Assay TC1200007859.hg.1 ENST00000300131.7 Ensembl
Affymetrix Human Clariom D Assay TC1200007859.hg.1 ENST00000300131.7 Ensembl
Affymetrix Human Clariom D Assay TC1200007859.hg.1 ENST00000300131.7 Ensembl
Affymetrix Human Clariom D Assay TC1200007859.hg.1 ENST00000300131.7 Ensembl
Affymetrix Human Clariom D Assay TC1200007859.hg.1 ENST00000300131.7 Ensembl
Affymetrix Human Clariom D Assay TC1200007859.hg.1 ENST00000300131.7 Ensembl
Affymetrix Human Clariom D Assay TC1200007859.hg.1 ENST00000300131.7 Ensembl
Affymetrix Human Clariom D Assay TC1200007859.hg.1 ENST00000300131.7 Ensembl
Affymetrix Human Clariom D Assay TC1200007859.hg.1 ENST00000342556.6 Ensembl
Affymetrix Human Clariom D Assay TC1200007859.hg.1 ENST00000342556.6 Ensembl
Affymetrix Human Clariom D Assay TC1200007859.hg.1 ENST00000342556.6 Ensembl
Affymetrix Human Clariom D Assay TC1200007859.hg.1 ENST00000342556.6 Ensembl
Affymetrix Human Clariom D Assay TC1200007859.hg.1 ENST00000342556.6 Ensembl
Affymetrix Human Clariom D Assay TC1200007859.hg.1 ENST00000342556.6 Ensembl
Affymetrix Human Clariom D Assay TC1200007859.hg.1 ENST00000342556.6 Ensembl
Affymetrix Human Clariom D Assay TC1200007859.hg.1 ENST00000342556.6 Ensembl
Affymetrix Human Clariom D Assay TC1200007859.hg.1 ENST00000342556.6 Ensembl
Affymetrix Human Clariom D Assay TC1200007859.hg.1 ENST00000342556.6 Ensembl
Affymetrix Human Clariom D Assay TC1200007859.hg.1 ENST00000342556.6 Ensembl
Affymetrix Human Clariom D Assay TC1200007859.hg.1 ENST00000342556.6 Ensembl
Affymetrix Human Clariom D Assay TC1200007859.hg.1 ENST00000342556.6 Ensembl
Affymetrix Human Clariom D Assay TC1200007859.hg.1 ENST00000342556.6 Ensembl
Affymetrix Human Clariom D Assay TC1200007859.hg.1 ENST00000554718.1 Ensembl
Affymetrix Human Clariom D Assay TC1200007859.hg.1 ENST00000554718.1 Ensembl
Affymetrix Human Clariom D Assay TC1200007859.hg.1 ENST00000554718.1 Ensembl
Affymetrix Human Clariom D Assay TC1200007859.hg.1 ENST00000554718.1 Ensembl
Affymetrix Human Clariom D Assay TC1200007859.hg.1 ENST00000554839.1 Ensembl
Affymetrix Human Clariom D Assay TC1200007859.hg.1 ENST00000554839.1 Ensembl
Affymetrix Human Clariom D Assay TC1200007859.hg.1 ENST00000554839.1 Ensembl
Affymetrix Human Clariom D Assay TC1200007859.hg.1 ENST00000554839.1 Ensembl
Affymetrix Human Clariom D Assay TC1200007859.hg.1 ENST00000555857.1 Ensembl
Affymetrix Human Clariom D Assay TC1200007859.hg.1 ENST00000555857.1 Ensembl
Affymetrix Human Clariom D Assay TC1200007859.hg.1 ENST00000555857.1 Ensembl
Affymetrix Human Clariom D Assay TC1200007859.hg.1 ENST00000555857.1 Ensembl
Affymetrix Human Clariom D Assay TC1200007859.hg.1 ENST00000555857.1 Ensembl
Affymetrix Human Clariom D Assay TC1200007859.hg.1 NM_005967.3 Refseq
Affymetrix Human Clariom D Assay TC1200007859.hg.1 NM_005967.3 Refseq
Affymetrix Human Clariom D Assay TC1200007859.hg.1 NM_005967.3 Refseq
Affymetrix Human Clariom D Assay TC1200007859.hg.1 NM_005967.3 Refseq
Affymetrix Human Clariom D Assay TC1200007859.hg.1 NM_005967.3 Refseq
Affymetrix Human Clariom D Assay TC1200007859.hg.1 NM_005967.3 Refseq
Affymetrix Human Clariom D Assay TC1200007859.hg.1 NM_005967.3 Refseq
Affymetrix Human Clariom D Assay TC1200007859.hg.1 NM_005967.3 Refseq
Affymetrix Human Clariom D Assay TC1200007859.hg.1 NM_005967.3 Refseq
Affymetrix Human Clariom D Assay TC1200007859.hg.1 NM_005967.3 Refseq
Affymetrix Human Clariom D Assay TC1200007859.hg.1 NM_005967.3 Refseq
Affymetrix Human Clariom D Assay TC1200007859.hg.1 NM_005967.3 Refseq
Affymetrix Human Clariom D Assay TC1200007859.hg.1 NM_005967.3 Refseq
Affymetrix Human Clariom D Assay TC1200007859.hg.1 NM_005967.3 Refseq
Affymetrix Human Clariom D Assay TC1200007859.hg.1 NM_005967.3 Refseq
Affymetrix Human Clariom D Assay TC1200007859.hg.1 NM_005967.3 Refseq
Affymetrix Human Clariom D Assay TC1200007859.hg.1 NM_005967.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0