This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05047515_s1
See other LY6K GE Assays ›
Gene Symbol:
LY6K
Gene Name:
lymphocyte antigen 6 complex, locus K
Gene Aliases:
CT97, HSJ001348, URLC10, ly-6K
Chromosome Location:
Chr.8: 142699791 - 142704170 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05047515_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LY6K
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK092545.1 - 2 313 80

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0800009157.hg.1 ENST00000292430.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009157.hg.1 ENST00000292430.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009157.hg.1 ENST00000292430.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009157.hg.1 ENST00000292430.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009157.hg.1 ENST00000292430.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009157.hg.1 ENST00000292430.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009157.hg.1 ENST00000292430.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009157.hg.1 ENST00000292430.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009157.hg.1 ENST00000292430.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009157.hg.1 ENST00000292430.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009157.hg.1 ENST00000292430.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009157.hg.1 ENST00000292430.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009157.hg.1 ENST00000518841.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009157.hg.1 ENST00000518841.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009157.hg.1 ENST00000518841.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009157.hg.1 ENST00000518841.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009157.hg.1 ENST00000518841.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009157.hg.1 ENST00000518841.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009157.hg.1 ENST00000519387.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009157.hg.1 ENST00000519387.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009157.hg.1 ENST00000519387.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009157.hg.1 ENST00000519387.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009157.hg.1 ENST00000519387.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009157.hg.1 ENST00000519387.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009157.hg.1 ENST00000519387.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009157.hg.1 ENST00000519387.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009157.hg.1 ENST00000519387.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009157.hg.1 ENST00000519390.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009157.hg.1 ENST00000519390.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009157.hg.1 ENST00000519390.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009157.hg.1 ENST00000519390.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009157.hg.1 ENST00000519390.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009157.hg.1 ENST00000519390.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009157.hg.1 ENST00000519390.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009157.hg.1 ENST00000522591.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009157.hg.1 ENST00000522591.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009157.hg.1 ENST00000522591.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009157.hg.1 ENST00000522591.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009157.hg.1 ENST00000522591.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009157.hg.1 ENST00000522591.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009157.hg.1 ENST00000522591.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009157.hg.1 ENST00000522591.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009157.hg.1 ENST00000522591.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009157.hg.1 ENST00000522591.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009157.hg.1 ENST00000522591.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009157.hg.1 ENST00000522591.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009157.hg.1 ENST00000522591.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009157.hg.1 NM_001160354.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800009157.hg.1 NM_001160354.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800009157.hg.1 NM_001160354.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800009157.hg.1 NM_001160354.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800009157.hg.1 NM_001160354.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800009157.hg.1 NM_001160354.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800009157.hg.1 NM_001160354.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800009157.hg.1 NM_001160354.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800009157.hg.1 NM_001160354.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800009157.hg.1 NM_001160354.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800009157.hg.1 NM_001160354.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800009157.hg.1 NM_001160354.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800009157.hg.1 NM_001160355.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800009157.hg.1 NM_001160355.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800009157.hg.1 NM_001160355.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800009157.hg.1 NM_001160355.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800009157.hg.1 NM_001160355.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800009157.hg.1 NM_001160355.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800009157.hg.1 NM_001160355.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800009157.hg.1 NM_001160355.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800009157.hg.1 NM_001160355.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800009157.hg.1 NM_001160355.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800009157.hg.1 NM_017527.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800009157.hg.1 NM_017527.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800009157.hg.1 NM_017527.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800009157.hg.1 NM_017527.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800009157.hg.1 NM_017527.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800009157.hg.1 NM_017527.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800009157.hg.1 NM_017527.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800009157.hg.1 NM_017527.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800009157.hg.1 NM_017527.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800009157.hg.1 NM_017527.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800009157.hg.1 NM_017527.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800009005.hg.1 ENST00000577345.1 Ensembl
Affymetrix Human Clariom D Assay TC0800009005.hg.1 ENST00000577345.1 Ensembl
Affymetrix Human Clariom D Assay TC0800009157.hg.1 ENST00000292430.10 Ensembl
Affymetrix Human Clariom D Assay TC0800009157.hg.1 ENST00000292430.10 Ensembl
Affymetrix Human Clariom D Assay TC0800009157.hg.1 ENST00000292430.10 Ensembl
Affymetrix Human Clariom D Assay TC0800009157.hg.1 ENST00000292430.10 Ensembl
Affymetrix Human Clariom D Assay TC0800009157.hg.1 ENST00000292430.10 Ensembl
Affymetrix Human Clariom D Assay TC0800009157.hg.1 ENST00000292430.10 Ensembl
Affymetrix Human Clariom D Assay TC0800009157.hg.1 ENST00000292430.10 Ensembl
Affymetrix Human Clariom D Assay TC0800009157.hg.1 ENST00000292430.10 Ensembl
Affymetrix Human Clariom D Assay TC0800009157.hg.1 ENST00000292430.10 Ensembl
Affymetrix Human Clariom D Assay TC0800009157.hg.1 ENST00000292430.10 Ensembl
Affymetrix Human Clariom D Assay TC0800009157.hg.1 ENST00000292430.10 Ensembl
Affymetrix Human Clariom D Assay TC0800009157.hg.1 ENST00000292430.10 Ensembl
Affymetrix Human Clariom D Assay TC0800009157.hg.1 ENST00000518841.5 Ensembl
Affymetrix Human Clariom D Assay TC0800009157.hg.1 ENST00000518841.5 Ensembl
Affymetrix Human Clariom D Assay TC0800009157.hg.1 ENST00000518841.5 Ensembl
Affymetrix Human Clariom D Assay TC0800009157.hg.1 ENST00000518841.5 Ensembl
Affymetrix Human Clariom D Assay TC0800009157.hg.1 ENST00000518841.5 Ensembl
Affymetrix Human Clariom D Assay TC0800009157.hg.1 ENST00000518841.5 Ensembl
Affymetrix Human Clariom D Assay TC0800009157.hg.1 ENST00000519387.1 Ensembl
Affymetrix Human Clariom D Assay TC0800009157.hg.1 ENST00000519387.1 Ensembl
Affymetrix Human Clariom D Assay TC0800009157.hg.1 ENST00000519387.1 Ensembl
Affymetrix Human Clariom D Assay TC0800009157.hg.1 ENST00000519387.1 Ensembl
Affymetrix Human Clariom D Assay TC0800009157.hg.1 ENST00000519387.1 Ensembl
Affymetrix Human Clariom D Assay TC0800009157.hg.1 ENST00000519387.1 Ensembl
Affymetrix Human Clariom D Assay TC0800009157.hg.1 ENST00000519387.1 Ensembl
Affymetrix Human Clariom D Assay TC0800009157.hg.1 ENST00000519387.1 Ensembl
Affymetrix Human Clariom D Assay TC0800009157.hg.1 ENST00000519387.1 Ensembl
Affymetrix Human Clariom D Assay TC0800009157.hg.1 ENST00000519390.1 Ensembl
Affymetrix Human Clariom D Assay TC0800009157.hg.1 ENST00000519390.1 Ensembl
Affymetrix Human Clariom D Assay TC0800009157.hg.1 ENST00000519390.1 Ensembl
Affymetrix Human Clariom D Assay TC0800009157.hg.1 ENST00000519390.1 Ensembl
Affymetrix Human Clariom D Assay TC0800009157.hg.1 ENST00000519390.1 Ensembl
Affymetrix Human Clariom D Assay TC0800009157.hg.1 ENST00000519390.1 Ensembl
Affymetrix Human Clariom D Assay TC0800009157.hg.1 ENST00000519390.1 Ensembl
Affymetrix Human Clariom D Assay TC0800009157.hg.1 ENST00000522591.1 Ensembl
Affymetrix Human Clariom D Assay TC0800009157.hg.1 ENST00000522591.1 Ensembl
Affymetrix Human Clariom D Assay TC0800009157.hg.1 ENST00000522591.1 Ensembl
Affymetrix Human Clariom D Assay TC0800009157.hg.1 ENST00000522591.1 Ensembl
Affymetrix Human Clariom D Assay TC0800009157.hg.1 ENST00000522591.1 Ensembl
Affymetrix Human Clariom D Assay TC0800009157.hg.1 ENST00000522591.1 Ensembl
Affymetrix Human Clariom D Assay TC0800009157.hg.1 ENST00000522591.1 Ensembl
Affymetrix Human Clariom D Assay TC0800009157.hg.1 ENST00000522591.1 Ensembl
Affymetrix Human Clariom D Assay TC0800009157.hg.1 ENST00000522591.1 Ensembl
Affymetrix Human Clariom D Assay TC0800009157.hg.1 ENST00000522591.1 Ensembl
Affymetrix Human Clariom D Assay TC0800009157.hg.1 ENST00000522591.1 Ensembl
Affymetrix Human Clariom D Assay TC0800009157.hg.1 ENST00000522591.1 Ensembl
Affymetrix Human Clariom D Assay TC0800009157.hg.1 ENST00000522591.1 Ensembl
Affymetrix Human Clariom D Assay TC0800009157.hg.1 NM_001160354.1 Refseq
Affymetrix Human Clariom D Assay TC0800009157.hg.1 NM_001160354.1 Refseq
Affymetrix Human Clariom D Assay TC0800009157.hg.1 NM_001160354.1 Refseq
Affymetrix Human Clariom D Assay TC0800009157.hg.1 NM_001160354.1 Refseq
Affymetrix Human Clariom D Assay TC0800009157.hg.1 NM_001160354.1 Refseq
Affymetrix Human Clariom D Assay TC0800009157.hg.1 NM_001160354.1 Refseq
Affymetrix Human Clariom D Assay TC0800009157.hg.1 NM_001160354.1 Refseq
Affymetrix Human Clariom D Assay TC0800009157.hg.1 NM_001160354.1 Refseq
Affymetrix Human Clariom D Assay TC0800009157.hg.1 NM_001160354.1 Refseq
Affymetrix Human Clariom D Assay TC0800009157.hg.1 NM_001160354.1 Refseq
Affymetrix Human Clariom D Assay TC0800009157.hg.1 NM_001160354.1 Refseq
Affymetrix Human Clariom D Assay TC0800009157.hg.1 NM_001160354.1 Refseq
Affymetrix Human Clariom D Assay TC0800009157.hg.1 NM_001160355.1 Refseq
Affymetrix Human Clariom D Assay TC0800009157.hg.1 NM_001160355.1 Refseq
Affymetrix Human Clariom D Assay TC0800009157.hg.1 NM_001160355.1 Refseq
Affymetrix Human Clariom D Assay TC0800009157.hg.1 NM_001160355.1 Refseq
Affymetrix Human Clariom D Assay TC0800009157.hg.1 NM_001160355.1 Refseq
Affymetrix Human Clariom D Assay TC0800009157.hg.1 NM_001160355.1 Refseq
Affymetrix Human Clariom D Assay TC0800009157.hg.1 NM_001160355.1 Refseq
Affymetrix Human Clariom D Assay TC0800009157.hg.1 NM_001160355.1 Refseq
Affymetrix Human Clariom D Assay TC0800009157.hg.1 NM_001160355.1 Refseq
Affymetrix Human Clariom D Assay TC0800009157.hg.1 NM_001160355.1 Refseq
Affymetrix Human Clariom D Assay TC0800009157.hg.1 NM_017527.3 Refseq
Affymetrix Human Clariom D Assay TC0800009157.hg.1 NM_017527.3 Refseq
Affymetrix Human Clariom D Assay TC0800009157.hg.1 NM_017527.3 Refseq
Affymetrix Human Clariom D Assay TC0800009157.hg.1 NM_017527.3 Refseq
Affymetrix Human Clariom D Assay TC0800009157.hg.1 NM_017527.3 Refseq
Affymetrix Human Clariom D Assay TC0800009157.hg.1 NM_017527.3 Refseq
Affymetrix Human Clariom D Assay TC0800009157.hg.1 NM_017527.3 Refseq
Affymetrix Human Clariom D Assay TC0800009157.hg.1 NM_017527.3 Refseq
Affymetrix Human Clariom D Assay TC0800009157.hg.1 NM_017527.3 Refseq
Affymetrix Human Clariom D Assay TC0800009157.hg.1 NM_017527.3 Refseq
Affymetrix Human Clariom D Assay TC0800009157.hg.1 NM_017527.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0