This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05045898_s1
See other OPN5 GE Assays ›
Gene Symbol:
OPN5
Gene Name:
opsin 5
Gene Aliases:
GPR136, GRP136, PGR12, TMEM13
Chromosome Location:
Chr.6: 47782037 - 47831088 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05045898_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: OPN5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_017010409.1 XP_016865898.1 1 1172 82
XM_017010414.1 XP_016865903.1 1 1177 82
GenBank mRNA AK097448.1 - 1 82

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_017010409.1, XM_017010414.1 transcripts.

Set Membership:

Amplicon length between 71 and 85 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

G-protein coupled receptor

Gene Ontology Categories:

Function(s) Process(es)

G-protein coupled receptor signaling pathway
visual perception
phototransduction
detection of visible light
protein-chromophore linkage
cellular response to light stimulus
11-cis retinal binding
G-protein coupled photoreceptor activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0600008190.hg.1 ENST00000244799.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008190.hg.1 ENST00000244799.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008190.hg.1 ENST00000244799.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008190.hg.1 ENST00000244799.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008190.hg.1 ENST00000244799.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008190.hg.1 ENST00000244799.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008190.hg.1 ENST00000244799.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008190.hg.1 ENST00000244799.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008190.hg.1 ENST00000244799.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008190.hg.1 ENST00000244799.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008190.hg.1 ENST00000244799.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008190.hg.1 ENST00000244799.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008190.hg.1 ENST00000244799.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008190.hg.1 ENST00000371211.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008190.hg.1 ENST00000371211.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008190.hg.1 ENST00000371211.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008190.hg.1 ENST00000371211.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008190.hg.1 ENST00000371211.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008190.hg.1 ENST00000371211.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008190.hg.1 ENST00000371211.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008190.hg.1 ENST00000371211.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008190.hg.1 ENST00000371211.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008190.hg.1 ENST00000371211.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008190.hg.1 ENST00000371211.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008190.hg.1 ENST00000371211.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008190.hg.1 ENST00000371211.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008190.hg.1 ENST00000393699.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008190.hg.1 ENST00000393699.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008190.hg.1 ENST00000393699.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008190.hg.1 ENST00000393699.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008190.hg.1 ENST00000393699.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008190.hg.1 ENST00000393699.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008190.hg.1 ENST00000393699.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008190.hg.1 ENST00000393699.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008190.hg.1 ENST00000393699.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008190.hg.1 ENST00000393699.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008190.hg.1 ENST00000393699.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008190.hg.1 ENST00000489301.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008190.hg.1 ENST00000489301.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008190.hg.1 ENST00000489301.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008190.hg.1 ENST00000489301.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008190.hg.1 ENST00000489301.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008190.hg.1 ENST00000489301.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008190.hg.1 ENST00000489301.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008190.hg.1 ENST00000489301.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008190.hg.1 ENST00000489301.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008190.hg.1 ENST00000489301.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008190.hg.1 ENST00000489301.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008190.hg.1 ENST00000489301.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008190.hg.1 ENST00000489301.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008190.hg.1 ENST00000510695.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008190.hg.1 ENST00000510695.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008190.hg.1 ENST00000510695.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008190.hg.1 ENST00000510695.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008190.hg.1 NM_181744.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008190.hg.1 NM_181744.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008190.hg.1 NM_181744.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008190.hg.1 NM_181744.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008190.hg.1 NM_181744.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008190.hg.1 NM_181744.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008190.hg.1 NM_181744.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008190.hg.1 NM_181744.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008190.hg.1 NM_181744.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008190.hg.1 NM_181744.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008190.hg.1 NM_181744.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008190.hg.1 NM_181744.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008190.hg.1 NM_181744.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008190.hg.1 NM_181744.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008190.hg.1 NR_033806.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008190.hg.1 NR_033806.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008190.hg.1 NR_033806.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008190.hg.1 NR_033806.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008190.hg.1 NR_033806.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008190.hg.1 NR_033806.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008190.hg.1 NR_033806.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008190.hg.1 NR_033806.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008190.hg.1 NR_033806.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008190.hg.1 NR_033806.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008190.hg.1 NR_033806.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008190.hg.1 NR_033806.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008190.hg.1 NR_033806.1 Refseq
Affymetrix Human Clariom D Assay TC0600008190.hg.1 ENST00000244799.4 Ensembl
Affymetrix Human Clariom D Assay TC0600008190.hg.1 ENST00000244799.4 Ensembl
Affymetrix Human Clariom D Assay TC0600008190.hg.1 ENST00000244799.4 Ensembl
Affymetrix Human Clariom D Assay TC0600008190.hg.1 ENST00000244799.4 Ensembl
Affymetrix Human Clariom D Assay TC0600008190.hg.1 ENST00000244799.4 Ensembl
Affymetrix Human Clariom D Assay TC0600008190.hg.1 ENST00000244799.4 Ensembl
Affymetrix Human Clariom D Assay TC0600008190.hg.1 ENST00000244799.4 Ensembl
Affymetrix Human Clariom D Assay TC0600008190.hg.1 ENST00000244799.4 Ensembl
Affymetrix Human Clariom D Assay TC0600008190.hg.1 ENST00000244799.4 Ensembl
Affymetrix Human Clariom D Assay TC0600008190.hg.1 ENST00000244799.4 Ensembl
Affymetrix Human Clariom D Assay TC0600008190.hg.1 ENST00000244799.4 Ensembl
Affymetrix Human Clariom D Assay TC0600008190.hg.1 ENST00000244799.4 Ensembl
Affymetrix Human Clariom D Assay TC0600008190.hg.1 ENST00000244799.4 Ensembl
Affymetrix Human Clariom D Assay TC0600008190.hg.1 ENST00000371211.6 Ensembl
Affymetrix Human Clariom D Assay TC0600008190.hg.1 ENST00000371211.6 Ensembl
Affymetrix Human Clariom D Assay TC0600008190.hg.1 ENST00000371211.6 Ensembl
Affymetrix Human Clariom D Assay TC0600008190.hg.1 ENST00000371211.6 Ensembl
Affymetrix Human Clariom D Assay TC0600008190.hg.1 ENST00000371211.6 Ensembl
Affymetrix Human Clariom D Assay TC0600008190.hg.1 ENST00000371211.6 Ensembl
Affymetrix Human Clariom D Assay TC0600008190.hg.1 ENST00000371211.6 Ensembl
Affymetrix Human Clariom D Assay TC0600008190.hg.1 ENST00000371211.6 Ensembl
Affymetrix Human Clariom D Assay TC0600008190.hg.1 ENST00000371211.6 Ensembl
Affymetrix Human Clariom D Assay TC0600008190.hg.1 ENST00000371211.6 Ensembl
Affymetrix Human Clariom D Assay TC0600008190.hg.1 ENST00000371211.6 Ensembl
Affymetrix Human Clariom D Assay TC0600008190.hg.1 ENST00000371211.6 Ensembl
Affymetrix Human Clariom D Assay TC0600008190.hg.1 ENST00000371211.6 Ensembl
Affymetrix Human Clariom D Assay TC0600008190.hg.1 ENST00000393699.2 Ensembl
Affymetrix Human Clariom D Assay TC0600008190.hg.1 ENST00000393699.2 Ensembl
Affymetrix Human Clariom D Assay TC0600008190.hg.1 ENST00000393699.2 Ensembl
Affymetrix Human Clariom D Assay TC0600008190.hg.1 ENST00000393699.2 Ensembl
Affymetrix Human Clariom D Assay TC0600008190.hg.1 ENST00000393699.2 Ensembl
Affymetrix Human Clariom D Assay TC0600008190.hg.1 ENST00000393699.2 Ensembl
Affymetrix Human Clariom D Assay TC0600008190.hg.1 ENST00000393699.2 Ensembl
Affymetrix Human Clariom D Assay TC0600008190.hg.1 ENST00000393699.2 Ensembl
Affymetrix Human Clariom D Assay TC0600008190.hg.1 ENST00000393699.2 Ensembl
Affymetrix Human Clariom D Assay TC0600008190.hg.1 ENST00000393699.2 Ensembl
Affymetrix Human Clariom D Assay TC0600008190.hg.1 ENST00000393699.2 Ensembl
Affymetrix Human Clariom D Assay TC0600008190.hg.1 ENST00000489301.6 Ensembl
Affymetrix Human Clariom D Assay TC0600008190.hg.1 ENST00000489301.6 Ensembl
Affymetrix Human Clariom D Assay TC0600008190.hg.1 ENST00000489301.6 Ensembl
Affymetrix Human Clariom D Assay TC0600008190.hg.1 ENST00000489301.6 Ensembl
Affymetrix Human Clariom D Assay TC0600008190.hg.1 ENST00000489301.6 Ensembl
Affymetrix Human Clariom D Assay TC0600008190.hg.1 ENST00000489301.6 Ensembl
Affymetrix Human Clariom D Assay TC0600008190.hg.1 ENST00000489301.6 Ensembl
Affymetrix Human Clariom D Assay TC0600008190.hg.1 ENST00000489301.6 Ensembl
Affymetrix Human Clariom D Assay TC0600008190.hg.1 ENST00000489301.6 Ensembl
Affymetrix Human Clariom D Assay TC0600008190.hg.1 ENST00000489301.6 Ensembl
Affymetrix Human Clariom D Assay TC0600008190.hg.1 ENST00000489301.6 Ensembl
Affymetrix Human Clariom D Assay TC0600008190.hg.1 ENST00000489301.6 Ensembl
Affymetrix Human Clariom D Assay TC0600008190.hg.1 ENST00000489301.6 Ensembl
Affymetrix Human Clariom D Assay TC0600008190.hg.1 ENST00000510695.1 Ensembl
Affymetrix Human Clariom D Assay TC0600008190.hg.1 ENST00000510695.1 Ensembl
Affymetrix Human Clariom D Assay TC0600008190.hg.1 ENST00000510695.1 Ensembl
Affymetrix Human Clariom D Assay TC0600008190.hg.1 ENST00000510695.1 Ensembl
Affymetrix Human Clariom D Assay TC0600008190.hg.1 NM_181744.3 Refseq
Affymetrix Human Clariom D Assay TC0600008190.hg.1 NM_181744.3 Refseq
Affymetrix Human Clariom D Assay TC0600008190.hg.1 NM_181744.3 Refseq
Affymetrix Human Clariom D Assay TC0600008190.hg.1 NM_181744.3 Refseq
Affymetrix Human Clariom D Assay TC0600008190.hg.1 NM_181744.3 Refseq
Affymetrix Human Clariom D Assay TC0600008190.hg.1 NM_181744.3 Refseq
Affymetrix Human Clariom D Assay TC0600008190.hg.1 NM_181744.3 Refseq
Affymetrix Human Clariom D Assay TC0600008190.hg.1 NM_181744.3 Refseq
Affymetrix Human Clariom D Assay TC0600008190.hg.1 NM_181744.3 Refseq
Affymetrix Human Clariom D Assay TC0600008190.hg.1 NM_181744.3 Refseq
Affymetrix Human Clariom D Assay TC0600008190.hg.1 NM_181744.3 Refseq
Affymetrix Human Clariom D Assay TC0600008190.hg.1 NM_181744.3 Refseq
Affymetrix Human Clariom D Assay TC0600008190.hg.1 NM_181744.3 Refseq
Affymetrix Human Clariom D Assay TC0600008190.hg.1 NM_181744.3 Refseq
Affymetrix Human Clariom D Assay TC0600008190.hg.1 NR_033806.1 Refseq
Affymetrix Human Clariom D Assay TC0600008190.hg.1 NR_033806.1 Refseq
Affymetrix Human Clariom D Assay TC0600008190.hg.1 NR_033806.1 Refseq
Affymetrix Human Clariom D Assay TC0600008190.hg.1 NR_033806.1 Refseq
Affymetrix Human Clariom D Assay TC0600008190.hg.1 NR_033806.1 Refseq
Affymetrix Human Clariom D Assay TC0600008190.hg.1 NR_033806.1 Refseq
Affymetrix Human Clariom D Assay TC0600008190.hg.1 NR_033806.1 Refseq
Affymetrix Human Clariom D Assay TC0600008190.hg.1 NR_033806.1 Refseq
Affymetrix Human Clariom D Assay TC0600008190.hg.1 NR_033806.1 Refseq
Affymetrix Human Clariom D Assay TC0600008190.hg.1 NR_033806.1 Refseq
Affymetrix Human Clariom D Assay TC0600008190.hg.1 NR_033806.1 Refseq
Affymetrix Human Clariom D Assay TC0600008190.hg.1 NR_033806.1 Refseq
Affymetrix Human Clariom D Assay TC0600008190.hg.1 NR_033806.1 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0