This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05044998_g1
See other LINC01018 GE Assays ›
Gene Symbol:
LINC01018
Gene Name:
long intergenic non-protein coding RNA 1018
Gene Aliases:
SRHC
Chromosome Location:
Chr.5: 6582136 - 6588500 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05044998_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LINC01018
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA DA240476.1 - 1 128 98

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0500006643.hg.1 ENST00000505626.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500006643.hg.1 ENST00000505626.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500006643.hg.1 ENST00000505626.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500006643.hg.1 ENST00000505626.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500006643.hg.1 ENST00000505626.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500006643.hg.1 ENST00000505626.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500006643.hg.1 ENST00000505626.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500006643.hg.1 ENST00000505626.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500006643.hg.1 ENST00000505626.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500006643.hg.1 ENST00000505626.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500006643.hg.1 ENST00000505626.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500006643.hg.1 ENST00000505626.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500006643.hg.1 ENST00000505626.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500006643.hg.1 ENST00000505626.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500006643.hg.1 ENST00000505626.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500006643.hg.1 ENST00000505626.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500006643.hg.1 ENST00000505626.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500006643.hg.1 ENST00000505626.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500006643.hg.1 ENST00000505626.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500006643.hg.1 ENST00000505626.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500006643.hg.1 ENST00000505626.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500006643.hg.1 ENST00000505626.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500006643.hg.1 ENST00000505626.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500006643.hg.1 ENST00000505626.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500006643.hg.1 ENST00000505626.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500006643.hg.1 ENST00000505626.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500006643.hg.1 ENST00000505626.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500006643.hg.1 ENST00000505626.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500006643.hg.1 ENST00000507582.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500006643.hg.1 ENST00000507582.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500006643.hg.1 ENST00000507582.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500006643.hg.1 ENST00000507582.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500006643.hg.1 ENST00000507582.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500006643.hg.1 ENST00000507582.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500006643.hg.1 ENST00000507582.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500006643.hg.1 ENST00000507582.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500006643.hg.1 ENST00000507582.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500006643.hg.1 ENST00000507628.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500006643.hg.1 ENST00000507628.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500006643.hg.1 ENST00000507628.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500006643.hg.1 ENST00000507628.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500006643.hg.1 ENST00000507628.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500006643.hg.1 ENST00000507628.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500006643.hg.1 ENST00000513844.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500006643.hg.1 ENST00000513844.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500006643.hg.1 ENST00000513844.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500006643.hg.1 ENST00000513844.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500006643.hg.1 ENST00000513844.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500006643.hg.1 ENST00000513844.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500006643.hg.1 NR_024423.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500006643.hg.1 NR_024423.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500006643.hg.1 NR_024423.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500006643.hg.1 NR_024423.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500006643.hg.1 NR_024423.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500006643.hg.1 NR_024423.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500006643.hg.1 NR_024423.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500006643.hg.1 NR_024423.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500006643.hg.1 NR_024423.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500006643.hg.1 NR_024423.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500006643.hg.1 NR_024423.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500006643.hg.1 NR_024423.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500006643.hg.1 NR_024423.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500006643.hg.1 NR_024423.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500006643.hg.1 NR_024424.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500006643.hg.1 NR_024424.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500006643.hg.1 NR_024424.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500006643.hg.1 NR_024424.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500006643.hg.1 NR_024424.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500006643.hg.1 NR_024424.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500006643.hg.1 NR_024424.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500006643.hg.1 NR_024424.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500006643.hg.1 NR_024424.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500006643.hg.1 NR_024424.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500006643.hg.1 NR_024424.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500006643.hg.1 NR_024424.2 Refseq
Affymetrix Human Clariom D Assay TC0500006643.hg.1 ENST00000505626.1 Ensembl
Affymetrix Human Clariom D Assay TC0500006643.hg.1 ENST00000505626.1 Ensembl
Affymetrix Human Clariom D Assay TC0500006643.hg.1 ENST00000505626.1 Ensembl
Affymetrix Human Clariom D Assay TC0500006643.hg.1 ENST00000505626.1 Ensembl
Affymetrix Human Clariom D Assay TC0500006643.hg.1 ENST00000505626.1 Ensembl
Affymetrix Human Clariom D Assay TC0500006643.hg.1 ENST00000505626.1 Ensembl
Affymetrix Human Clariom D Assay TC0500006643.hg.1 ENST00000505626.1 Ensembl
Affymetrix Human Clariom D Assay TC0500006643.hg.1 ENST00000505626.1 Ensembl
Affymetrix Human Clariom D Assay TC0500006643.hg.1 ENST00000505626.1 Ensembl
Affymetrix Human Clariom D Assay TC0500006643.hg.1 ENST00000505626.1 Ensembl
Affymetrix Human Clariom D Assay TC0500006643.hg.1 ENST00000505626.1 Ensembl
Affymetrix Human Clariom D Assay TC0500006643.hg.1 ENST00000505626.1 Ensembl
Affymetrix Human Clariom D Assay TC0500006643.hg.1 ENST00000505626.1 Ensembl
Affymetrix Human Clariom D Assay TC0500006643.hg.1 ENST00000505626.1 Ensembl
Affymetrix Human Clariom D Assay TC0500006643.hg.1 ENST00000505626.1 Ensembl
Affymetrix Human Clariom D Assay TC0500006643.hg.1 ENST00000505626.1 Ensembl
Affymetrix Human Clariom D Assay TC0500006643.hg.1 ENST00000505626.1 Ensembl
Affymetrix Human Clariom D Assay TC0500006643.hg.1 ENST00000505626.1 Ensembl
Affymetrix Human Clariom D Assay TC0500006643.hg.1 ENST00000505626.1 Ensembl
Affymetrix Human Clariom D Assay TC0500006643.hg.1 ENST00000505626.1 Ensembl
Affymetrix Human Clariom D Assay TC0500006643.hg.1 ENST00000505626.1 Ensembl
Affymetrix Human Clariom D Assay TC0500006643.hg.1 ENST00000505626.1 Ensembl
Affymetrix Human Clariom D Assay TC0500006643.hg.1 ENST00000505626.1 Ensembl
Affymetrix Human Clariom D Assay TC0500006643.hg.1 ENST00000505626.1 Ensembl
Affymetrix Human Clariom D Assay TC0500006643.hg.1 ENST00000505626.1 Ensembl
Affymetrix Human Clariom D Assay TC0500006643.hg.1 ENST00000505626.1 Ensembl
Affymetrix Human Clariom D Assay TC0500006643.hg.1 ENST00000505626.1 Ensembl
Affymetrix Human Clariom D Assay TC0500006643.hg.1 ENST00000505626.1 Ensembl
Affymetrix Human Clariom D Assay TC0500006643.hg.1 ENST00000507582.1 Ensembl
Affymetrix Human Clariom D Assay TC0500006643.hg.1 ENST00000507582.1 Ensembl
Affymetrix Human Clariom D Assay TC0500006643.hg.1 ENST00000507582.1 Ensembl
Affymetrix Human Clariom D Assay TC0500006643.hg.1 ENST00000507582.1 Ensembl
Affymetrix Human Clariom D Assay TC0500006643.hg.1 ENST00000507582.1 Ensembl
Affymetrix Human Clariom D Assay TC0500006643.hg.1 ENST00000507582.1 Ensembl
Affymetrix Human Clariom D Assay TC0500006643.hg.1 ENST00000507582.1 Ensembl
Affymetrix Human Clariom D Assay TC0500006643.hg.1 ENST00000507582.1 Ensembl
Affymetrix Human Clariom D Assay TC0500006643.hg.1 ENST00000507582.1 Ensembl
Affymetrix Human Clariom D Assay TC0500006643.hg.1 ENST00000507628.1 Ensembl
Affymetrix Human Clariom D Assay TC0500006643.hg.1 ENST00000507628.1 Ensembl
Affymetrix Human Clariom D Assay TC0500006643.hg.1 ENST00000507628.1 Ensembl
Affymetrix Human Clariom D Assay TC0500006643.hg.1 ENST00000507628.1 Ensembl
Affymetrix Human Clariom D Assay TC0500006643.hg.1 ENST00000507628.1 Ensembl
Affymetrix Human Clariom D Assay TC0500006643.hg.1 ENST00000507628.1 Ensembl
Affymetrix Human Clariom D Assay TC0500006643.hg.1 ENST00000513844.1 Ensembl
Affymetrix Human Clariom D Assay TC0500006643.hg.1 ENST00000513844.1 Ensembl
Affymetrix Human Clariom D Assay TC0500006643.hg.1 ENST00000513844.1 Ensembl
Affymetrix Human Clariom D Assay TC0500006643.hg.1 ENST00000513844.1 Ensembl
Affymetrix Human Clariom D Assay TC0500006643.hg.1 ENST00000513844.1 Ensembl
Affymetrix Human Clariom D Assay TC0500006643.hg.1 ENST00000513844.1 Ensembl
Affymetrix Human Clariom D Assay TC0500006643.hg.1 NR_024423.2 Refseq
Affymetrix Human Clariom D Assay TC0500006643.hg.1 NR_024423.2 Refseq
Affymetrix Human Clariom D Assay TC0500006643.hg.1 NR_024423.2 Refseq
Affymetrix Human Clariom D Assay TC0500006643.hg.1 NR_024423.2 Refseq
Affymetrix Human Clariom D Assay TC0500006643.hg.1 NR_024423.2 Refseq
Affymetrix Human Clariom D Assay TC0500006643.hg.1 NR_024423.2 Refseq
Affymetrix Human Clariom D Assay TC0500006643.hg.1 NR_024423.2 Refseq
Affymetrix Human Clariom D Assay TC0500006643.hg.1 NR_024423.2 Refseq
Affymetrix Human Clariom D Assay TC0500006643.hg.1 NR_024423.2 Refseq
Affymetrix Human Clariom D Assay TC0500006643.hg.1 NR_024423.2 Refseq
Affymetrix Human Clariom D Assay TC0500006643.hg.1 NR_024423.2 Refseq
Affymetrix Human Clariom D Assay TC0500006643.hg.1 NR_024423.2 Refseq
Affymetrix Human Clariom D Assay TC0500006643.hg.1 NR_024423.2 Refseq
Affymetrix Human Clariom D Assay TC0500006643.hg.1 NR_024423.2 Refseq
Affymetrix Human Clariom D Assay TC0500006643.hg.1 NR_024424.2 Refseq
Affymetrix Human Clariom D Assay TC0500006643.hg.1 NR_024424.2 Refseq
Affymetrix Human Clariom D Assay TC0500006643.hg.1 NR_024424.2 Refseq
Affymetrix Human Clariom D Assay TC0500006643.hg.1 NR_024424.2 Refseq
Affymetrix Human Clariom D Assay TC0500006643.hg.1 NR_024424.2 Refseq
Affymetrix Human Clariom D Assay TC0500006643.hg.1 NR_024424.2 Refseq
Affymetrix Human Clariom D Assay TC0500006643.hg.1 NR_024424.2 Refseq
Affymetrix Human Clariom D Assay TC0500006643.hg.1 NR_024424.2 Refseq
Affymetrix Human Clariom D Assay TC0500006643.hg.1 NR_024424.2 Refseq
Affymetrix Human Clariom D Assay TC0500006643.hg.1 NR_024424.2 Refseq
Affymetrix Human Clariom D Assay TC0500006643.hg.1 NR_024424.2 Refseq
Affymetrix Human Clariom D Assay TC0500006643.hg.1 NR_024424.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0