Assay Details
Gene Symbol: C3orf67-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_110820.1 | 3 | 501 | 107 | ||
GenBank mRNA | AK090895.1 | - | 3 | 501 | 107 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0300007625.hg.1 | ENST00000463703.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007625.hg.1 | ENST00000463703.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007625.hg.1 | ENST00000463703.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007625.hg.1 | ENST00000463703.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007625.hg.1 | ENST00000463703.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007625.hg.1 | ENST00000463703.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007625.hg.1 | ENST00000463703.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007625.hg.1 | ENST00000463703.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007625.hg.1 | ENST00000463703.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007625.hg.1 | ENST00000463703.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007625.hg.1 | ENST00000482372.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007625.hg.1 | ENST00000482372.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007625.hg.1 | ENST00000482372.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007625.hg.1 | ENST00000482372.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007625.hg.1 | ENST00000492031.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007625.hg.1 | ENST00000492031.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007625.hg.1 | ENST00000492031.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007625.hg.1 | ENST00000492031.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007625.hg.1 | ENST00000492031.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007625.hg.1 | ENST00000492031.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007625.hg.1 | ENST00000492031.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007625.hg.1 | ENST00000492031.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007625.hg.1 | ENST00000492031.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007625.hg.1 | ENST00000492031.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007625.hg.1 | ENST00000492031.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007625.hg.1 | ENST00000492031.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007625.hg.1 | ENST00000493123.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007625.hg.1 | ENST00000493123.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007625.hg.1 | ENST00000493123.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007625.hg.1 | NR_110820.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007625.hg.1 | NR_110820.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007625.hg.1 | NR_110820.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007625.hg.1 | NR_110820.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007625.hg.1 | ENST00000463703.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007625.hg.1 | ENST00000463703.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007625.hg.1 | ENST00000463703.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007625.hg.1 | ENST00000463703.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007625.hg.1 | ENST00000463703.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007625.hg.1 | ENST00000463703.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007625.hg.1 | ENST00000463703.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007625.hg.1 | ENST00000463703.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007625.hg.1 | ENST00000463703.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007625.hg.1 | ENST00000463703.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007625.hg.1 | ENST00000482372.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007625.hg.1 | ENST00000482372.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007625.hg.1 | ENST00000482372.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007625.hg.1 | ENST00000482372.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007625.hg.1 | ENST00000492031.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007625.hg.1 | ENST00000492031.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007625.hg.1 | ENST00000492031.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007625.hg.1 | ENST00000492031.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007625.hg.1 | ENST00000492031.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007625.hg.1 | ENST00000492031.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007625.hg.1 | ENST00000492031.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007625.hg.1 | ENST00000492031.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007625.hg.1 | ENST00000492031.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007625.hg.1 | ENST00000492031.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007625.hg.1 | ENST00000492031.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007625.hg.1 | ENST00000492031.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007625.hg.1 | ENST00000493123.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007625.hg.1 | ENST00000493123.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007625.hg.1 | ENST00000493123.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007625.hg.1 | NR_110820.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007625.hg.1 | NR_110820.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007625.hg.1 | NR_110820.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007625.hg.1 | NR_110820.1 | Refseq |
