This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05043135_s1
See other EGR4 GE Assays ›
Gene Symbol:
EGR4
Gene Name:
early growth response 4
Gene Aliases:
NGFI-C, NGFIC, PAT133
Chromosome Location:
Chr.2: 73290929 - 73293701 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05043135_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: EGR4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001965.3 NP_001956.3 1 126 88
GenBank mRNA AK314154.1 - 1 132 88
DA711319.1 - 1 132 88

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK314154.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene EGR4

Panther Classification:

Molecular Function -

C2H2 zinc finger transcription factor

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
positive regulation of cell proliferation
DNA binding
transcription factor activity, sequence-specific DNA binding
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0200013063.hg.1 ENST00000436467.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013063.hg.1 ENST00000436467.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013063.hg.1 ENST00000436467.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013063.hg.1 ENST00000436467.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013063.hg.1 ENST00000436467.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013063.hg.1 ENST00000436467.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013063.hg.1 ENST00000545030.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013063.hg.1 ENST00000545030.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013063.hg.1 ENST00000545030.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013063.hg.1 ENST00000545030.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013063.hg.1 ENST00000545030.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013063.hg.1 ENST00000545030.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013063.hg.1 NM_001965.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200013063.hg.1 NM_001965.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200013063.hg.1 NM_001965.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200013063.hg.1 NM_001965.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200013063.hg.1 NM_001965.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200013063.hg.1 NM_001965.3 Refseq
Affymetrix Human Clariom D Assay TC0200013063.hg.1 ENST00000436467.3 Ensembl
Affymetrix Human Clariom D Assay TC0200013063.hg.1 ENST00000436467.3 Ensembl
Affymetrix Human Clariom D Assay TC0200013063.hg.1 ENST00000436467.3 Ensembl
Affymetrix Human Clariom D Assay TC0200013063.hg.1 ENST00000436467.3 Ensembl
Affymetrix Human Clariom D Assay TC0200013063.hg.1 ENST00000436467.3 Ensembl
Affymetrix Human Clariom D Assay TC0200013063.hg.1 ENST00000436467.3 Ensembl
Affymetrix Human Clariom D Assay TC0200013063.hg.1 ENST00000545030.1 Ensembl
Affymetrix Human Clariom D Assay TC0200013063.hg.1 ENST00000545030.1 Ensembl
Affymetrix Human Clariom D Assay TC0200013063.hg.1 ENST00000545030.1 Ensembl
Affymetrix Human Clariom D Assay TC0200013063.hg.1 ENST00000545030.1 Ensembl
Affymetrix Human Clariom D Assay TC0200013063.hg.1 ENST00000545030.1 Ensembl
Affymetrix Human Clariom D Assay TC0200013063.hg.1 ENST00000545030.1 Ensembl
Affymetrix Human Clariom D Assay TC0200013063.hg.1 NM_001965.3 Refseq
Affymetrix Human Clariom D Assay TC0200013063.hg.1 NM_001965.3 Refseq
Affymetrix Human Clariom D Assay TC0200013063.hg.1 NM_001965.3 Refseq
Affymetrix Human Clariom D Assay TC0200013063.hg.1 NM_001965.3 Refseq
Affymetrix Human Clariom D Assay TC0200013063.hg.1 NM_001965.3 Refseq
Affymetrix Human Clariom D Assay TC0200013063.hg.1 NM_001965.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7db4fb6694-gx648:80/100.66.128.142:80.
git-commit: 672a5a8c50d6b616e949be631a941c86748ade1b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header