This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05042865_s1
See other SOS1-IT1 GE Assays ›
Gene Symbol:
SOS1-IT1
Gene Name:
SOS1 intronic transcript 1
Gene Aliases:
-
Chromosome Location:
-
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05042865_s1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SOS1-IT1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA BF769867.1 - 1 128 124

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Homo sapiens Both primers and probe map within a single exon

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0200012308.hg.1 ENST00000420644.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012308.hg.1 ENST00000420644.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012308.hg.1 ENST00000594472.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012308.hg.1 ENST00000594472.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012308.hg.1 ENST00000594472.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012308.hg.1 ENST00000594472.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012308.hg.1 ENST00000609942.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012308.hg.1 ENST00000609942.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012308.hg.1 ENST00000609942.1 Ensembl
Affymetrix Human Clariom D Assay TC0200012308.hg.1 ENST00000420644.1 Ensembl
Affymetrix Human Clariom D Assay TC0200012308.hg.1 ENST00000420644.1 Ensembl
Affymetrix Human Clariom D Assay TC0200012308.hg.1 ENST00000594472.1 Ensembl
Affymetrix Human Clariom D Assay TC0200012308.hg.1 ENST00000594472.1 Ensembl
Affymetrix Human Clariom D Assay TC0200012308.hg.1 ENST00000594472.1 Ensembl
Affymetrix Human Clariom D Assay TC0200012308.hg.1 ENST00000594472.1 Ensembl
Affymetrix Human Clariom D Assay TC0200012308.hg.1 ENST00000609942.1 Ensembl
Affymetrix Human Clariom D Assay TC0200012308.hg.1 ENST00000609942.1 Ensembl
Affymetrix Human Clariom D Assay TC0200012308.hg.1 ENST00000609942.1 Ensembl

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0