This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05042389_s1
See other IGLL1 GE Assays ›
Gene Symbol:
IGLL1
Gene Name:
immunoglobulin lambda like polypeptide 1
Gene Aliases:
14.1, AGM2, CD179b, IGL1, IGL5, IGLJ14.1, IGLL, IGO, IGVPB, VPREB2
Chromosome Location:
Chr.22: 23573125 - 23580548 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05042389_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: IGLL1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_020070.3 NP_064455.1 1 127 118
NM_152855.2 NP_690594.1 1 127 118
XM_011530169.2 XP_011528471.1 1 367 118
GenBank mRNA BC012293.1 - 1 67 4508892 118
BC030239.1 - 1 69 118
M27749.1 - 1 127 118

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the BC012293.1, BC030239.1, M27749.1, NM_020070.3, NM_152855.2, XM_011530169.2 transcripts.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene IGLL1

Panther Classification:

Molecular Function -

immunoglobulin major histocompatibility complex protein

Gene Ontology Categories:

Function(s) Process(es)

phagocytosis, recognition
phagocytosis, engulfment
immune response
complement activation, classical pathway
defense response to bacterium
innate immune response
B cell receptor signaling pathway
positive regulation of B cell activation
antigen binding
immunoglobulin receptor binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC2200008174.hg.1 ENST00000249053.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008174.hg.1 ENST00000249053.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008174.hg.1 ENST00000249053.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008174.hg.1 ENST00000249053.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008174.hg.1 ENST00000249053.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008174.hg.1 ENST00000249053.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008174.hg.1 ENST00000330377.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008174.hg.1 ENST00000330377.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008174.hg.1 ENST00000330377.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008174.hg.1 ENST00000330377.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008174.hg.1 ENST00000330377.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008174.hg.1 ENST00000330377.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008174.hg.1 ENST00000330377.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008174.hg.1 ENST00000330377.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008174.hg.1 ENST00000438703.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008174.hg.1 ENST00000438703.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008174.hg.1 ENST00000438703.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008174.hg.1 ENST00000438703.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008174.hg.1 ENST00000438703.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008174.hg.1 ENST00000438703.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200008174.hg.1 NM_020070.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008174.hg.1 NM_020070.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008174.hg.1 NM_020070.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008174.hg.1 NM_020070.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008174.hg.1 NM_020070.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008174.hg.1 NM_020070.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008174.hg.1 NM_020070.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008174.hg.1 NM_020070.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008174.hg.1 NM_152855.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008174.hg.1 NM_152855.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008174.hg.1 NM_152855.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008174.hg.1 NM_152855.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008174.hg.1 NM_152855.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008174.hg.1 NM_152855.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200008174.hg.1 NM_152855.2 Refseq
Affymetrix Human Clariom D Assay TC2200008174.hg.1 ENST00000249053.3 Ensembl
Affymetrix Human Clariom D Assay TC2200008174.hg.1 ENST00000249053.3 Ensembl
Affymetrix Human Clariom D Assay TC2200008174.hg.1 ENST00000249053.3 Ensembl
Affymetrix Human Clariom D Assay TC2200008174.hg.1 ENST00000249053.3 Ensembl
Affymetrix Human Clariom D Assay TC2200008174.hg.1 ENST00000249053.3 Ensembl
Affymetrix Human Clariom D Assay TC2200008174.hg.1 ENST00000249053.3 Ensembl
Affymetrix Human Clariom D Assay TC2200008174.hg.1 ENST00000330377.2 Ensembl
Affymetrix Human Clariom D Assay TC2200008174.hg.1 ENST00000330377.2 Ensembl
Affymetrix Human Clariom D Assay TC2200008174.hg.1 ENST00000330377.2 Ensembl
Affymetrix Human Clariom D Assay TC2200008174.hg.1 ENST00000330377.2 Ensembl
Affymetrix Human Clariom D Assay TC2200008174.hg.1 ENST00000330377.2 Ensembl
Affymetrix Human Clariom D Assay TC2200008174.hg.1 ENST00000330377.2 Ensembl
Affymetrix Human Clariom D Assay TC2200008174.hg.1 ENST00000330377.2 Ensembl
Affymetrix Human Clariom D Assay TC2200008174.hg.1 ENST00000330377.2 Ensembl
Affymetrix Human Clariom D Assay TC2200008174.hg.1 ENST00000438703.1 Ensembl
Affymetrix Human Clariom D Assay TC2200008174.hg.1 ENST00000438703.1 Ensembl
Affymetrix Human Clariom D Assay TC2200008174.hg.1 ENST00000438703.1 Ensembl
Affymetrix Human Clariom D Assay TC2200008174.hg.1 ENST00000438703.1 Ensembl
Affymetrix Human Clariom D Assay TC2200008174.hg.1 ENST00000438703.1 Ensembl
Affymetrix Human Clariom D Assay TC2200008174.hg.1 ENST00000438703.1 Ensembl
Affymetrix Human Clariom D Assay TC2200008174.hg.1 NM_020070.3 Refseq
Affymetrix Human Clariom D Assay TC2200008174.hg.1 NM_020070.3 Refseq
Affymetrix Human Clariom D Assay TC2200008174.hg.1 NM_020070.3 Refseq
Affymetrix Human Clariom D Assay TC2200008174.hg.1 NM_020070.3 Refseq
Affymetrix Human Clariom D Assay TC2200008174.hg.1 NM_020070.3 Refseq
Affymetrix Human Clariom D Assay TC2200008174.hg.1 NM_020070.3 Refseq
Affymetrix Human Clariom D Assay TC2200008174.hg.1 NM_020070.3 Refseq
Affymetrix Human Clariom D Assay TC2200008174.hg.1 NM_020070.3 Refseq
Affymetrix Human Clariom D Assay TC2200008174.hg.1 NM_152855.2 Refseq
Affymetrix Human Clariom D Assay TC2200008174.hg.1 NM_152855.2 Refseq
Affymetrix Human Clariom D Assay TC2200008174.hg.1 NM_152855.2 Refseq
Affymetrix Human Clariom D Assay TC2200008174.hg.1 NM_152855.2 Refseq
Affymetrix Human Clariom D Assay TC2200008174.hg.1 NM_152855.2 Refseq
Affymetrix Human Clariom D Assay TC2200008174.hg.1 NM_152855.2 Refseq
Affymetrix Human Clariom D Assay TC2200008174.hg.1 NM_152855.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0