Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008232.hg.1 |
ENST00000379751.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008232.hg.1 |
ENST00000379751.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008232.hg.1 |
ENST00000379751.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008232.hg.1 |
ENST00000379751.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008232.hg.1 |
NM_001810.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008232.hg.1 |
NM_001810.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008232.hg.1 |
NM_001810.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008232.hg.1 |
NM_001810.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008232.hg.1 |
ENST00000379751.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008232.hg.1 |
ENST00000379751.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008232.hg.1 |
ENST00000379751.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008232.hg.1 |
ENST00000379751.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008232.hg.1 |
NM_001810.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008232.hg.1 |
NM_001810.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008232.hg.1 |
NM_001810.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008232.hg.1 |
NM_001810.5 |
Refseq |