Assay Details
Gene Symbol: WNK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | DQ925669.1 | - | 3 | 127 | 67 | |
JQ358908.1 | - | 30 | 8007 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() |

Gene Symbol: WNK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | DQ925669.1 | - | 3 | 127 | 67 | |
JQ358908.1 | - | 30 | 8007 | 67 |
Set Membership: |
![]() ![]() ![]() |