This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05030068_s1
See other OPN5 GE Assays ›
Gene Symbol:
OPN5
Gene Name:
opsin 5
Gene Aliases:
GPR136, GRP136, PGR12, TMEM13
Chromosome Location:
Chr.6: 47782037 - 47831088 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05030068_s1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: OPN5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_181744.3 NP_859528.1 4 117
NR_033806.1 4 117
XM_017010409.1 XP_016865898.1 2 117
XM_017010411.1 XP_016865900.1 3 117
XM_017010412.1 XP_016865901.1 3 117
XM_017010413.1 XP_016865902.1 3 117
XM_017010415.1 XP_016865904.1 4 117
XM_017010416.1 XP_016865905.1 4 117
GenBank mRNA AK097448.1 - 1 154 117
AY255615.1 - 2 117
AY288419.1 - 4 117
AY377391.1 - 4 117
BC126194.1 - 4 8991910 117
BC126198.1 - 4 8991914 117
BX647224.1 - 4 117

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene OPN5

Panther Classification:

Molecular Function -

G-protein coupled receptor

Gene Ontology Categories:

Function(s) Process(es)

G-protein coupled receptor signaling pathway
visual perception
phototransduction
detection of visible light
protein-chromophore linkage
cellular response to light stimulus
11-cis retinal binding
G-protein coupled photoreceptor activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0600008190.hg.1 ENST00000244799.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008190.hg.1 ENST00000244799.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008190.hg.1 ENST00000244799.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008190.hg.1 ENST00000244799.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008190.hg.1 ENST00000244799.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008190.hg.1 ENST00000244799.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008190.hg.1 ENST00000244799.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008190.hg.1 ENST00000244799.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008190.hg.1 ENST00000244799.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008190.hg.1 ENST00000244799.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008190.hg.1 ENST00000244799.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008190.hg.1 ENST00000244799.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008190.hg.1 ENST00000244799.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008190.hg.1 ENST00000371211.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008190.hg.1 ENST00000371211.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008190.hg.1 ENST00000371211.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008190.hg.1 ENST00000371211.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008190.hg.1 ENST00000371211.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008190.hg.1 ENST00000371211.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008190.hg.1 ENST00000371211.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008190.hg.1 ENST00000371211.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008190.hg.1 ENST00000371211.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008190.hg.1 ENST00000371211.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008190.hg.1 ENST00000371211.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008190.hg.1 ENST00000371211.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008190.hg.1 ENST00000371211.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008190.hg.1 ENST00000393699.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008190.hg.1 ENST00000393699.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008190.hg.1 ENST00000393699.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008190.hg.1 ENST00000393699.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008190.hg.1 ENST00000393699.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008190.hg.1 ENST00000393699.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008190.hg.1 ENST00000393699.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008190.hg.1 ENST00000393699.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008190.hg.1 ENST00000393699.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008190.hg.1 ENST00000393699.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008190.hg.1 ENST00000393699.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008190.hg.1 ENST00000489301.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008190.hg.1 ENST00000489301.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008190.hg.1 ENST00000489301.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008190.hg.1 ENST00000489301.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008190.hg.1 ENST00000489301.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008190.hg.1 ENST00000489301.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008190.hg.1 ENST00000489301.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008190.hg.1 ENST00000489301.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008190.hg.1 ENST00000489301.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008190.hg.1 ENST00000489301.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008190.hg.1 ENST00000489301.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008190.hg.1 ENST00000489301.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008190.hg.1 ENST00000489301.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008190.hg.1 ENST00000510695.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008190.hg.1 ENST00000510695.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008190.hg.1 ENST00000510695.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008190.hg.1 ENST00000510695.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600008190.hg.1 NM_181744.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008190.hg.1 NM_181744.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008190.hg.1 NM_181744.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008190.hg.1 NM_181744.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008190.hg.1 NM_181744.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008190.hg.1 NM_181744.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008190.hg.1 NM_181744.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008190.hg.1 NM_181744.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008190.hg.1 NM_181744.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008190.hg.1 NM_181744.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008190.hg.1 NM_181744.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008190.hg.1 NM_181744.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008190.hg.1 NM_181744.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008190.hg.1 NM_181744.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008190.hg.1 NR_033806.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008190.hg.1 NR_033806.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008190.hg.1 NR_033806.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008190.hg.1 NR_033806.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008190.hg.1 NR_033806.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008190.hg.1 NR_033806.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008190.hg.1 NR_033806.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008190.hg.1 NR_033806.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008190.hg.1 NR_033806.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008190.hg.1 NR_033806.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008190.hg.1 NR_033806.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008190.hg.1 NR_033806.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600008190.hg.1 NR_033806.1 Refseq
Affymetrix Human Clariom D Assay TC0600008190.hg.1 ENST00000244799.4 Ensembl
Affymetrix Human Clariom D Assay TC0600008190.hg.1 ENST00000244799.4 Ensembl
Affymetrix Human Clariom D Assay TC0600008190.hg.1 ENST00000244799.4 Ensembl
Affymetrix Human Clariom D Assay TC0600008190.hg.1 ENST00000244799.4 Ensembl
Affymetrix Human Clariom D Assay TC0600008190.hg.1 ENST00000244799.4 Ensembl
Affymetrix Human Clariom D Assay TC0600008190.hg.1 ENST00000244799.4 Ensembl
Affymetrix Human Clariom D Assay TC0600008190.hg.1 ENST00000244799.4 Ensembl
Affymetrix Human Clariom D Assay TC0600008190.hg.1 ENST00000244799.4 Ensembl
Affymetrix Human Clariom D Assay TC0600008190.hg.1 ENST00000244799.4 Ensembl
Affymetrix Human Clariom D Assay TC0600008190.hg.1 ENST00000244799.4 Ensembl
Affymetrix Human Clariom D Assay TC0600008190.hg.1 ENST00000244799.4 Ensembl
Affymetrix Human Clariom D Assay TC0600008190.hg.1 ENST00000244799.4 Ensembl
Affymetrix Human Clariom D Assay TC0600008190.hg.1 ENST00000244799.4 Ensembl
Affymetrix Human Clariom D Assay TC0600008190.hg.1 ENST00000371211.6 Ensembl
Affymetrix Human Clariom D Assay TC0600008190.hg.1 ENST00000371211.6 Ensembl
Affymetrix Human Clariom D Assay TC0600008190.hg.1 ENST00000371211.6 Ensembl
Affymetrix Human Clariom D Assay TC0600008190.hg.1 ENST00000371211.6 Ensembl
Affymetrix Human Clariom D Assay TC0600008190.hg.1 ENST00000371211.6 Ensembl
Affymetrix Human Clariom D Assay TC0600008190.hg.1 ENST00000371211.6 Ensembl
Affymetrix Human Clariom D Assay TC0600008190.hg.1 ENST00000371211.6 Ensembl
Affymetrix Human Clariom D Assay TC0600008190.hg.1 ENST00000371211.6 Ensembl
Affymetrix Human Clariom D Assay TC0600008190.hg.1 ENST00000371211.6 Ensembl
Affymetrix Human Clariom D Assay TC0600008190.hg.1 ENST00000371211.6 Ensembl
Affymetrix Human Clariom D Assay TC0600008190.hg.1 ENST00000371211.6 Ensembl
Affymetrix Human Clariom D Assay TC0600008190.hg.1 ENST00000371211.6 Ensembl
Affymetrix Human Clariom D Assay TC0600008190.hg.1 ENST00000371211.6 Ensembl
Affymetrix Human Clariom D Assay TC0600008190.hg.1 ENST00000393699.2 Ensembl
Affymetrix Human Clariom D Assay TC0600008190.hg.1 ENST00000393699.2 Ensembl
Affymetrix Human Clariom D Assay TC0600008190.hg.1 ENST00000393699.2 Ensembl
Affymetrix Human Clariom D Assay TC0600008190.hg.1 ENST00000393699.2 Ensembl
Affymetrix Human Clariom D Assay TC0600008190.hg.1 ENST00000393699.2 Ensembl
Affymetrix Human Clariom D Assay TC0600008190.hg.1 ENST00000393699.2 Ensembl
Affymetrix Human Clariom D Assay TC0600008190.hg.1 ENST00000393699.2 Ensembl
Affymetrix Human Clariom D Assay TC0600008190.hg.1 ENST00000393699.2 Ensembl
Affymetrix Human Clariom D Assay TC0600008190.hg.1 ENST00000393699.2 Ensembl
Affymetrix Human Clariom D Assay TC0600008190.hg.1 ENST00000393699.2 Ensembl
Affymetrix Human Clariom D Assay TC0600008190.hg.1 ENST00000393699.2 Ensembl
Affymetrix Human Clariom D Assay TC0600008190.hg.1 ENST00000489301.6 Ensembl
Affymetrix Human Clariom D Assay TC0600008190.hg.1 ENST00000489301.6 Ensembl
Affymetrix Human Clariom D Assay TC0600008190.hg.1 ENST00000489301.6 Ensembl
Affymetrix Human Clariom D Assay TC0600008190.hg.1 ENST00000489301.6 Ensembl
Affymetrix Human Clariom D Assay TC0600008190.hg.1 ENST00000489301.6 Ensembl
Affymetrix Human Clariom D Assay TC0600008190.hg.1 ENST00000489301.6 Ensembl
Affymetrix Human Clariom D Assay TC0600008190.hg.1 ENST00000489301.6 Ensembl
Affymetrix Human Clariom D Assay TC0600008190.hg.1 ENST00000489301.6 Ensembl
Affymetrix Human Clariom D Assay TC0600008190.hg.1 ENST00000489301.6 Ensembl
Affymetrix Human Clariom D Assay TC0600008190.hg.1 ENST00000489301.6 Ensembl
Affymetrix Human Clariom D Assay TC0600008190.hg.1 ENST00000489301.6 Ensembl
Affymetrix Human Clariom D Assay TC0600008190.hg.1 ENST00000489301.6 Ensembl
Affymetrix Human Clariom D Assay TC0600008190.hg.1 ENST00000489301.6 Ensembl
Affymetrix Human Clariom D Assay TC0600008190.hg.1 ENST00000510695.1 Ensembl
Affymetrix Human Clariom D Assay TC0600008190.hg.1 ENST00000510695.1 Ensembl
Affymetrix Human Clariom D Assay TC0600008190.hg.1 ENST00000510695.1 Ensembl
Affymetrix Human Clariom D Assay TC0600008190.hg.1 ENST00000510695.1 Ensembl
Affymetrix Human Clariom D Assay TC0600008190.hg.1 NM_181744.3 Refseq
Affymetrix Human Clariom D Assay TC0600008190.hg.1 NM_181744.3 Refseq
Affymetrix Human Clariom D Assay TC0600008190.hg.1 NM_181744.3 Refseq
Affymetrix Human Clariom D Assay TC0600008190.hg.1 NM_181744.3 Refseq
Affymetrix Human Clariom D Assay TC0600008190.hg.1 NM_181744.3 Refseq
Affymetrix Human Clariom D Assay TC0600008190.hg.1 NM_181744.3 Refseq
Affymetrix Human Clariom D Assay TC0600008190.hg.1 NM_181744.3 Refseq
Affymetrix Human Clariom D Assay TC0600008190.hg.1 NM_181744.3 Refseq
Affymetrix Human Clariom D Assay TC0600008190.hg.1 NM_181744.3 Refseq
Affymetrix Human Clariom D Assay TC0600008190.hg.1 NM_181744.3 Refseq
Affymetrix Human Clariom D Assay TC0600008190.hg.1 NM_181744.3 Refseq
Affymetrix Human Clariom D Assay TC0600008190.hg.1 NM_181744.3 Refseq
Affymetrix Human Clariom D Assay TC0600008190.hg.1 NM_181744.3 Refseq
Affymetrix Human Clariom D Assay TC0600008190.hg.1 NM_181744.3 Refseq
Affymetrix Human Clariom D Assay TC0600008190.hg.1 NR_033806.1 Refseq
Affymetrix Human Clariom D Assay TC0600008190.hg.1 NR_033806.1 Refseq
Affymetrix Human Clariom D Assay TC0600008190.hg.1 NR_033806.1 Refseq
Affymetrix Human Clariom D Assay TC0600008190.hg.1 NR_033806.1 Refseq
Affymetrix Human Clariom D Assay TC0600008190.hg.1 NR_033806.1 Refseq
Affymetrix Human Clariom D Assay TC0600008190.hg.1 NR_033806.1 Refseq
Affymetrix Human Clariom D Assay TC0600008190.hg.1 NR_033806.1 Refseq
Affymetrix Human Clariom D Assay TC0600008190.hg.1 NR_033806.1 Refseq
Affymetrix Human Clariom D Assay TC0600008190.hg.1 NR_033806.1 Refseq
Affymetrix Human Clariom D Assay TC0600008190.hg.1 NR_033806.1 Refseq
Affymetrix Human Clariom D Assay TC0600008190.hg.1 NR_033806.1 Refseq
Affymetrix Human Clariom D Assay TC0600008190.hg.1 NR_033806.1 Refseq
Affymetrix Human Clariom D Assay TC0600008190.hg.1 NR_033806.1 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0