This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05028902_s1
See other SLC35A3 GE Assays ›
Gene Symbol:
SLC35A3 LOC730081
Gene Name:
solute carrier family 35 member A3
uncharacterized LOC730081
Gene Aliases:
AMRS
-
Chromosome Location:
Chr.1: 99969367 - 100026979 on Build GRCh38
Chromosome Location:
-
Species:
Human
Species Specific ID (Flybase ID):
-
-
Assay ID Hs05028902_s1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SLC35A3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005270691.4 XP_005270748.1 1 65 88
XM_017000869.1 XP_016856358.1 1 65 88
XM_017000870.1 XP_016856359.1 1 65 88
XM_017000871.1 XP_016856360.1 1 65 88
XM_017000872.1 XP_016856361.1 1 65 88
Gene Symbol: LOC730081
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK097502.1 - 1 88

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_005270691.4, XM_017000869.1, XM_017000870.1, XM_017000871.1, XM_017000872.1 transcripts.

Set Membership:

Amplicon length between 86 and 100 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

transporter

Gene Ontology Categories:

Function(s) Process(es)

UDP-N-acetylglucosamine metabolic process
carbohydrate transport
UDP-N-acetylglucosamine transport
proton transport
UDP-N-acetylglucosamine transmembrane transport
sugar:proton symporter activity
UDP-N-acetylglucosamine transmembrane transporter activity
protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0100015053.hg.1 ENST00000454219.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015053.hg.1 ENST00000454219.1 Ensembl
Affymetrix Human Clariom D Assay TC0100015053.hg.1 ENST00000454219.1 Ensembl
Affymetrix Human Clariom D Assay TC0100015053.hg.1 ENST00000454219.1 Ensembl

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0