This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05028115_s1
See other PDZD8 GE Assays ›
Gene Symbol:
PDZD8
Gene Name:
PDZ domain containing 8
Gene Aliases:
PDZK8
Chromosome Location:
Chr.10: 117280489 - 117375467 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05028115_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PDZD8
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_173791.4 NP_776152.1 4 1439 87
XM_005269518.3 XP_005269575.1 3 1289 87
GenBank mRNA AK058141.1 - 2 173 87
AK091746.1 - 4 1412 87
AY211929.1 - 4 508 87
BC028375.1 - 4 1401 4837939 87

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene PDZD8

Panther Classification:

Molecular Function -

scaffold/adaptor protein

Gene Ontology Categories:

Function(s) Process(es)

cytoskeleton organization
viral process
regulation of cell morphogenesis
intracellular signal transduction
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1000011948.hg.1 ENST00000334464.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011948.hg.1 ENST00000334464.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011948.hg.1 ENST00000334464.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011948.hg.1 ENST00000334464.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011948.hg.1 ENST00000334464.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011948.hg.1 ENST00000334464.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011948.hg.1 ENST00000334464.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011948.hg.1 ENST00000334464.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011948.hg.1 ENST00000334464.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011948.hg.1 ENST00000334464.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011948.hg.1 ENST00000334464.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011948.hg.1 ENST00000334464.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011948.hg.1 ENST00000334464.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011948.hg.1 ENST00000482496.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011948.hg.1 ENST00000482496.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011948.hg.1 ENST00000482496.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011948.hg.1 ENST00000482496.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011948.hg.1 ENST00000489302.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011948.hg.1 ENST00000489302.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011948.hg.1 ENST00000489302.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011948.hg.1 ENST00000489302.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011948.hg.1 ENST00000489491.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011948.hg.1 ENST00000489491.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011948.hg.1 NM_173791.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011948.hg.1 NM_173791.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011948.hg.1 NM_173791.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011948.hg.1 NM_173791.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011948.hg.1 NM_173791.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011948.hg.1 NM_173791.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011948.hg.1 NM_173791.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011948.hg.1 NM_173791.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011948.hg.1 NM_173791.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011948.hg.1 NM_173791.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011948.hg.1 NM_173791.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011948.hg.1 NM_173791.4 Refseq
Affymetrix Human Clariom D Assay TC1000011948.hg.1 ENST00000334464.6 Ensembl
Affymetrix Human Clariom D Assay TC1000011948.hg.1 ENST00000334464.6 Ensembl
Affymetrix Human Clariom D Assay TC1000011948.hg.1 ENST00000334464.6 Ensembl
Affymetrix Human Clariom D Assay TC1000011948.hg.1 ENST00000334464.6 Ensembl
Affymetrix Human Clariom D Assay TC1000011948.hg.1 ENST00000334464.6 Ensembl
Affymetrix Human Clariom D Assay TC1000011948.hg.1 ENST00000334464.6 Ensembl
Affymetrix Human Clariom D Assay TC1000011948.hg.1 ENST00000334464.6 Ensembl
Affymetrix Human Clariom D Assay TC1000011948.hg.1 ENST00000334464.6 Ensembl
Affymetrix Human Clariom D Assay TC1000011948.hg.1 ENST00000334464.6 Ensembl
Affymetrix Human Clariom D Assay TC1000011948.hg.1 ENST00000334464.6 Ensembl
Affymetrix Human Clariom D Assay TC1000011948.hg.1 ENST00000334464.6 Ensembl
Affymetrix Human Clariom D Assay TC1000011948.hg.1 ENST00000334464.6 Ensembl
Affymetrix Human Clariom D Assay TC1000011948.hg.1 ENST00000334464.6 Ensembl
Affymetrix Human Clariom D Assay TC1000011948.hg.1 ENST00000482496.5 Ensembl
Affymetrix Human Clariom D Assay TC1000011948.hg.1 ENST00000482496.5 Ensembl
Affymetrix Human Clariom D Assay TC1000011948.hg.1 ENST00000482496.5 Ensembl
Affymetrix Human Clariom D Assay TC1000011948.hg.1 ENST00000482496.5 Ensembl
Affymetrix Human Clariom D Assay TC1000011948.hg.1 ENST00000489302.5 Ensembl
Affymetrix Human Clariom D Assay TC1000011948.hg.1 ENST00000489302.5 Ensembl
Affymetrix Human Clariom D Assay TC1000011948.hg.1 ENST00000489302.5 Ensembl
Affymetrix Human Clariom D Assay TC1000011948.hg.1 ENST00000489302.5 Ensembl
Affymetrix Human Clariom D Assay TC1000011948.hg.1 ENST00000489491.1 Ensembl
Affymetrix Human Clariom D Assay TC1000011948.hg.1 ENST00000489491.1 Ensembl
Affymetrix Human Clariom D Assay TC1000011948.hg.1 NM_173791.4 Refseq
Affymetrix Human Clariom D Assay TC1000011948.hg.1 NM_173791.4 Refseq
Affymetrix Human Clariom D Assay TC1000011948.hg.1 NM_173791.4 Refseq
Affymetrix Human Clariom D Assay TC1000011948.hg.1 NM_173791.4 Refseq
Affymetrix Human Clariom D Assay TC1000011948.hg.1 NM_173791.4 Refseq
Affymetrix Human Clariom D Assay TC1000011948.hg.1 NM_173791.4 Refseq
Affymetrix Human Clariom D Assay TC1000011948.hg.1 NM_173791.4 Refseq
Affymetrix Human Clariom D Assay TC1000011948.hg.1 NM_173791.4 Refseq
Affymetrix Human Clariom D Assay TC1000011948.hg.1 NM_173791.4 Refseq
Affymetrix Human Clariom D Assay TC1000011948.hg.1 NM_173791.4 Refseq
Affymetrix Human Clariom D Assay TC1000011948.hg.1 NM_173791.4 Refseq
Affymetrix Human Clariom D Assay TC1000011948.hg.1 NM_173791.4 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0