This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05027668_s1
See other ALDH1L1-AS2 GE Assays ›
Gene Symbol:
ALDH1L1-AS2
Gene Name:
ALDH1L1 antisense RNA 2
Gene Aliases:
-
Chromosome Location:
Chr.3: 126180065 - 126210169 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05027668_s1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ALDH1L1-AS2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_046383.1 2 612 75
GenBank mRNA AK097508.1 - 4 523 75
BC039521.1 - 2 183 75

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Homo sapiens Both primers and probe map within a single exon

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0300008661.hg.1 ENST00000500232.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008661.hg.1 ENST00000500232.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008661.hg.1 ENST00000500232.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008661.hg.1 ENST00000500232.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008661.hg.1 ENST00000500232.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008661.hg.1 ENST00000500232.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008661.hg.1 ENST00000500525.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008661.hg.1 ENST00000500525.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008661.hg.1 ENST00000500525.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008661.hg.1 ENST00000500525.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008661.hg.1 ENST00000500525.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008661.hg.1 ENST00000500525.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008661.hg.1 ENST00000500525.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008661.hg.1 ENST00000502278.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008661.hg.1 ENST00000502278.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008661.hg.1 ENST00000502278.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008661.hg.1 NR_046383.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008661.hg.1 NR_046383.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008661.hg.1 NR_046383.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008661.hg.1 NR_046383.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008661.hg.1 NR_046383.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008661.hg.1 NR_046383.1 Refseq
Affymetrix Human Clariom D Assay TC0300008661.hg.1 ENST00000500232.2 Ensembl
Affymetrix Human Clariom D Assay TC0300008661.hg.1 ENST00000500232.2 Ensembl
Affymetrix Human Clariom D Assay TC0300008661.hg.1 ENST00000500232.2 Ensembl
Affymetrix Human Clariom D Assay TC0300008661.hg.1 ENST00000500232.2 Ensembl
Affymetrix Human Clariom D Assay TC0300008661.hg.1 ENST00000500232.2 Ensembl
Affymetrix Human Clariom D Assay TC0300008661.hg.1 ENST00000500232.2 Ensembl
Affymetrix Human Clariom D Assay TC0300008661.hg.1 ENST00000500525.1 Ensembl
Affymetrix Human Clariom D Assay TC0300008661.hg.1 ENST00000500525.1 Ensembl
Affymetrix Human Clariom D Assay TC0300008661.hg.1 ENST00000500525.1 Ensembl
Affymetrix Human Clariom D Assay TC0300008661.hg.1 ENST00000500525.1 Ensembl
Affymetrix Human Clariom D Assay TC0300008661.hg.1 ENST00000500525.1 Ensembl
Affymetrix Human Clariom D Assay TC0300008661.hg.1 ENST00000500525.1 Ensembl
Affymetrix Human Clariom D Assay TC0300008661.hg.1 ENST00000500525.1 Ensembl
Affymetrix Human Clariom D Assay TC0300008661.hg.1 ENST00000502278.1 Ensembl
Affymetrix Human Clariom D Assay TC0300008661.hg.1 ENST00000502278.1 Ensembl
Affymetrix Human Clariom D Assay TC0300008661.hg.1 ENST00000502278.1 Ensembl
Affymetrix Human Clariom D Assay TC0300008661.hg.1 NR_046383.1 Refseq
Affymetrix Human Clariom D Assay TC0300008661.hg.1 NR_046383.1 Refseq
Affymetrix Human Clariom D Assay TC0300008661.hg.1 NR_046383.1 Refseq
Affymetrix Human Clariom D Assay TC0300008661.hg.1 NR_046383.1 Refseq
Affymetrix Human Clariom D Assay TC0300008661.hg.1 NR_046383.1 Refseq
Affymetrix Human Clariom D Assay TC0300008661.hg.1 NR_046383.1 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0