Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008900.hg.1 |
ENST00000362494.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011641.hg.1 |
ENST00000516277.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012418.hg.1 |
ENST00000365355.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012418.hg.1 |
ENST00000365355.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008174.hg.1 |
ENST00000363916.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008174.hg.1 |
ENST00000363916.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008900.hg.1 |
ENST00000362494.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008174.hg.1 |
ENST00000363916.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008174.hg.1 |
ENST00000363916.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011641.hg.1 |
ENST00000516277.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012418.hg.1 |
ENST00000365355.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012418.hg.1 |
ENST00000365355.1 |
Ensembl |