This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05025720_m1
See other PDE3B GE Assays ›
Gene Symbol:
PDE3B
Gene Name:
phosphodiesterase 3B
Gene Aliases:
HcGIP1, cGIPDE1
Chromosome Location:
Chr.11: 14643691 - 14872059 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05025720_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PDE3B
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_000922.3 NP_000913.2 1 - 2 1327 67
XM_006718249.3 XP_006718312.1 1 - 2 1359 67
XM_017017911.1 XP_016873400.1 1 - 2 1359 67
XM_017017912.1 XP_016873401.1 1 - 2 1359 67
GenBank mRNA AB209326.1 - 1 - 2 790 67
AY459346.1 - 1 - 2 1265 67
BC136565.1 - 1 - 2 1220 9020555 67
BC136566.1 - 1 - 2 1220 9020556 67
BC144248.1 - 1 - 2 1220 9052768 67
BC150307.1 - 1 - 2 1327 8860429 67
X95520.1 - 1 - 2 974 67

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene PDE3B

Panther Classification:

Molecular Function -

phosphodiesterase

Gene Ontology Categories:

Function(s) Process(es)

angiogenesis
cAMP catabolic process
negative regulation of cell adhesion
signal transduction
negative regulation of angiogenesis
cellular response to insulin stimulus
negative regulation of cell adhesion mediated by integrin
negative regulation of cAMP-mediated signaling
negative regulation of lipid catabolic process
3',5'-cyclic-nucleotide phosphodiesterase activity
3',5'-cyclic-AMP phosphodiesterase activity
cGMP-inhibited cyclic-nucleotide phosphodiesterase activity
protein binding
protein kinase B binding
metal ion binding
3',5'-cyclic-GMP phosphodiesterase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1100006923.hg.1 ENST00000282096.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100006923.hg.1 ENST00000282096.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100006923.hg.1 ENST00000282096.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100006923.hg.1 ENST00000282096.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100006923.hg.1 ENST00000282096.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100006923.hg.1 ENST00000282096.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100006923.hg.1 ENST00000282096.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100006923.hg.1 ENST00000282096.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100006923.hg.1 ENST00000282096.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100006923.hg.1 ENST00000282096.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100006923.hg.1 ENST00000282096.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100006923.hg.1 ENST00000282096.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100006923.hg.1 ENST00000282096.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100006923.hg.1 ENST00000282096.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100006923.hg.1 ENST00000282096.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100006923.hg.1 ENST00000282096.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100006923.hg.1 ENST00000282096.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100006923.hg.1 ENST00000282096.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100006923.hg.1 ENST00000282096.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100006923.hg.1 ENST00000282096.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100006923.hg.1 ENST00000282096.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100006923.hg.1 ENST00000282096.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100006923.hg.1 ENST00000282096.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100006923.hg.1 ENST00000455098.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100006923.hg.1 ENST00000455098.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100006923.hg.1 ENST00000455098.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100006923.hg.1 ENST00000455098.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100006923.hg.1 ENST00000455098.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100006923.hg.1 ENST00000455098.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100006923.hg.1 ENST00000455098.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100006923.hg.1 ENST00000455098.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100006923.hg.1 ENST00000455098.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100006923.hg.1 ENST00000455098.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100006923.hg.1 ENST00000455098.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100006923.hg.1 ENST00000455098.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100006923.hg.1 ENST00000455098.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100006923.hg.1 ENST00000455098.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100006923.hg.1 ENST00000455098.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100006923.hg.1 ENST00000455098.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100006923.hg.1 ENST00000455098.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100006923.hg.1 ENST00000455098.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100006923.hg.1 ENST00000455098.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100006923.hg.1 ENST00000525439.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100006923.hg.1 ENST00000525439.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100006923.hg.1 ENST00000525439.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100006923.hg.1 ENST00000525439.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100006923.hg.1 ENST00000525439.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100006923.hg.1 ENST00000534317.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100006923.hg.1 ENST00000534317.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100006923.hg.1 ENST00000534317.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100006923.hg.1 NM_000922.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100006923.hg.1 NM_000922.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100006923.hg.1 NM_000922.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100006923.hg.1 NM_000922.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100006923.hg.1 NM_000922.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100006923.hg.1 NM_000922.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100006923.hg.1 NM_000922.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100006923.hg.1 NM_000922.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100006923.hg.1 NM_000922.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100006923.hg.1 NM_000922.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100006923.hg.1 NM_000922.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100006923.hg.1 NM_000922.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100006923.hg.1 NM_000922.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100006923.hg.1 NM_000922.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100006923.hg.1 NM_000922.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100006923.hg.1 NM_000922.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100006923.hg.1 NM_000922.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100006923.hg.1 NM_000922.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100006923.hg.1 NM_000922.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100006923.hg.1 NM_000922.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100006923.hg.1 NM_000922.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100006923.hg.1 NM_000922.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100006923.hg.1 NM_000922.3 Refseq
Affymetrix Human Clariom D Assay TC1100006923.hg.1 ENST00000282096.8 Ensembl
Affymetrix Human Clariom D Assay TC1100006923.hg.1 ENST00000282096.8 Ensembl
Affymetrix Human Clariom D Assay TC1100006923.hg.1 ENST00000282096.8 Ensembl
Affymetrix Human Clariom D Assay TC1100006923.hg.1 ENST00000282096.8 Ensembl
Affymetrix Human Clariom D Assay TC1100006923.hg.1 ENST00000282096.8 Ensembl
Affymetrix Human Clariom D Assay TC1100006923.hg.1 ENST00000282096.8 Ensembl
Affymetrix Human Clariom D Assay TC1100006923.hg.1 ENST00000282096.8 Ensembl
Affymetrix Human Clariom D Assay TC1100006923.hg.1 ENST00000282096.8 Ensembl
Affymetrix Human Clariom D Assay TC1100006923.hg.1 ENST00000282096.8 Ensembl
Affymetrix Human Clariom D Assay TC1100006923.hg.1 ENST00000282096.8 Ensembl
Affymetrix Human Clariom D Assay TC1100006923.hg.1 ENST00000282096.8 Ensembl
Affymetrix Human Clariom D Assay TC1100006923.hg.1 ENST00000282096.8 Ensembl
Affymetrix Human Clariom D Assay TC1100006923.hg.1 ENST00000282096.8 Ensembl
Affymetrix Human Clariom D Assay TC1100006923.hg.1 ENST00000282096.8 Ensembl
Affymetrix Human Clariom D Assay TC1100006923.hg.1 ENST00000282096.8 Ensembl
Affymetrix Human Clariom D Assay TC1100006923.hg.1 ENST00000282096.8 Ensembl
Affymetrix Human Clariom D Assay TC1100006923.hg.1 ENST00000282096.8 Ensembl
Affymetrix Human Clariom D Assay TC1100006923.hg.1 ENST00000282096.8 Ensembl
Affymetrix Human Clariom D Assay TC1100006923.hg.1 ENST00000282096.8 Ensembl
Affymetrix Human Clariom D Assay TC1100006923.hg.1 ENST00000282096.8 Ensembl
Affymetrix Human Clariom D Assay TC1100006923.hg.1 ENST00000282096.8 Ensembl
Affymetrix Human Clariom D Assay TC1100006923.hg.1 ENST00000282096.8 Ensembl
Affymetrix Human Clariom D Assay TC1100006923.hg.1 ENST00000282096.8 Ensembl
Affymetrix Human Clariom D Assay TC1100006923.hg.1 ENST00000455098.2 Ensembl
Affymetrix Human Clariom D Assay TC1100006923.hg.1 ENST00000455098.2 Ensembl
Affymetrix Human Clariom D Assay TC1100006923.hg.1 ENST00000455098.2 Ensembl
Affymetrix Human Clariom D Assay TC1100006923.hg.1 ENST00000455098.2 Ensembl
Affymetrix Human Clariom D Assay TC1100006923.hg.1 ENST00000455098.2 Ensembl
Affymetrix Human Clariom D Assay TC1100006923.hg.1 ENST00000455098.2 Ensembl
Affymetrix Human Clariom D Assay TC1100006923.hg.1 ENST00000455098.2 Ensembl
Affymetrix Human Clariom D Assay TC1100006923.hg.1 ENST00000455098.2 Ensembl
Affymetrix Human Clariom D Assay TC1100006923.hg.1 ENST00000455098.2 Ensembl
Affymetrix Human Clariom D Assay TC1100006923.hg.1 ENST00000455098.2 Ensembl
Affymetrix Human Clariom D Assay TC1100006923.hg.1 ENST00000455098.2 Ensembl
Affymetrix Human Clariom D Assay TC1100006923.hg.1 ENST00000455098.2 Ensembl
Affymetrix Human Clariom D Assay TC1100006923.hg.1 ENST00000455098.2 Ensembl
Affymetrix Human Clariom D Assay TC1100006923.hg.1 ENST00000455098.2 Ensembl
Affymetrix Human Clariom D Assay TC1100006923.hg.1 ENST00000455098.2 Ensembl
Affymetrix Human Clariom D Assay TC1100006923.hg.1 ENST00000455098.2 Ensembl
Affymetrix Human Clariom D Assay TC1100006923.hg.1 ENST00000455098.2 Ensembl
Affymetrix Human Clariom D Assay TC1100006923.hg.1 ENST00000455098.2 Ensembl
Affymetrix Human Clariom D Assay TC1100006923.hg.1 ENST00000455098.2 Ensembl
Affymetrix Human Clariom D Assay TC1100006923.hg.1 ENST00000525439.1 Ensembl
Affymetrix Human Clariom D Assay TC1100006923.hg.1 ENST00000525439.1 Ensembl
Affymetrix Human Clariom D Assay TC1100006923.hg.1 ENST00000525439.1 Ensembl
Affymetrix Human Clariom D Assay TC1100006923.hg.1 ENST00000525439.1 Ensembl
Affymetrix Human Clariom D Assay TC1100006923.hg.1 ENST00000525439.1 Ensembl
Affymetrix Human Clariom D Assay TC1100006923.hg.1 ENST00000534317.1 Ensembl
Affymetrix Human Clariom D Assay TC1100006923.hg.1 ENST00000534317.1 Ensembl
Affymetrix Human Clariom D Assay TC1100006923.hg.1 ENST00000534317.1 Ensembl
Affymetrix Human Clariom D Assay TC1100006923.hg.1 NM_000922.3 Refseq
Affymetrix Human Clariom D Assay TC1100006923.hg.1 NM_000922.3 Refseq
Affymetrix Human Clariom D Assay TC1100006923.hg.1 NM_000922.3 Refseq
Affymetrix Human Clariom D Assay TC1100006923.hg.1 NM_000922.3 Refseq
Affymetrix Human Clariom D Assay TC1100006923.hg.1 NM_000922.3 Refseq
Affymetrix Human Clariom D Assay TC1100006923.hg.1 NM_000922.3 Refseq
Affymetrix Human Clariom D Assay TC1100006923.hg.1 NM_000922.3 Refseq
Affymetrix Human Clariom D Assay TC1100006923.hg.1 NM_000922.3 Refseq
Affymetrix Human Clariom D Assay TC1100006923.hg.1 NM_000922.3 Refseq
Affymetrix Human Clariom D Assay TC1100006923.hg.1 NM_000922.3 Refseq
Affymetrix Human Clariom D Assay TC1100006923.hg.1 NM_000922.3 Refseq
Affymetrix Human Clariom D Assay TC1100006923.hg.1 NM_000922.3 Refseq
Affymetrix Human Clariom D Assay TC1100006923.hg.1 NM_000922.3 Refseq
Affymetrix Human Clariom D Assay TC1100006923.hg.1 NM_000922.3 Refseq
Affymetrix Human Clariom D Assay TC1100006923.hg.1 NM_000922.3 Refseq
Affymetrix Human Clariom D Assay TC1100006923.hg.1 NM_000922.3 Refseq
Affymetrix Human Clariom D Assay TC1100006923.hg.1 NM_000922.3 Refseq
Affymetrix Human Clariom D Assay TC1100006923.hg.1 NM_000922.3 Refseq
Affymetrix Human Clariom D Assay TC1100006923.hg.1 NM_000922.3 Refseq
Affymetrix Human Clariom D Assay TC1100006923.hg.1 NM_000922.3 Refseq
Affymetrix Human Clariom D Assay TC1100006923.hg.1 NM_000922.3 Refseq
Affymetrix Human Clariom D Assay TC1100006923.hg.1 NM_000922.3 Refseq
Affymetrix Human Clariom D Assay TC1100006923.hg.1 NM_000922.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0