This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05024103_m1
See other HSD17B7P2 GE Assays ›
Gene Symbol:
HSD17B7P2
Gene Name:
hydroxysteroid 17-beta dehydrogenase 7 pseudogene 2
Gene Aliases:
HSD17B7, Hsd17b_2, bA291L22.1
Chromosome Location:
Chr.10: 38356380 - 38378505 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05024103_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: HSD17B7P2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AF165514.1 - 2 - 3 309 95
AK307174.1 - 2 - 3 299 95

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Homo sapiens Probe spans exons

Panther Classification:

Molecular Function -

reductase

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0100010389.hg.1 ENST00000254521.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010389.hg.1 ENST00000254521.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010389.hg.1 ENST00000254521.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010389.hg.1 ENST00000254521.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010389.hg.1 ENST00000254521.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010389.hg.1 ENST00000254521.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010389.hg.1 ENST00000254521.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010389.hg.1 ENST00000254521.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010389.hg.1 ENST00000254521.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010389.hg.1 ENST00000254521.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010389.hg.1 ENST00000254521.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010389.hg.1 ENST00000254521.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010389.hg.1 ENST00000254521.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010389.hg.1 ENST00000367913.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010389.hg.1 ENST00000367913.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010389.hg.1 ENST00000367913.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010389.hg.1 ENST00000367913.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010389.hg.1 ENST00000367913.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010389.hg.1 ENST00000367915.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010389.hg.1 ENST00000367915.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010389.hg.1 ENST00000367915.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010389.hg.1 ENST00000367915.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010389.hg.1 ENST00000463037.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010389.hg.1 ENST00000463037.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010389.hg.1 ENST00000463037.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010389.hg.1 ENST00000463037.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010389.hg.1 ENST00000463037.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010389.hg.1 ENST00000463037.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010389.hg.1 ENST00000463037.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010389.hg.1 ENST00000463037.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010389.hg.1 ENST00000463037.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010389.hg.1 ENST00000463037.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010389.hg.1 ENST00000466176.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010389.hg.1 ENST00000466176.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010389.hg.1 ENST00000466176.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010389.hg.1 ENST00000466176.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010389.hg.1 ENST00000466176.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010389.hg.1 ENST00000466176.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010389.hg.1 ENST00000466176.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010389.hg.1 ENST00000466176.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010389.hg.1 ENST00000470195.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010389.hg.1 ENST00000470195.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010389.hg.1 ENST00000470195.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010389.hg.1 ENST00000470195.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010389.hg.1 ENST00000470195.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010389.hg.1 ENST00000470195.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010389.hg.1 ENST00000484251.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010389.hg.1 ENST00000484251.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010389.hg.1 ENST00000484251.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010389.hg.1 ENST00000484251.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010389.hg.1 ENST00000484251.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010389.hg.1 ENST00000485405.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010389.hg.1 ENST00000485405.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010389.hg.1 ENST00000485405.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010389.hg.1 ENST00000485405.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010389.hg.1 ENST00000485405.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010389.hg.1 ENST00000485405.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010389.hg.1 ENST00000485405.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010389.hg.1 ENST00000485405.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010389.hg.1 ENST00000485405.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010389.hg.1 ENST00000485405.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010389.hg.1 ENST00000485405.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010389.hg.1 ENST00000485405.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010389.hg.1 ENST00000485405.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010389.hg.1 ENST00000485405.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010389.hg.1 ENST00000485405.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010389.hg.1 ENST00000485405.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010389.hg.1 ENST00000488656.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010389.hg.1 ENST00000488656.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010389.hg.1 ENST00000488656.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010389.hg.1 ENST00000494450.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010389.hg.1 ENST00000494450.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010389.hg.1 ENST00000494450.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010389.hg.1 ENST00000494450.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010389.hg.1 ENST00000494450.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010389.hg.1 NM_001304512.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100010389.hg.1 NM_001304512.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100010389.hg.1 NM_001304512.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100010389.hg.1 NM_001304512.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100010389.hg.1 NM_001304512.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100010389.hg.1 NM_001304512.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100010389.hg.1 NM_001304512.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100010389.hg.1 NM_001304513.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100010389.hg.1 NM_001304513.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100010389.hg.1 NM_001304513.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100010389.hg.1 NM_001304513.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100010389.hg.1 NM_001304513.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100010389.hg.1 NM_001304513.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100010389.hg.1 NM_016371.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100010389.hg.1 NM_016371.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100010389.hg.1 NM_016371.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100010389.hg.1 NM_016371.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100010389.hg.1 NM_016371.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100010389.hg.1 NM_016371.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100010389.hg.1 NM_016371.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100010389.hg.1 NM_016371.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100010389.hg.1 NM_016371.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100010389.hg.1 NM_016371.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100010389.hg.1 NM_016371.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100010389.hg.1 NM_016371.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100010389.hg.1 NM_016371.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100010389.hg.1 NM_016371.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000007346.hg.1 ENST00000471365.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007346.hg.1 ENST00000471365.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007346.hg.1 ENST00000471365.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007346.hg.1 ENST00000471365.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007346.hg.1 ENST00000471365.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007346.hg.1 ENST00000471365.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007346.hg.1 ENST00000471365.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007346.hg.1 ENST00000471365.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007346.hg.1 ENST00000471365.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007346.hg.1 ENST00000494540.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007346.hg.1 ENST00000494540.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007346.hg.1 ENST00000494540.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007346.hg.1 ENST00000494540.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007346.hg.1 ENST00000494540.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007346.hg.1 ENST00000494540.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007346.hg.1 ENST00000494540.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007346.hg.1 ENST00000494540.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007346.hg.1 ENST00000494540.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007346.hg.1 ENST00000494540.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007346.hg.1 NR_003086.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000007346.hg.1 NR_003086.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000007346.hg.1 NR_003086.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000007346.hg.1 NR_003086.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000007346.hg.1 NR_003086.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000007346.hg.1 NR_003086.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000007346.hg.1 NR_003086.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000007346.hg.1 NR_003086.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000007346.hg.1 NR_003086.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000007346.hg.1 NR_003086.1 Refseq
Affymetrix Human Clariom D Assay TC1000007346.hg.1 ENST00000471365.1 Ensembl
Affymetrix Human Clariom D Assay TC1000007346.hg.1 ENST00000471365.1 Ensembl
Affymetrix Human Clariom D Assay TC1000007346.hg.1 ENST00000471365.1 Ensembl
Affymetrix Human Clariom D Assay TC1000007346.hg.1 ENST00000471365.1 Ensembl
Affymetrix Human Clariom D Assay TC1000007346.hg.1 ENST00000471365.1 Ensembl
Affymetrix Human Clariom D Assay TC1000007346.hg.1 ENST00000471365.1 Ensembl
Affymetrix Human Clariom D Assay TC1000007346.hg.1 ENST00000471365.1 Ensembl
Affymetrix Human Clariom D Assay TC1000007346.hg.1 ENST00000471365.1 Ensembl
Affymetrix Human Clariom D Assay TC1000007346.hg.1 ENST00000471365.1 Ensembl
Affymetrix Human Clariom D Assay TC1000007346.hg.1 ENST00000494540.5 Ensembl
Affymetrix Human Clariom D Assay TC1000007346.hg.1 ENST00000494540.5 Ensembl
Affymetrix Human Clariom D Assay TC1000007346.hg.1 ENST00000494540.5 Ensembl
Affymetrix Human Clariom D Assay TC1000007346.hg.1 ENST00000494540.5 Ensembl
Affymetrix Human Clariom D Assay TC1000007346.hg.1 ENST00000494540.5 Ensembl
Affymetrix Human Clariom D Assay TC1000007346.hg.1 ENST00000494540.5 Ensembl
Affymetrix Human Clariom D Assay TC1000007346.hg.1 ENST00000494540.5 Ensembl
Affymetrix Human Clariom D Assay TC1000007346.hg.1 ENST00000494540.5 Ensembl
Affymetrix Human Clariom D Assay TC1000007346.hg.1 ENST00000494540.5 Ensembl
Affymetrix Human Clariom D Assay TC1000007346.hg.1 ENST00000494540.5 Ensembl
Affymetrix Human Clariom D Assay TC1000007346.hg.1 NR_003086.1 Refseq
Affymetrix Human Clariom D Assay TC1000007346.hg.1 NR_003086.1 Refseq
Affymetrix Human Clariom D Assay TC1000007346.hg.1 NR_003086.1 Refseq
Affymetrix Human Clariom D Assay TC1000007346.hg.1 NR_003086.1 Refseq
Affymetrix Human Clariom D Assay TC1000007346.hg.1 NR_003086.1 Refseq
Affymetrix Human Clariom D Assay TC1000007346.hg.1 NR_003086.1 Refseq
Affymetrix Human Clariom D Assay TC1000007346.hg.1 NR_003086.1 Refseq
Affymetrix Human Clariom D Assay TC1000007346.hg.1 NR_003086.1 Refseq
Affymetrix Human Clariom D Assay TC1000007346.hg.1 NR_003086.1 Refseq
Affymetrix Human Clariom D Assay TC1000007346.hg.1 NR_003086.1 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0