This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05023237_g1
See other MYH7B GE Assays ›
Gene Symbol:
MYH7B
Gene Name:
myosin heavy chain 7B
Gene Aliases:
MHC14, MYH14
Chromosome Location:
Chr.20: 34955835 - 35002437 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05023237_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MYH7B
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_020884.4 NP_065935.3 42 - 43 6086 113
XM_006723840.3 XP_006723903.1 40 - 41 5727 113
XM_011528941.2 XP_011527243.1 40 - 41 5751 113
XM_011528947.2 XP_011527249.1 34 - 35 6124 113
XM_011528948.2 XP_011527250.1 34 - 35 5283 113
XM_017027986.1 XP_016883475.1 41 - 42 5787 113
XM_017027987.1 XP_016883476.1 36 - 37 5302 113
GenBank mRNA AB040945.2 - 40 - 41 5795 113
BC007808.1 - 15 - 16 2773 113
BC151242.1 - 40 - 41 5807 8860515 113
BG189104.1 - 1 - 2 107 114

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene MYH7B

Gene Ontology Categories:

Process(es)

motor activity
actin binding
protein binding
ATP binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0