This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05022708_s1
See other CHML GE Assays ›
Gene Symbol:
CHML
Gene Name:
CHM like, Rab escort protein 2
Gene Aliases:
REP2
Chromosome Location:
Chr.1: 241628864 - 241635930 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05022708_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CHML
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001821.3 NP_001812.2 1 6123 103
GenBank mRNA AK000933.1 - 1 1615 103
AK023423.1 - 1 1127 103
BU187416.1 - 1 530 103
BU195066.1 - 1 242 103

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene CHML

Panther Classification:

Molecular Function -

G-protein modulator protein-binding activity modulator

Gene Ontology Categories:

Function(s) Process(es)

intracellular protein transport
small GTPase mediated signal transduction
protein geranylgeranylation
positive regulation of GTPase activity
oxidation-reduction process
GDP-dissociation inhibitor activity
GTPase activator activity
oxidoreductase activity
Rab GTPase binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0100017948.hg.1 ENST00000366553.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017948.hg.1 ENST00000366553.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017948.hg.1 ENST00000366553.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017948.hg.1 ENST00000366553.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017948.hg.1 ENST00000366553.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017948.hg.1 ENST00000366553.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017948.hg.1 NM_001821.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100017948.hg.1 NM_001821.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100017948.hg.1 NM_001821.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100017948.hg.1 NM_001821.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100017948.hg.1 NM_001821.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100017948.hg.1 NM_001821.3 Refseq
Affymetrix Human Clariom D Assay TC0100017948.hg.1 ENST00000366553.1 Ensembl
Affymetrix Human Clariom D Assay TC0100017948.hg.1 ENST00000366553.1 Ensembl
Affymetrix Human Clariom D Assay TC0100017948.hg.1 ENST00000366553.1 Ensembl
Affymetrix Human Clariom D Assay TC0100017948.hg.1 ENST00000366553.1 Ensembl
Affymetrix Human Clariom D Assay TC0100017948.hg.1 ENST00000366553.1 Ensembl
Affymetrix Human Clariom D Assay TC0100017948.hg.1 ENST00000366553.1 Ensembl
Affymetrix Human Clariom D Assay TC0100017948.hg.1 NM_001821.3 Refseq
Affymetrix Human Clariom D Assay TC0100017948.hg.1 NM_001821.3 Refseq
Affymetrix Human Clariom D Assay TC0100017948.hg.1 NM_001821.3 Refseq
Affymetrix Human Clariom D Assay TC0100017948.hg.1 NM_001821.3 Refseq
Affymetrix Human Clariom D Assay TC0100017948.hg.1 NM_001821.3 Refseq
Affymetrix Human Clariom D Assay TC0100017948.hg.1 NM_001821.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0