This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05021700_m1
See other LINC01141 GE Assays ›
Gene Symbol:
LINC01141
Gene Name:
long intergenic non-protein coding RNA 1141
Gene Aliases:
-
Chromosome Location:
Chr.1: 20359801 - 20428794 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05021700_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LINC01141
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_033887.1 10 - 11 1289 90
GenBank mRNA BC040588.1 - 10 - 11 1277 90
BC131526.1 - 2 - 3 270 90

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0100013177.hg.1 ENST00000418743.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013177.hg.1 ENST00000418743.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013177.hg.1 ENST00000418743.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013177.hg.1 ENST00000418743.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013177.hg.1 ENST00000418743.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013177.hg.1 ENST00000418743.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013177.hg.1 ENST00000418743.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013177.hg.1 ENST00000418743.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013177.hg.1 ENST00000418743.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013177.hg.1 ENST00000418743.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013177.hg.1 ENST00000418743.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013177.hg.1 ENST00000418743.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013177.hg.1 ENST00000418743.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013177.hg.1 ENST00000418743.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013177.hg.1 ENST00000418743.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013177.hg.1 ENST00000418743.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013177.hg.1 ENST00000423486.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013177.hg.1 ENST00000423486.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013177.hg.1 ENST00000423486.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013177.hg.1 ENST00000423486.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013177.hg.1 ENST00000423486.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013177.hg.1 ENST00000426428.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013177.hg.1 ENST00000426428.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013177.hg.1 ENST00000426428.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013177.hg.1 ENST00000426428.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013177.hg.1 ENST00000426428.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013177.hg.1 ENST00000426428.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013177.hg.1 ENST00000426428.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013177.hg.1 ENST00000426428.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013177.hg.1 ENST00000426428.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013177.hg.1 ENST00000426428.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013177.hg.1 ENST00000426428.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013177.hg.1 ENST00000426428.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013177.hg.1 ENST00000426428.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013177.hg.1 ENST00000426428.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013177.hg.1 ENST00000426428.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013177.hg.1 ENST00000426428.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013177.hg.1 ENST00000426428.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013177.hg.1 ENST00000426428.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013177.hg.1 ENST00000426428.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013177.hg.1 ENST00000426428.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013177.hg.1 ENST00000426428.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013177.hg.1 ENST00000426428.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013177.hg.1 ENST00000426428.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013177.hg.1 ENST00000426428.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013177.hg.1 ENST00000426428.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013177.hg.1 ENST00000426428.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013177.hg.1 ENST00000426428.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013177.hg.1 ENST00000426428.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013177.hg.1 ENST00000426428.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013177.hg.1 ENST00000426428.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013177.hg.1 ENST00000426428.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013177.hg.1 ENST00000426428.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013177.hg.1 ENST00000426428.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013177.hg.1 ENST00000426428.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013177.hg.1 ENST00000426428.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013177.hg.1 ENST00000426428.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100013177.hg.1 NR_033887.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100013177.hg.1 NR_033887.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100013177.hg.1 NR_033887.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100013177.hg.1 NR_033887.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100013177.hg.1 NR_033887.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100013177.hg.1 NR_033887.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100013177.hg.1 NR_033887.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100013177.hg.1 NR_033887.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100013177.hg.1 NR_033887.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100013177.hg.1 NR_033887.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100013177.hg.1 NR_033887.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100013177.hg.1 NR_033887.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100013177.hg.1 NR_033887.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100013177.hg.1 NR_033887.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100013177.hg.1 NR_033887.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100013177.hg.1 NR_033887.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100013177.hg.1 NR_033887.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100013177.hg.1 NR_033887.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100013177.hg.1 NR_033887.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100013177.hg.1 NR_033887.1 Refseq
Affymetrix Human Clariom D Assay TC0100013177.hg.1 ENST00000418743.1 Ensembl
Affymetrix Human Clariom D Assay TC0100013177.hg.1 ENST00000418743.1 Ensembl
Affymetrix Human Clariom D Assay TC0100013177.hg.1 ENST00000418743.1 Ensembl
Affymetrix Human Clariom D Assay TC0100013177.hg.1 ENST00000418743.1 Ensembl
Affymetrix Human Clariom D Assay TC0100013177.hg.1 ENST00000418743.1 Ensembl
Affymetrix Human Clariom D Assay TC0100013177.hg.1 ENST00000418743.1 Ensembl
Affymetrix Human Clariom D Assay TC0100013177.hg.1 ENST00000418743.1 Ensembl
Affymetrix Human Clariom D Assay TC0100013177.hg.1 ENST00000418743.1 Ensembl
Affymetrix Human Clariom D Assay TC0100013177.hg.1 ENST00000418743.1 Ensembl
Affymetrix Human Clariom D Assay TC0100013177.hg.1 ENST00000418743.1 Ensembl
Affymetrix Human Clariom D Assay TC0100013177.hg.1 ENST00000418743.1 Ensembl
Affymetrix Human Clariom D Assay TC0100013177.hg.1 ENST00000418743.1 Ensembl
Affymetrix Human Clariom D Assay TC0100013177.hg.1 ENST00000418743.1 Ensembl
Affymetrix Human Clariom D Assay TC0100013177.hg.1 ENST00000418743.1 Ensembl
Affymetrix Human Clariom D Assay TC0100013177.hg.1 ENST00000418743.1 Ensembl
Affymetrix Human Clariom D Assay TC0100013177.hg.1 ENST00000418743.1 Ensembl
Affymetrix Human Clariom D Assay TC0100013177.hg.1 ENST00000423486.1 Ensembl
Affymetrix Human Clariom D Assay TC0100013177.hg.1 ENST00000423486.1 Ensembl
Affymetrix Human Clariom D Assay TC0100013177.hg.1 ENST00000423486.1 Ensembl
Affymetrix Human Clariom D Assay TC0100013177.hg.1 ENST00000423486.1 Ensembl
Affymetrix Human Clariom D Assay TC0100013177.hg.1 ENST00000423486.1 Ensembl
Affymetrix Human Clariom D Assay TC0100013177.hg.1 ENST00000426428.1 Ensembl
Affymetrix Human Clariom D Assay TC0100013177.hg.1 ENST00000426428.1 Ensembl
Affymetrix Human Clariom D Assay TC0100013177.hg.1 ENST00000426428.1 Ensembl
Affymetrix Human Clariom D Assay TC0100013177.hg.1 ENST00000426428.1 Ensembl
Affymetrix Human Clariom D Assay TC0100013177.hg.1 ENST00000426428.1 Ensembl
Affymetrix Human Clariom D Assay TC0100013177.hg.1 ENST00000426428.1 Ensembl
Affymetrix Human Clariom D Assay TC0100013177.hg.1 ENST00000426428.1 Ensembl
Affymetrix Human Clariom D Assay TC0100013177.hg.1 ENST00000426428.1 Ensembl
Affymetrix Human Clariom D Assay TC0100013177.hg.1 ENST00000426428.1 Ensembl
Affymetrix Human Clariom D Assay TC0100013177.hg.1 ENST00000426428.1 Ensembl
Affymetrix Human Clariom D Assay TC0100013177.hg.1 ENST00000426428.1 Ensembl
Affymetrix Human Clariom D Assay TC0100013177.hg.1 ENST00000426428.1 Ensembl
Affymetrix Human Clariom D Assay TC0100013177.hg.1 ENST00000426428.1 Ensembl
Affymetrix Human Clariom D Assay TC0100013177.hg.1 ENST00000426428.1 Ensembl
Affymetrix Human Clariom D Assay TC0100013177.hg.1 ENST00000426428.1 Ensembl
Affymetrix Human Clariom D Assay TC0100013177.hg.1 ENST00000426428.1 Ensembl
Affymetrix Human Clariom D Assay TC0100013177.hg.1 ENST00000426428.1 Ensembl
Affymetrix Human Clariom D Assay TC0100013177.hg.1 ENST00000426428.1 Ensembl
Affymetrix Human Clariom D Assay TC0100013177.hg.1 ENST00000426428.1 Ensembl
Affymetrix Human Clariom D Assay TC0100013177.hg.1 ENST00000426428.1 Ensembl
Affymetrix Human Clariom D Assay TC0100013177.hg.1 ENST00000426428.1 Ensembl
Affymetrix Human Clariom D Assay TC0100013177.hg.1 ENST00000426428.1 Ensembl
Affymetrix Human Clariom D Assay TC0100013177.hg.1 ENST00000426428.1 Ensembl
Affymetrix Human Clariom D Assay TC0100013177.hg.1 ENST00000426428.1 Ensembl
Affymetrix Human Clariom D Assay TC0100013177.hg.1 ENST00000426428.1 Ensembl
Affymetrix Human Clariom D Assay TC0100013177.hg.1 ENST00000426428.1 Ensembl
Affymetrix Human Clariom D Assay TC0100013177.hg.1 ENST00000426428.1 Ensembl
Affymetrix Human Clariom D Assay TC0100013177.hg.1 ENST00000426428.1 Ensembl
Affymetrix Human Clariom D Assay TC0100013177.hg.1 ENST00000426428.1 Ensembl
Affymetrix Human Clariom D Assay TC0100013177.hg.1 ENST00000426428.1 Ensembl
Affymetrix Human Clariom D Assay TC0100013177.hg.1 ENST00000426428.1 Ensembl
Affymetrix Human Clariom D Assay TC0100013177.hg.1 ENST00000426428.1 Ensembl
Affymetrix Human Clariom D Assay TC0100013177.hg.1 ENST00000426428.1 Ensembl
Affymetrix Human Clariom D Assay TC0100013177.hg.1 ENST00000426428.1 Ensembl
Affymetrix Human Clariom D Assay TC0100013177.hg.1 ENST00000426428.1 Ensembl
Affymetrix Human Clariom D Assay TC0100013177.hg.1 ENST00000426428.1 Ensembl
Affymetrix Human Clariom D Assay TC0100013177.hg.1 NR_033887.1 Refseq
Affymetrix Human Clariom D Assay TC0100013177.hg.1 NR_033887.1 Refseq
Affymetrix Human Clariom D Assay TC0100013177.hg.1 NR_033887.1 Refseq
Affymetrix Human Clariom D Assay TC0100013177.hg.1 NR_033887.1 Refseq
Affymetrix Human Clariom D Assay TC0100013177.hg.1 NR_033887.1 Refseq
Affymetrix Human Clariom D Assay TC0100013177.hg.1 NR_033887.1 Refseq
Affymetrix Human Clariom D Assay TC0100013177.hg.1 NR_033887.1 Refseq
Affymetrix Human Clariom D Assay TC0100013177.hg.1 NR_033887.1 Refseq
Affymetrix Human Clariom D Assay TC0100013177.hg.1 NR_033887.1 Refseq
Affymetrix Human Clariom D Assay TC0100013177.hg.1 NR_033887.1 Refseq
Affymetrix Human Clariom D Assay TC0100013177.hg.1 NR_033887.1 Refseq
Affymetrix Human Clariom D Assay TC0100013177.hg.1 NR_033887.1 Refseq
Affymetrix Human Clariom D Assay TC0100013177.hg.1 NR_033887.1 Refseq
Affymetrix Human Clariom D Assay TC0100013177.hg.1 NR_033887.1 Refseq
Affymetrix Human Clariom D Assay TC0100013177.hg.1 NR_033887.1 Refseq
Affymetrix Human Clariom D Assay TC0100013177.hg.1 NR_033887.1 Refseq
Affymetrix Human Clariom D Assay TC0100013177.hg.1 NR_033887.1 Refseq
Affymetrix Human Clariom D Assay TC0100013177.hg.1 NR_033887.1 Refseq
Affymetrix Human Clariom D Assay TC0100013177.hg.1 NR_033887.1 Refseq
Affymetrix Human Clariom D Assay TC0100013177.hg.1 NR_033887.1 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0