This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05019860_g1
See other RPL22 GE Assays ›
Gene Symbol:
RPL22
Gene Name:
ribosomal protein L22
Gene Aliases:
EAP, HBP15, HBP15/L22, L22
Chromosome Location:
Chr.1: 6185020 - 6199619 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05019860_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: RPL22
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_000983.3 NP_000974.1 2 - 3 163 74
GenBank mRNA AK097075.1 - 2 - 3 139 74
AK311749.1 - 2 - 3 137 74
BC035566.1 - 2 - 3 163 5109124 74
BC058887.1 - 2 - 3 121 6141451 74
BC066314.1 - 2 - 3 139 5303658 74
BU927988.1 - 177 74
CA336134.1 - 2 - 3 166 74
CR456873.1 - 2 - 3 117 74
D17652.1 - 2 - 3 139 74
X59357.1 - 1 - 2 168 74

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene RPL22

Panther Classification:

Molecular Function -

ribosomal protein

Gene Ontology Categories:

Function(s) Process(es)

nuclear-transcribed mRNA catabolic process, nonsense-mediated decay
cytoplasmic translation
rRNA processing
translation
translational initiation
SRP-dependent cotranslational protein targeting to membrane
viral transcription
alpha-beta T cell differentiation
RNA binding
structural constituent of ribosome
protein binding
heparin binding
poly(A) RNA binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0