This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05015380_g1
See other EIF1 GE Assays ›
Gene Symbol:
EIF1
Gene Name:
eukaryotic translation initiation factor 1
Gene Aliases:
A121, EIF-1, EIF1A, ISO1, SUI1
Chromosome Location:
Chr.17: 41688875 - 41691646 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05015380_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: EIF1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_005801.3 NP_005792.1 4 1216 86
GenBank mRNA AA159721.1 - 86
AF083441.1 - 4 1199 86
AF100737.1 - 4 1183 86
AJ012375.1 - 4 1188 86
AK311209.1 - 3 1037 86
AL050005.1 - 2 2063 86
BC005118.1 - 4 1196 2823520 86
BC008710.1 - 4 1190 3350981 86

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene EIF1

Panther Classification:

Molecular Function -

translation initiation factor

Gene Ontology Categories:

Function(s) Process(es)

translational initiation
regulation of translational initiation
response to stress
dosage compensation by inactivation of X chromosome
translation initiation factor activity
translation factor activity, RNA binding
poly(A) RNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1700007851.hg.1 ENST00000310837.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007851.hg.1 ENST00000310837.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007851.hg.1 ENST00000310837.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007851.hg.1 ENST00000310837.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007851.hg.1 ENST00000310837.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007851.hg.1 ENST00000310837.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007851.hg.1 ENST00000310837.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007851.hg.1 ENST00000310837.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007851.hg.1 ENST00000310837.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007851.hg.1 ENST00000462917.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007851.hg.1 ENST00000462917.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007851.hg.1 ENST00000462917.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007851.hg.1 ENST00000462917.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007851.hg.1 ENST00000462917.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007851.hg.1 ENST00000462917.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007851.hg.1 ENST00000462917.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007851.hg.1 ENST00000462917.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007851.hg.1 ENST00000462917.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007851.hg.1 ENST00000462917.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007851.hg.1 ENST00000462917.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007851.hg.1 ENST00000462917.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007851.hg.1 ENST00000462917.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007851.hg.1 ENST00000462917.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007851.hg.1 ENST00000469257.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007851.hg.1 ENST00000469257.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007851.hg.1 ENST00000469257.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007851.hg.1 ENST00000469257.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007851.hg.1 ENST00000469257.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007851.hg.1 ENST00000469257.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007851.hg.1 ENST00000469257.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007851.hg.1 ENST00000469257.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007851.hg.1 ENST00000469257.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007851.hg.1 ENST00000469257.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007851.hg.1 ENST00000469257.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007851.hg.1 ENST00000469257.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007851.hg.1 ENST00000469308.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007851.hg.1 ENST00000469308.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007851.hg.1 ENST00000469308.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007851.hg.1 ENST00000469308.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007851.hg.1 ENST00000469308.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007851.hg.1 ENST00000469308.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007851.hg.1 ENST00000469308.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007851.hg.1 ENST00000469308.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007851.hg.1 ENST00000469308.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007851.hg.1 ENST00000482111.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007851.hg.1 ENST00000482111.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007851.hg.1 ENST00000482111.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007851.hg.1 ENST00000482111.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007851.hg.1 ENST00000482111.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007851.hg.1 ENST00000482111.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007851.hg.1 ENST00000482111.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007851.hg.1 ENST00000482111.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007851.hg.1 ENST00000482111.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007851.hg.1 ENST00000482111.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007851.hg.1 ENST00000482111.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007851.hg.1 ENST00000482111.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007851.hg.1 ENST00000586699.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007851.hg.1 ENST00000586699.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007851.hg.1 ENST00000586699.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007851.hg.1 ENST00000591776.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007851.hg.1 ENST00000591776.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007851.hg.1 ENST00000591776.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007851.hg.1 ENST00000591776.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007851.hg.1 ENST00000591776.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007851.hg.1 ENST00000591776.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007851.hg.1 ENST00000591776.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007851.hg.1 ENST00000591776.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007851.hg.1 ENST00000591776.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700007851.hg.1 NM_005801.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700007851.hg.1 NM_005801.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700007851.hg.1 NM_005801.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700007851.hg.1 NM_005801.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700007851.hg.1 NM_005801.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700007851.hg.1 NM_005801.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700007851.hg.1 NM_005801.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700007851.hg.1 NM_005801.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700007851.hg.1 NM_005801.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700007851.hg.1 NM_005801.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700007851.hg.1 NM_005801.3 Refseq
Affymetrix Human Clariom D Assay TC1700007851.hg.1 ENST00000310837.8 Ensembl
Affymetrix Human Clariom D Assay TC1700007851.hg.1 ENST00000310837.8 Ensembl
Affymetrix Human Clariom D Assay TC1700007851.hg.1 ENST00000310837.8 Ensembl
Affymetrix Human Clariom D Assay TC1700007851.hg.1 ENST00000310837.8 Ensembl
Affymetrix Human Clariom D Assay TC1700007851.hg.1 ENST00000310837.8 Ensembl
Affymetrix Human Clariom D Assay TC1700007851.hg.1 ENST00000310837.8 Ensembl
Affymetrix Human Clariom D Assay TC1700007851.hg.1 ENST00000310837.8 Ensembl
Affymetrix Human Clariom D Assay TC1700007851.hg.1 ENST00000310837.8 Ensembl
Affymetrix Human Clariom D Assay TC1700007851.hg.1 ENST00000310837.8 Ensembl
Affymetrix Human Clariom D Assay TC1700007851.hg.1 ENST00000462917.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007851.hg.1 ENST00000462917.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007851.hg.1 ENST00000462917.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007851.hg.1 ENST00000462917.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007851.hg.1 ENST00000462917.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007851.hg.1 ENST00000462917.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007851.hg.1 ENST00000462917.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007851.hg.1 ENST00000462917.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007851.hg.1 ENST00000462917.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007851.hg.1 ENST00000462917.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007851.hg.1 ENST00000462917.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007851.hg.1 ENST00000462917.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007851.hg.1 ENST00000462917.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007851.hg.1 ENST00000462917.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007851.hg.1 ENST00000469257.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007851.hg.1 ENST00000469257.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007851.hg.1 ENST00000469257.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007851.hg.1 ENST00000469257.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007851.hg.1 ENST00000469257.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007851.hg.1 ENST00000469257.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007851.hg.1 ENST00000469257.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007851.hg.1 ENST00000469257.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007851.hg.1 ENST00000469257.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007851.hg.1 ENST00000469257.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007851.hg.1 ENST00000469257.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007851.hg.1 ENST00000469257.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007851.hg.1 ENST00000469308.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007851.hg.1 ENST00000469308.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007851.hg.1 ENST00000469308.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007851.hg.1 ENST00000469308.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007851.hg.1 ENST00000469308.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007851.hg.1 ENST00000469308.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007851.hg.1 ENST00000469308.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007851.hg.1 ENST00000469308.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007851.hg.1 ENST00000469308.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007851.hg.1 ENST00000482111.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007851.hg.1 ENST00000482111.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007851.hg.1 ENST00000482111.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007851.hg.1 ENST00000482111.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007851.hg.1 ENST00000482111.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007851.hg.1 ENST00000482111.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007851.hg.1 ENST00000482111.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007851.hg.1 ENST00000482111.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007851.hg.1 ENST00000482111.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007851.hg.1 ENST00000482111.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007851.hg.1 ENST00000482111.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007851.hg.1 ENST00000482111.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007851.hg.1 ENST00000586699.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007851.hg.1 ENST00000586699.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007851.hg.1 ENST00000586699.1 Ensembl
Affymetrix Human Clariom D Assay TC1700007851.hg.1 ENST00000591776.5 Ensembl
Affymetrix Human Clariom D Assay TC1700007851.hg.1 ENST00000591776.5 Ensembl
Affymetrix Human Clariom D Assay TC1700007851.hg.1 ENST00000591776.5 Ensembl
Affymetrix Human Clariom D Assay TC1700007851.hg.1 ENST00000591776.5 Ensembl
Affymetrix Human Clariom D Assay TC1700007851.hg.1 ENST00000591776.5 Ensembl
Affymetrix Human Clariom D Assay TC1700007851.hg.1 ENST00000591776.5 Ensembl
Affymetrix Human Clariom D Assay TC1700007851.hg.1 ENST00000591776.5 Ensembl
Affymetrix Human Clariom D Assay TC1700007851.hg.1 ENST00000591776.5 Ensembl
Affymetrix Human Clariom D Assay TC1700007851.hg.1 ENST00000591776.5 Ensembl
Affymetrix Human Clariom D Assay TC1700007851.hg.1 NM_005801.3 Refseq
Affymetrix Human Clariom D Assay TC1700007851.hg.1 NM_005801.3 Refseq
Affymetrix Human Clariom D Assay TC1700007851.hg.1 NM_005801.3 Refseq
Affymetrix Human Clariom D Assay TC1700007851.hg.1 NM_005801.3 Refseq
Affymetrix Human Clariom D Assay TC1700007851.hg.1 NM_005801.3 Refseq
Affymetrix Human Clariom D Assay TC1700007851.hg.1 NM_005801.3 Refseq
Affymetrix Human Clariom D Assay TC1700007851.hg.1 NM_005801.3 Refseq
Affymetrix Human Clariom D Assay TC1700007851.hg.1 NM_005801.3 Refseq
Affymetrix Human Clariom D Assay TC1700007851.hg.1 NM_005801.3 Refseq
Affymetrix Human Clariom D Assay TC1700007851.hg.1 NM_005801.3 Refseq
Affymetrix Human Clariom D Assay TC1700007851.hg.1 NM_005801.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header