This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05011589_s1
See other FAM63B GE Assays ›
Gene Symbol:
FAM63B
Gene Name:
family with sequence similarity 63 member B
Gene Aliases:
-
Chromosome Location:
Chr.15: 58771192 - 58861900 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05011589_s1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: FAM63B
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001040450.2 NP_001035540.1 9 3405 98
NM_001040453.2 NP_001035543.1 9 3232 98
XM_011521687.1 XP_011519989.1 10 3457 98
XM_011521688.1 XP_011519990.1 10 3454 98
XM_011521689.1 XP_011519991.1 10 3355 98
XM_011521690.1 XP_011519992.1 9 3301 98
XM_017022332.1 XP_016877821.1 8 3211 98
XM_017022333.1 XP_016877822.1 7 3157 98
XM_017022334.1 XP_016877823.1 7 3154 98
GenBank mRNA AB032990.1 - 9 2509 98
AK022891.1 - 1 994 98
AL049226.1 - 1 122 98
AL833610.1 - 8 3259 97

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1500007379.hg.1 ENST00000316848.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007379.hg.1 ENST00000316848.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007379.hg.1 ENST00000316848.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007379.hg.1 ENST00000316848.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007379.hg.1 ENST00000316848.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007379.hg.1 ENST00000316848.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007379.hg.1 ENST00000316848.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007379.hg.1 ENST00000316848.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007379.hg.1 ENST00000316848.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007379.hg.1 ENST00000316848.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007379.hg.1 ENST00000316848.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007379.hg.1 ENST00000316848.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007379.hg.1 ENST00000316848.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007379.hg.1 ENST00000316848.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007379.hg.1 ENST00000316848.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007379.hg.1 ENST00000316848.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007379.hg.1 ENST00000450403.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007379.hg.1 ENST00000450403.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007379.hg.1 ENST00000450403.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007379.hg.1 ENST00000450403.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007379.hg.1 ENST00000450403.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007379.hg.1 ENST00000450403.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007379.hg.1 ENST00000450403.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007379.hg.1 ENST00000450403.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007379.hg.1 ENST00000450403.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007379.hg.1 ENST00000450403.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007379.hg.1 ENST00000450403.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007379.hg.1 ENST00000450403.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007379.hg.1 ENST00000450403.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007379.hg.1 ENST00000450403.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007379.hg.1 ENST00000450403.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007379.hg.1 ENST00000559228.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007379.hg.1 ENST00000559228.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007379.hg.1 ENST00000559228.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007379.hg.1 ENST00000559228.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007379.hg.1 ENST00000559228.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007379.hg.1 ENST00000559228.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007379.hg.1 ENST00000559228.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007379.hg.1 ENST00000559228.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007379.hg.1 ENST00000559228.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007379.hg.1 ENST00000559228.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007379.hg.1 ENST00000559228.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007379.hg.1 ENST00000559228.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007379.hg.1 ENST00000559228.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007379.hg.1 ENST00000559228.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007379.hg.1 ENST00000559228.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007379.hg.1 ENST00000559228.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007379.hg.1 ENST00000559228.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007379.hg.1 ENST00000559228.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007379.hg.1 ENST00000559745.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007379.hg.1 ENST00000559745.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007379.hg.1 ENST00000559745.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007379.hg.1 ENST00000559745.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007379.hg.1 ENST00000559745.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007379.hg.1 ENST00000559745.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007379.hg.1 ENST00000560289.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007379.hg.1 ENST00000560289.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007379.hg.1 ENST00000560289.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007379.hg.1 ENST00000560289.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007379.hg.1 ENST00000560289.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007379.hg.1 ENST00000560289.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007379.hg.1 ENST00000560289.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007379.hg.1 ENST00000560289.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007379.hg.1 ENST00000560289.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007379.hg.1 ENST00000560289.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007379.hg.1 ENST00000560289.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007379.hg.1 ENST00000560289.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007379.hg.1 ENST00000560289.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500007379.hg.1 NM_001040450.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500007379.hg.1 NM_001040450.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500007379.hg.1 NM_001040450.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500007379.hg.1 NM_001040450.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500007379.hg.1 NM_001040450.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500007379.hg.1 NM_001040450.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500007379.hg.1 NM_001040450.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500007379.hg.1 NM_001040450.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500007379.hg.1 NM_001040450.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500007379.hg.1 NM_001040450.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500007379.hg.1 NM_001040450.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500007379.hg.1 NM_001040450.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500007379.hg.1 NM_001040450.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500007379.hg.1 NM_001040450.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500007379.hg.1 NM_001040450.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500007379.hg.1 NM_001040450.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500007379.hg.1 NM_001040450.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500007379.hg.1 NM_001040450.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500007379.hg.1 NM_001040453.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500007379.hg.1 NM_001040453.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500007379.hg.1 NM_001040453.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500007379.hg.1 NM_001040453.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500007379.hg.1 NM_001040453.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500007379.hg.1 NM_001040453.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500007379.hg.1 NM_001040453.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500007379.hg.1 NM_001040453.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500007379.hg.1 NM_001040453.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500007379.hg.1 NM_001040453.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500007379.hg.1 NM_001040453.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500007379.hg.1 NM_001040453.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500007379.hg.1 NM_001040453.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500007379.hg.1 NM_001040453.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500007379.hg.1 NM_001040453.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500007379.hg.1 NM_001040453.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500007379.hg.1 NM_001040453.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500007379.hg.1 NM_001040453.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200007304.hg.1 ENST00000615981.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009583.hg.1 ENST00000558042.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009583.hg.1 ENST00000558042.1 Ensembl
Affymetrix Human Clariom D Assay TC1500009583.hg.1 ENST00000558042.1 Ensembl
Affymetrix Human Clariom D Assay TC1500009583.hg.1 ENST00000558042.1 Ensembl
Affymetrix Human Clariom D Assay TC2200007304.hg.1 ENST00000615981.1 Ensembl
Affymetrix Human Clariom D Assay TC1500007379.hg.1 ENST00000316848.9 Ensembl
Affymetrix Human Clariom D Assay TC1500007379.hg.1 ENST00000316848.9 Ensembl
Affymetrix Human Clariom D Assay TC1500007379.hg.1 ENST00000316848.9 Ensembl
Affymetrix Human Clariom D Assay TC1500007379.hg.1 ENST00000316848.9 Ensembl
Affymetrix Human Clariom D Assay TC1500007379.hg.1 ENST00000316848.9 Ensembl
Affymetrix Human Clariom D Assay TC1500007379.hg.1 ENST00000316848.9 Ensembl
Affymetrix Human Clariom D Assay TC1500007379.hg.1 ENST00000316848.9 Ensembl
Affymetrix Human Clariom D Assay TC1500007379.hg.1 ENST00000316848.9 Ensembl
Affymetrix Human Clariom D Assay TC1500007379.hg.1 ENST00000316848.9 Ensembl
Affymetrix Human Clariom D Assay TC1500007379.hg.1 ENST00000316848.9 Ensembl
Affymetrix Human Clariom D Assay TC1500007379.hg.1 ENST00000316848.9 Ensembl
Affymetrix Human Clariom D Assay TC1500007379.hg.1 ENST00000316848.9 Ensembl
Affymetrix Human Clariom D Assay TC1500007379.hg.1 ENST00000316848.9 Ensembl
Affymetrix Human Clariom D Assay TC1500007379.hg.1 ENST00000316848.9 Ensembl
Affymetrix Human Clariom D Assay TC1500007379.hg.1 ENST00000316848.9 Ensembl
Affymetrix Human Clariom D Assay TC1500007379.hg.1 ENST00000316848.9 Ensembl
Affymetrix Human Clariom D Assay TC1500007379.hg.1 ENST00000450403.3 Ensembl
Affymetrix Human Clariom D Assay TC1500007379.hg.1 ENST00000450403.3 Ensembl
Affymetrix Human Clariom D Assay TC1500007379.hg.1 ENST00000450403.3 Ensembl
Affymetrix Human Clariom D Assay TC1500007379.hg.1 ENST00000450403.3 Ensembl
Affymetrix Human Clariom D Assay TC1500007379.hg.1 ENST00000450403.3 Ensembl
Affymetrix Human Clariom D Assay TC1500007379.hg.1 ENST00000450403.3 Ensembl
Affymetrix Human Clariom D Assay TC1500007379.hg.1 ENST00000450403.3 Ensembl
Affymetrix Human Clariom D Assay TC1500007379.hg.1 ENST00000450403.3 Ensembl
Affymetrix Human Clariom D Assay TC1500007379.hg.1 ENST00000450403.3 Ensembl
Affymetrix Human Clariom D Assay TC1500007379.hg.1 ENST00000450403.3 Ensembl
Affymetrix Human Clariom D Assay TC1500007379.hg.1 ENST00000450403.3 Ensembl
Affymetrix Human Clariom D Assay TC1500007379.hg.1 ENST00000450403.3 Ensembl
Affymetrix Human Clariom D Assay TC1500007379.hg.1 ENST00000450403.3 Ensembl
Affymetrix Human Clariom D Assay TC1500007379.hg.1 ENST00000450403.3 Ensembl
Affymetrix Human Clariom D Assay TC1500007379.hg.1 ENST00000450403.3 Ensembl
Affymetrix Human Clariom D Assay TC1500007379.hg.1 ENST00000559228.5 Ensembl
Affymetrix Human Clariom D Assay TC1500007379.hg.1 ENST00000559228.5 Ensembl
Affymetrix Human Clariom D Assay TC1500007379.hg.1 ENST00000559228.5 Ensembl
Affymetrix Human Clariom D Assay TC1500007379.hg.1 ENST00000559228.5 Ensembl
Affymetrix Human Clariom D Assay TC1500007379.hg.1 ENST00000559228.5 Ensembl
Affymetrix Human Clariom D Assay TC1500007379.hg.1 ENST00000559228.5 Ensembl
Affymetrix Human Clariom D Assay TC1500007379.hg.1 ENST00000559228.5 Ensembl
Affymetrix Human Clariom D Assay TC1500007379.hg.1 ENST00000559228.5 Ensembl
Affymetrix Human Clariom D Assay TC1500007379.hg.1 ENST00000559228.5 Ensembl
Affymetrix Human Clariom D Assay TC1500007379.hg.1 ENST00000559228.5 Ensembl
Affymetrix Human Clariom D Assay TC1500007379.hg.1 ENST00000559228.5 Ensembl
Affymetrix Human Clariom D Assay TC1500007379.hg.1 ENST00000559228.5 Ensembl
Affymetrix Human Clariom D Assay TC1500007379.hg.1 ENST00000559228.5 Ensembl
Affymetrix Human Clariom D Assay TC1500007379.hg.1 ENST00000559228.5 Ensembl
Affymetrix Human Clariom D Assay TC1500007379.hg.1 ENST00000559228.5 Ensembl
Affymetrix Human Clariom D Assay TC1500007379.hg.1 ENST00000559228.5 Ensembl
Affymetrix Human Clariom D Assay TC1500007379.hg.1 ENST00000559228.5 Ensembl
Affymetrix Human Clariom D Assay TC1500007379.hg.1 ENST00000559228.5 Ensembl
Affymetrix Human Clariom D Assay TC1500007379.hg.1 ENST00000559745.2 Ensembl
Affymetrix Human Clariom D Assay TC1500007379.hg.1 ENST00000559745.2 Ensembl
Affymetrix Human Clariom D Assay TC1500007379.hg.1 ENST00000559745.2 Ensembl
Affymetrix Human Clariom D Assay TC1500007379.hg.1 ENST00000559745.2 Ensembl
Affymetrix Human Clariom D Assay TC1500007379.hg.1 ENST00000559745.2 Ensembl
Affymetrix Human Clariom D Assay TC1500007379.hg.1 ENST00000559745.2 Ensembl
Affymetrix Human Clariom D Assay TC1500007379.hg.1 ENST00000560289.5 Ensembl
Affymetrix Human Clariom D Assay TC1500007379.hg.1 ENST00000560289.5 Ensembl
Affymetrix Human Clariom D Assay TC1500007379.hg.1 ENST00000560289.5 Ensembl
Affymetrix Human Clariom D Assay TC1500007379.hg.1 ENST00000560289.5 Ensembl
Affymetrix Human Clariom D Assay TC1500007379.hg.1 ENST00000560289.5 Ensembl
Affymetrix Human Clariom D Assay TC1500007379.hg.1 ENST00000560289.5 Ensembl
Affymetrix Human Clariom D Assay TC1500007379.hg.1 ENST00000560289.5 Ensembl
Affymetrix Human Clariom D Assay TC1500007379.hg.1 ENST00000560289.5 Ensembl
Affymetrix Human Clariom D Assay TC1500007379.hg.1 ENST00000560289.5 Ensembl
Affymetrix Human Clariom D Assay TC1500007379.hg.1 ENST00000560289.5 Ensembl
Affymetrix Human Clariom D Assay TC1500007379.hg.1 ENST00000560289.5 Ensembl
Affymetrix Human Clariom D Assay TC1500007379.hg.1 ENST00000560289.5 Ensembl
Affymetrix Human Clariom D Assay TC1500007379.hg.1 ENST00000560289.5 Ensembl
Affymetrix Human Clariom D Assay TC1500007379.hg.1 NM_001040450.2 Refseq
Affymetrix Human Clariom D Assay TC1500007379.hg.1 NM_001040450.2 Refseq
Affymetrix Human Clariom D Assay TC1500007379.hg.1 NM_001040450.2 Refseq
Affymetrix Human Clariom D Assay TC1500007379.hg.1 NM_001040450.2 Refseq
Affymetrix Human Clariom D Assay TC1500007379.hg.1 NM_001040450.2 Refseq
Affymetrix Human Clariom D Assay TC1500007379.hg.1 NM_001040450.2 Refseq
Affymetrix Human Clariom D Assay TC1500007379.hg.1 NM_001040450.2 Refseq
Affymetrix Human Clariom D Assay TC1500007379.hg.1 NM_001040450.2 Refseq
Affymetrix Human Clariom D Assay TC1500007379.hg.1 NM_001040450.2 Refseq
Affymetrix Human Clariom D Assay TC1500007379.hg.1 NM_001040450.2 Refseq
Affymetrix Human Clariom D Assay TC1500007379.hg.1 NM_001040450.2 Refseq
Affymetrix Human Clariom D Assay TC1500007379.hg.1 NM_001040450.2 Refseq
Affymetrix Human Clariom D Assay TC1500007379.hg.1 NM_001040450.2 Refseq
Affymetrix Human Clariom D Assay TC1500007379.hg.1 NM_001040450.2 Refseq
Affymetrix Human Clariom D Assay TC1500007379.hg.1 NM_001040450.2 Refseq
Affymetrix Human Clariom D Assay TC1500007379.hg.1 NM_001040450.2 Refseq
Affymetrix Human Clariom D Assay TC1500007379.hg.1 NM_001040450.2 Refseq
Affymetrix Human Clariom D Assay TC1500007379.hg.1 NM_001040450.2 Refseq
Affymetrix Human Clariom D Assay TC1500007379.hg.1 NM_001040453.2 Refseq
Affymetrix Human Clariom D Assay TC1500007379.hg.1 NM_001040453.2 Refseq
Affymetrix Human Clariom D Assay TC1500007379.hg.1 NM_001040453.2 Refseq
Affymetrix Human Clariom D Assay TC1500007379.hg.1 NM_001040453.2 Refseq
Affymetrix Human Clariom D Assay TC1500007379.hg.1 NM_001040453.2 Refseq
Affymetrix Human Clariom D Assay TC1500007379.hg.1 NM_001040453.2 Refseq
Affymetrix Human Clariom D Assay TC1500007379.hg.1 NM_001040453.2 Refseq
Affymetrix Human Clariom D Assay TC1500007379.hg.1 NM_001040453.2 Refseq
Affymetrix Human Clariom D Assay TC1500007379.hg.1 NM_001040453.2 Refseq
Affymetrix Human Clariom D Assay TC1500007379.hg.1 NM_001040453.2 Refseq
Affymetrix Human Clariom D Assay TC1500007379.hg.1 NM_001040453.2 Refseq
Affymetrix Human Clariom D Assay TC1500007379.hg.1 NM_001040453.2 Refseq
Affymetrix Human Clariom D Assay TC1500007379.hg.1 NM_001040453.2 Refseq
Affymetrix Human Clariom D Assay TC1500007379.hg.1 NM_001040453.2 Refseq
Affymetrix Human Clariom D Assay TC1500007379.hg.1 NM_001040453.2 Refseq
Affymetrix Human Clariom D Assay TC1500007379.hg.1 NM_001040453.2 Refseq
Affymetrix Human Clariom D Assay TC1500007379.hg.1 NM_001040453.2 Refseq
Affymetrix Human Clariom D Assay TC1500007379.hg.1 NM_001040453.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0