This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05010074_s1
See other FAM63B GE Assays ›
Gene Symbol:
FAM63B
Gene Name:
family with sequence similarity 63 member B
Gene Aliases:
-
Chromosome Location:
Chr.15: 58771192 - 58861900 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05010074_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: FAM63B
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001040450.2 NP_001035540.1 9 8954 130
NM_001040453.2 NP_001035543.1 9 8781 130
XM_011521687.1 XP_011519989.1 10 9006 130
XM_011521688.1 XP_011519990.1 10 9003 130
XM_011521689.1 XP_011519991.1 10 8904 130
XM_011521690.1 XP_011519992.1 9 8850 130
XM_017022332.1 XP_016877821.1 8 8760 130
XM_017022333.1 XP_016877822.1 7 8706 130
XM_017022334.1 XP_016877823.1 7 8703 130
GenBank mRNA BC037808.2 - 1 713 130
BC042116.1 - 1 713 130
CB321913.1 - 1 130

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0