Assay Details
Gene Symbol: DPF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK124615.1 | - | 1 | 415 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |

Gene Symbol: DPF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK124615.1 | - | 1 | 415 | 72 |
Set Membership: |
![]() ![]() ![]() ![]() |