This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05007069_s1
See other GPR180 GE Assays ›
Gene Symbol:
GPR180
Gene Name:
G protein-coupled receptor 180
Gene Aliases:
ITR
Chromosome Location:
Chr.13: 94601850 - 94634645 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05007069_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GPR180
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_180989.5 NP_851320.1 9 8817 118
GenBank mRNA AL049452.1 - 1 519 118

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene GPR180

Gene Ontology Categories:

Function(s)

G-protein coupled receptor signaling pathway
response to pheromone

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1300007731.hg.1 ENST00000376958.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300007731.hg.1 ENST00000376958.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300007731.hg.1 ENST00000376958.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300007731.hg.1 ENST00000376958.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300007731.hg.1 ENST00000376958.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300007731.hg.1 ENST00000376958.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300007731.hg.1 ENST00000376958.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300007731.hg.1 ENST00000376958.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300007731.hg.1 ENST00000376958.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300007731.hg.1 ENST00000376958.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300007731.hg.1 ENST00000376958.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300007731.hg.1 ENST00000376958.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300007731.hg.1 ENST00000376958.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300007731.hg.1 ENST00000376958.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300007731.hg.1 NM_180989.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300007731.hg.1 NM_180989.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300007731.hg.1 NM_180989.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300007731.hg.1 NM_180989.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300007731.hg.1 NM_180989.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300007731.hg.1 NM_180989.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300007731.hg.1 NM_180989.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300007731.hg.1 NM_180989.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300007731.hg.1 NM_180989.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300007731.hg.1 NM_180989.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300007731.hg.1 NM_180989.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300007731.hg.1 NM_180989.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300007731.hg.1 NM_180989.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300007731.hg.1 NM_180989.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300007731.hg.1 NM_180989.5 Refseq
Affymetrix Human Clariom D Assay TC1300007731.hg.1 ENST00000376958.4 Ensembl
Affymetrix Human Clariom D Assay TC1300007731.hg.1 ENST00000376958.4 Ensembl
Affymetrix Human Clariom D Assay TC1300007731.hg.1 ENST00000376958.4 Ensembl
Affymetrix Human Clariom D Assay TC1300007731.hg.1 ENST00000376958.4 Ensembl
Affymetrix Human Clariom D Assay TC1300007731.hg.1 ENST00000376958.4 Ensembl
Affymetrix Human Clariom D Assay TC1300007731.hg.1 ENST00000376958.4 Ensembl
Affymetrix Human Clariom D Assay TC1300007731.hg.1 ENST00000376958.4 Ensembl
Affymetrix Human Clariom D Assay TC1300007731.hg.1 ENST00000376958.4 Ensembl
Affymetrix Human Clariom D Assay TC1300007731.hg.1 ENST00000376958.4 Ensembl
Affymetrix Human Clariom D Assay TC1300007731.hg.1 ENST00000376958.4 Ensembl
Affymetrix Human Clariom D Assay TC1300007731.hg.1 ENST00000376958.4 Ensembl
Affymetrix Human Clariom D Assay TC1300007731.hg.1 ENST00000376958.4 Ensembl
Affymetrix Human Clariom D Assay TC1300007731.hg.1 ENST00000376958.4 Ensembl
Affymetrix Human Clariom D Assay TC1300007731.hg.1 ENST00000376958.4 Ensembl
Affymetrix Human Clariom D Assay TC1300007731.hg.1 NM_180989.5 Refseq
Affymetrix Human Clariom D Assay TC1300007731.hg.1 NM_180989.5 Refseq
Affymetrix Human Clariom D Assay TC1300007731.hg.1 NM_180989.5 Refseq
Affymetrix Human Clariom D Assay TC1300007731.hg.1 NM_180989.5 Refseq
Affymetrix Human Clariom D Assay TC1300007731.hg.1 NM_180989.5 Refseq
Affymetrix Human Clariom D Assay TC1300007731.hg.1 NM_180989.5 Refseq
Affymetrix Human Clariom D Assay TC1300007731.hg.1 NM_180989.5 Refseq
Affymetrix Human Clariom D Assay TC1300007731.hg.1 NM_180989.5 Refseq
Affymetrix Human Clariom D Assay TC1300007731.hg.1 NM_180989.5 Refseq
Affymetrix Human Clariom D Assay TC1300007731.hg.1 NM_180989.5 Refseq
Affymetrix Human Clariom D Assay TC1300007731.hg.1 NM_180989.5 Refseq
Affymetrix Human Clariom D Assay TC1300007731.hg.1 NM_180989.5 Refseq
Affymetrix Human Clariom D Assay TC1300007731.hg.1 NM_180989.5 Refseq
Affymetrix Human Clariom D Assay TC1300007731.hg.1 NM_180989.5 Refseq
Affymetrix Human Clariom D Assay TC1300007731.hg.1 NM_180989.5 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0