This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05006563_s1
See other DGKH GE Assays ›
Gene Symbol:
DGKH
Gene Name:
diacylglycerol kinase eta
Gene Aliases:
DGKeta
Chromosome Location:
Chr.13: 42040036 - 42256584 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05006563_s1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: DGKH
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001204504.2 NP_001191433.1 30 14076 115
NM_001204505.2 NP_001191434.1 29 13876 115
NM_001204506.2 NP_001191435.1 28 13828 115
NM_001297429.1 NP_001284358.1 25 13464 115
NM_152910.5 NP_690874.2 29 14039 115
NM_178009.4 NP_821077.1 30 14170 115
NR_123714.1 28 13930 115
GenBank mRNA AK090591.1 - 1 420 115
AK130015.1 - 1 565 115

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene DGKH

Panther Classification:

Molecular Function -

kinase

Gene Ontology Categories:

Function(s) Process(es)

protein kinase C-activating G-protein coupled receptor signaling pathway
phosphorylation
platelet activation
intracellular signal transduction
protein oligomerization
diacylglycerol kinase activity
ATP binding
metal ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0