Assay Details
Gene Symbol: MTMR6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_011535306.1 | XP_011533608.1 | 7 - 8 | 1105 | 118 | |
XM_011535307.1 | XP_011533609.1 | 7 - 8 | 1105 | 118 | ||
XM_017020846.1 | XP_016876335.1 | 3 - 4 | 310 | 118 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
protein dephosphorylation
phosphatidylinositol biosynthetic process
peptidyl-tyrosine dephosphorylation
phosphatidylinositol dephosphorylation
potassium ion transmembrane transport
phosphatidylinositol-3-phosphatase activity
protein serine/threonine phosphatase activity
protein tyrosine phosphatase activity
protein binding
calcium-activated potassium channel activity
phosphatidylinositol-3,5-bisphosphate 3-phosphatase activity
protein dephosphorylation
phosphatidylinositol biosynthetic process
peptidyl-tyrosine dephosphorylation
phosphatidylinositol dephosphorylation
potassium ion transmembrane transport
phosphatidylinositol-3-phosphatase activity
protein serine/threonine phosphatase activity
protein tyrosine phosphatase activity
protein binding
calcium-activated potassium channel activity
phosphatidylinositol-3,5-bisphosphate 3-phosphatase activity
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1300008386.hg.1 | ENST00000381801.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008386.hg.1 | ENST00000381801.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008386.hg.1 | ENST00000381801.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008386.hg.1 | ENST00000381801.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008386.hg.1 | ENST00000381801.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008386.hg.1 | ENST00000381801.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008386.hg.1 | ENST00000381801.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008386.hg.1 | ENST00000381801.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008386.hg.1 | ENST00000381801.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008386.hg.1 | ENST00000381801.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008386.hg.1 | ENST00000381801.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008386.hg.1 | ENST00000381801.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008386.hg.1 | ENST00000381801.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008386.hg.1 | ENST00000381801.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008386.hg.1 | ENST00000381801.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008386.hg.1 | ENST00000381801.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008386.hg.1 | ENST00000381801.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008386.hg.1 | ENST00000381801.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008386.hg.1 | ENST00000381801.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008386.hg.1 | ENST00000482345.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008386.hg.1 | ENST00000482345.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008386.hg.1 | ENST00000482345.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008386.hg.1 | NM_004685.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008386.hg.1 | NM_004685.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008386.hg.1 | NM_004685.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008386.hg.1 | NM_004685.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008386.hg.1 | NM_004685.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008386.hg.1 | NM_004685.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008386.hg.1 | NM_004685.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008386.hg.1 | NM_004685.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008386.hg.1 | NM_004685.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008386.hg.1 | NM_004685.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008386.hg.1 | NM_004685.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008386.hg.1 | NM_004685.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008386.hg.1 | NM_004685.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008386.hg.1 | NM_004685.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008386.hg.1 | NM_004685.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008386.hg.1 | NM_004685.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008386.hg.1 | NM_004685.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300008386.hg.1 | NM_004685.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008386.hg.1 | ENST00000381801.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008386.hg.1 | ENST00000381801.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008386.hg.1 | ENST00000381801.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008386.hg.1 | ENST00000381801.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008386.hg.1 | ENST00000381801.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008386.hg.1 | ENST00000381801.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008386.hg.1 | ENST00000381801.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008386.hg.1 | ENST00000381801.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008386.hg.1 | ENST00000381801.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008386.hg.1 | ENST00000381801.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008386.hg.1 | ENST00000381801.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008386.hg.1 | ENST00000381801.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008386.hg.1 | ENST00000381801.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008386.hg.1 | ENST00000381801.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008386.hg.1 | ENST00000381801.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008386.hg.1 | ENST00000381801.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008386.hg.1 | ENST00000381801.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008386.hg.1 | ENST00000381801.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008386.hg.1 | ENST00000381801.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008386.hg.1 | ENST00000482345.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008386.hg.1 | ENST00000482345.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008386.hg.1 | ENST00000482345.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008386.hg.1 | NM_004685.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008386.hg.1 | NM_004685.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008386.hg.1 | NM_004685.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008386.hg.1 | NM_004685.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008386.hg.1 | NM_004685.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008386.hg.1 | NM_004685.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008386.hg.1 | NM_004685.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008386.hg.1 | NM_004685.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008386.hg.1 | NM_004685.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008386.hg.1 | NM_004685.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008386.hg.1 | NM_004685.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008386.hg.1 | NM_004685.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008386.hg.1 | NM_004685.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008386.hg.1 | NM_004685.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008386.hg.1 | NM_004685.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008386.hg.1 | NM_004685.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008386.hg.1 | NM_004685.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1300008386.hg.1 | NM_004685.3 | Refseq |
