This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05005432_m1
See other PGAM5 GE Assays ›
Gene Symbol:
PGAM5
Gene Name:
PGAM family member 5, mitochondrial serine/threonine protein phosphatase
Gene Aliases:
BXLBV68
Chromosome Location:
Chr.12: 132710807 - 132722737 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05005432_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PGAM5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001170543.1 NP_001164014.1 5 - 6 784 64
NM_001170544.1 NP_001164015.1 5 - 6 781 64
GenBank mRNA AK097688.1 - 5 - 6 1131 64
BC047906.2 - 4 - 5 436 64
CA307621.1 - 1 - 2 64
DR155760.1 - 5 - 6 749 64
EU249757.1 - 5 - 6 772 64

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Homo sapiens Probe spans exons

Panther Classification:

Molecular Function -

isomerase mutase metabolite interconversion enzyme

Gene Ontology Categories:

Function(s) Process(es)

protein dephosphorylation
signal transduction
positive regulation of GTPase activity
necroptotic process
GTPase activator activity
receptor signaling protein serine/threonine phosphatase activity
phosphatase activity
protein complex binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1200012731.hg.1 ENST00000317555.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012731.hg.1 ENST00000317555.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012731.hg.1 ENST00000317555.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012731.hg.1 ENST00000317555.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012731.hg.1 ENST00000317555.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012731.hg.1 ENST00000317555.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012731.hg.1 ENST00000317555.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012731.hg.1 ENST00000317555.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012731.hg.1 ENST00000317555.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012731.hg.1 ENST00000317555.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012731.hg.1 ENST00000454808.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012731.hg.1 ENST00000454808.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012731.hg.1 ENST00000454808.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012731.hg.1 ENST00000454808.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012731.hg.1 ENST00000454808.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012731.hg.1 ENST00000454808.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012731.hg.1 ENST00000454808.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012731.hg.1 ENST00000454808.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012731.hg.1 ENST00000454808.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012731.hg.1 ENST00000498926.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012731.hg.1 ENST00000498926.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012731.hg.1 ENST00000498926.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012731.hg.1 ENST00000498926.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012731.hg.1 ENST00000498926.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012731.hg.1 ENST00000498926.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012731.hg.1 ENST00000498926.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012731.hg.1 ENST00000498926.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012731.hg.1 ENST00000498926.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012731.hg.1 ENST00000498926.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012731.hg.1 ENST00000498926.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012731.hg.1 ENST00000498926.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012731.hg.1 ENST00000498926.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012731.hg.1 ENST00000498926.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012731.hg.1 ENST00000543955.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012731.hg.1 ENST00000543955.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012731.hg.1 ENST00000543955.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012731.hg.1 ENST00000543955.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012731.hg.1 ENST00000543955.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012731.hg.1 ENST00000543955.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012731.hg.1 ENST00000543955.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012731.hg.1 ENST00000543955.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012731.hg.1 ENST00000543955.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012731.hg.1 ENST00000543955.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012731.hg.1 ENST00000543955.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200012731.hg.1 NM_001170543.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012731.hg.1 NM_001170543.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012731.hg.1 NM_001170543.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012731.hg.1 NM_001170543.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012731.hg.1 NM_001170543.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012731.hg.1 NM_001170543.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012731.hg.1 NM_001170543.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012731.hg.1 NM_001170543.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012731.hg.1 NM_001170543.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012731.hg.1 NM_001170543.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012731.hg.1 NM_001170543.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012731.hg.1 NM_001170543.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012731.hg.1 NM_001170543.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012731.hg.1 NM_001170543.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012731.hg.1 NM_001170544.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012731.hg.1 NM_001170544.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012731.hg.1 NM_001170544.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012731.hg.1 NM_001170544.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012731.hg.1 NM_001170544.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012731.hg.1 NM_001170544.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012731.hg.1 NM_001170544.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012731.hg.1 NM_001170544.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012731.hg.1 NM_001170544.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012731.hg.1 NM_001170544.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012731.hg.1 NM_001170544.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012731.hg.1 NM_001170544.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012731.hg.1 NM_001170544.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012731.hg.1 NM_001170544.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012731.hg.1 NM_138575.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012731.hg.1 NM_138575.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012731.hg.1 NM_138575.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012731.hg.1 NM_138575.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012731.hg.1 NM_138575.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012731.hg.1 NM_138575.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012731.hg.1 NM_138575.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012731.hg.1 NM_138575.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012731.hg.1 NM_138575.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200012731.hg.1 NM_138575.3 Refseq
Affymetrix Human Clariom D Assay TC1200012731.hg.1 ENST00000317555.6 Ensembl
Affymetrix Human Clariom D Assay TC1200012731.hg.1 ENST00000317555.6 Ensembl
Affymetrix Human Clariom D Assay TC1200012731.hg.1 ENST00000317555.6 Ensembl
Affymetrix Human Clariom D Assay TC1200012731.hg.1 ENST00000317555.6 Ensembl
Affymetrix Human Clariom D Assay TC1200012731.hg.1 ENST00000317555.6 Ensembl
Affymetrix Human Clariom D Assay TC1200012731.hg.1 ENST00000317555.6 Ensembl
Affymetrix Human Clariom D Assay TC1200012731.hg.1 ENST00000317555.6 Ensembl
Affymetrix Human Clariom D Assay TC1200012731.hg.1 ENST00000317555.6 Ensembl
Affymetrix Human Clariom D Assay TC1200012731.hg.1 ENST00000317555.6 Ensembl
Affymetrix Human Clariom D Assay TC1200012731.hg.1 ENST00000317555.6 Ensembl
Affymetrix Human Clariom D Assay TC1200012731.hg.1 ENST00000454808.2 Ensembl
Affymetrix Human Clariom D Assay TC1200012731.hg.1 ENST00000454808.2 Ensembl
Affymetrix Human Clariom D Assay TC1200012731.hg.1 ENST00000454808.2 Ensembl
Affymetrix Human Clariom D Assay TC1200012731.hg.1 ENST00000454808.2 Ensembl
Affymetrix Human Clariom D Assay TC1200012731.hg.1 ENST00000454808.2 Ensembl
Affymetrix Human Clariom D Assay TC1200012731.hg.1 ENST00000454808.2 Ensembl
Affymetrix Human Clariom D Assay TC1200012731.hg.1 ENST00000454808.2 Ensembl
Affymetrix Human Clariom D Assay TC1200012731.hg.1 ENST00000454808.2 Ensembl
Affymetrix Human Clariom D Assay TC1200012731.hg.1 ENST00000454808.2 Ensembl
Affymetrix Human Clariom D Assay TC1200012731.hg.1 ENST00000498926.6 Ensembl
Affymetrix Human Clariom D Assay TC1200012731.hg.1 ENST00000498926.6 Ensembl
Affymetrix Human Clariom D Assay TC1200012731.hg.1 ENST00000498926.6 Ensembl
Affymetrix Human Clariom D Assay TC1200012731.hg.1 ENST00000498926.6 Ensembl
Affymetrix Human Clariom D Assay TC1200012731.hg.1 ENST00000498926.6 Ensembl
Affymetrix Human Clariom D Assay TC1200012731.hg.1 ENST00000498926.6 Ensembl
Affymetrix Human Clariom D Assay TC1200012731.hg.1 ENST00000498926.6 Ensembl
Affymetrix Human Clariom D Assay TC1200012731.hg.1 ENST00000498926.6 Ensembl
Affymetrix Human Clariom D Assay TC1200012731.hg.1 ENST00000498926.6 Ensembl
Affymetrix Human Clariom D Assay TC1200012731.hg.1 ENST00000498926.6 Ensembl
Affymetrix Human Clariom D Assay TC1200012731.hg.1 ENST00000498926.6 Ensembl
Affymetrix Human Clariom D Assay TC1200012731.hg.1 ENST00000498926.6 Ensembl
Affymetrix Human Clariom D Assay TC1200012731.hg.1 ENST00000498926.6 Ensembl
Affymetrix Human Clariom D Assay TC1200012731.hg.1 ENST00000498926.6 Ensembl
Affymetrix Human Clariom D Assay TC1200012731.hg.1 ENST00000543955.5 Ensembl
Affymetrix Human Clariom D Assay TC1200012731.hg.1 ENST00000543955.5 Ensembl
Affymetrix Human Clariom D Assay TC1200012731.hg.1 ENST00000543955.5 Ensembl
Affymetrix Human Clariom D Assay TC1200012731.hg.1 ENST00000543955.5 Ensembl
Affymetrix Human Clariom D Assay TC1200012731.hg.1 ENST00000543955.5 Ensembl
Affymetrix Human Clariom D Assay TC1200012731.hg.1 ENST00000543955.5 Ensembl
Affymetrix Human Clariom D Assay TC1200012731.hg.1 ENST00000543955.5 Ensembl
Affymetrix Human Clariom D Assay TC1200012731.hg.1 ENST00000543955.5 Ensembl
Affymetrix Human Clariom D Assay TC1200012731.hg.1 ENST00000543955.5 Ensembl
Affymetrix Human Clariom D Assay TC1200012731.hg.1 ENST00000543955.5 Ensembl
Affymetrix Human Clariom D Assay TC1200012731.hg.1 ENST00000543955.5 Ensembl
Affymetrix Human Clariom D Assay TC1200012731.hg.1 NM_001170543.1 Refseq
Affymetrix Human Clariom D Assay TC1200012731.hg.1 NM_001170543.1 Refseq
Affymetrix Human Clariom D Assay TC1200012731.hg.1 NM_001170543.1 Refseq
Affymetrix Human Clariom D Assay TC1200012731.hg.1 NM_001170543.1 Refseq
Affymetrix Human Clariom D Assay TC1200012731.hg.1 NM_001170543.1 Refseq
Affymetrix Human Clariom D Assay TC1200012731.hg.1 NM_001170543.1 Refseq
Affymetrix Human Clariom D Assay TC1200012731.hg.1 NM_001170543.1 Refseq
Affymetrix Human Clariom D Assay TC1200012731.hg.1 NM_001170543.1 Refseq
Affymetrix Human Clariom D Assay TC1200012731.hg.1 NM_001170543.1 Refseq
Affymetrix Human Clariom D Assay TC1200012731.hg.1 NM_001170543.1 Refseq
Affymetrix Human Clariom D Assay TC1200012731.hg.1 NM_001170543.1 Refseq
Affymetrix Human Clariom D Assay TC1200012731.hg.1 NM_001170543.1 Refseq
Affymetrix Human Clariom D Assay TC1200012731.hg.1 NM_001170543.1 Refseq
Affymetrix Human Clariom D Assay TC1200012731.hg.1 NM_001170543.1 Refseq
Affymetrix Human Clariom D Assay TC1200012731.hg.1 NM_001170544.1 Refseq
Affymetrix Human Clariom D Assay TC1200012731.hg.1 NM_001170544.1 Refseq
Affymetrix Human Clariom D Assay TC1200012731.hg.1 NM_001170544.1 Refseq
Affymetrix Human Clariom D Assay TC1200012731.hg.1 NM_001170544.1 Refseq
Affymetrix Human Clariom D Assay TC1200012731.hg.1 NM_001170544.1 Refseq
Affymetrix Human Clariom D Assay TC1200012731.hg.1 NM_001170544.1 Refseq
Affymetrix Human Clariom D Assay TC1200012731.hg.1 NM_001170544.1 Refseq
Affymetrix Human Clariom D Assay TC1200012731.hg.1 NM_001170544.1 Refseq
Affymetrix Human Clariom D Assay TC1200012731.hg.1 NM_001170544.1 Refseq
Affymetrix Human Clariom D Assay TC1200012731.hg.1 NM_001170544.1 Refseq
Affymetrix Human Clariom D Assay TC1200012731.hg.1 NM_001170544.1 Refseq
Affymetrix Human Clariom D Assay TC1200012731.hg.1 NM_001170544.1 Refseq
Affymetrix Human Clariom D Assay TC1200012731.hg.1 NM_001170544.1 Refseq
Affymetrix Human Clariom D Assay TC1200012731.hg.1 NM_001170544.1 Refseq
Affymetrix Human Clariom D Assay TC1200012731.hg.1 NM_138575.3 Refseq
Affymetrix Human Clariom D Assay TC1200012731.hg.1 NM_138575.3 Refseq
Affymetrix Human Clariom D Assay TC1200012731.hg.1 NM_138575.3 Refseq
Affymetrix Human Clariom D Assay TC1200012731.hg.1 NM_138575.3 Refseq
Affymetrix Human Clariom D Assay TC1200012731.hg.1 NM_138575.3 Refseq
Affymetrix Human Clariom D Assay TC1200012731.hg.1 NM_138575.3 Refseq
Affymetrix Human Clariom D Assay TC1200012731.hg.1 NM_138575.3 Refseq
Affymetrix Human Clariom D Assay TC1200012731.hg.1 NM_138575.3 Refseq
Affymetrix Human Clariom D Assay TC1200012731.hg.1 NM_138575.3 Refseq
Affymetrix Human Clariom D Assay TC1200012731.hg.1 NM_138575.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7db4fb6694-gx648:80/100.66.128.142:80.
git-commit: 672a5a8c50d6b616e949be631a941c86748ade1b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header