This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05005387_s1
See other LRIT1 GE Assays ›
Gene Symbol:
LRIT1
Gene Name:
leucine rich repeat, Ig-like and transmembrane domains 1
Gene Aliases:
FIGLER9, LRRC21, PAL
Chromosome Location:
Chr.10: 84231520 - 84241461 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05005387_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LRIT1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA DQ426881.1 - 1 120 118
DQ426882.1 - 1 82 118

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the DQ426882.1 transcripts.

Set Membership:

Amplicon length greater than or equal to 101 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1000011267.hg.1 ENST00000372105.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011267.hg.1 ENST00000372105.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011267.hg.1 ENST00000372105.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011267.hg.1 ENST00000372105.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011267.hg.1 ENST00000372105.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011267.hg.1 ENST00000372105.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011267.hg.1 NM_015613.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011267.hg.1 NM_015613.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011267.hg.1 NM_015613.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011267.hg.1 NM_015613.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011267.hg.1 NM_015613.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011267.hg.1 NM_015613.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011267.hg.1 NM_015613.2 Refseq
Affymetrix Human Clariom D Assay TC1000011267.hg.1 ENST00000372105.3 Ensembl
Affymetrix Human Clariom D Assay TC1000011267.hg.1 ENST00000372105.3 Ensembl
Affymetrix Human Clariom D Assay TC1000011267.hg.1 ENST00000372105.3 Ensembl
Affymetrix Human Clariom D Assay TC1000011267.hg.1 ENST00000372105.3 Ensembl
Affymetrix Human Clariom D Assay TC1000011267.hg.1 ENST00000372105.3 Ensembl
Affymetrix Human Clariom D Assay TC1000011267.hg.1 ENST00000372105.3 Ensembl
Affymetrix Human Clariom D Assay TC1000011267.hg.1 NM_015613.2 Refseq
Affymetrix Human Clariom D Assay TC1000011267.hg.1 NM_015613.2 Refseq
Affymetrix Human Clariom D Assay TC1000011267.hg.1 NM_015613.2 Refseq
Affymetrix Human Clariom D Assay TC1000011267.hg.1 NM_015613.2 Refseq
Affymetrix Human Clariom D Assay TC1000011267.hg.1 NM_015613.2 Refseq
Affymetrix Human Clariom D Assay TC1000011267.hg.1 NM_015613.2 Refseq
Affymetrix Human Clariom D Assay TC1000011267.hg.1 NM_015613.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0