This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05005339_m1
See other ACTA2 GE Assays ›
Gene Symbol:
ACTA2
Gene Name:
actin, alpha 2, smooth muscle, aorta
Gene Aliases:
AAT6, ACTSA, MYMY5
Chromosome Location:
Chr.10: 88935074 - 88991397 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05005339_m1
Size
Availability Inventoried
Catalog # 4331182
Price 244.00
Your Price
Online offer: 200.00
200.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ACTA2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001613.2 NP_001604.1 1 - 2 90 102
GenBank mRNA BP379410.1 - 1 - 2 276 102

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the NM_001613.2 transcripts.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Homo sapiens Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene ACTA2

Panther Classification:

Molecular Function -

actin and actin related protein actin or actin-binding cytoskeletal protein cytoskeletal protein

Gene Ontology Categories:

Function(s) Process(es)

muscle contraction
regulation of blood pressure
response to virus
positive regulation of gene expression
vascular smooth muscle contraction
glomerular mesangial cell development
mesenchyme migration
ATP binding
protein kinase binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0