This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05005296_m1
See other PLCE1-AS1 GE Assays ›
Gene Symbol:
PLCE1-AS1
Gene Name:
PLCE1 antisense RNA 1
Gene Aliases:
-
Chromosome Location:
Chr.10: 94279290 - 94287070 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05005296_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PLCE1-AS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_033969.1 3 - 4 366 96

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Recommended Assay Homo sapiens Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1000011467.hg.1 ENST00000425267.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011467.hg.1 ENST00000425267.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011467.hg.1 ENST00000425267.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011467.hg.1 ENST00000425267.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011467.hg.1 ENST00000425267.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011467.hg.1 ENST00000425267.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011467.hg.1 ENST00000425267.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011467.hg.1 ENST00000425267.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011467.hg.1 ENST00000425267.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011467.hg.1 ENST00000425267.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011467.hg.1 ENST00000425267.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011467.hg.1 ENST00000425267.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011467.hg.1 ENST00000425267.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011467.hg.1 ENST00000425267.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011467.hg.1 ENST00000440198.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011467.hg.1 ENST00000440198.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011467.hg.1 ENST00000440198.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011467.hg.1 ENST00000440198.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011467.hg.1 ENST00000440198.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011467.hg.1 ENST00000440198.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011467.hg.1 ENST00000440198.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011467.hg.1 ENST00000440198.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011467.hg.1 ENST00000440198.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011467.hg.1 ENST00000440198.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011467.hg.1 ENST00000440198.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011467.hg.1 ENST00000596633.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011467.hg.1 ENST00000596633.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011467.hg.1 ENST00000596633.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011467.hg.1 ENST00000596633.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011467.hg.1 ENST00000596633.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011467.hg.1 ENST00000596633.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011467.hg.1 ENST00000596633.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011467.hg.1 ENST00000596633.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011467.hg.1 ENST00000596633.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011467.hg.1 ENST00000596633.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011467.hg.1 ENST00000596633.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011467.hg.1 ENST00000596633.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011467.hg.1 ENST00000596633.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011467.hg.1 ENST00000596633.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011467.hg.1 ENST00000596633.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011467.hg.1 ENST00000596633.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011467.hg.1 ENST00000611247.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011467.hg.1 ENST00000611247.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011467.hg.1 ENST00000611247.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011467.hg.1 ENST00000611247.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011467.hg.1 NR_033969.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011467.hg.1 NR_033969.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011467.hg.1 NR_033969.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011467.hg.1 NR_033969.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011467.hg.1 NR_033969.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011467.hg.1 NR_033969.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011467.hg.1 NR_033969.1 Refseq
Affymetrix Human Clariom D Assay TC1000011467.hg.1 ENST00000425267.3 Ensembl
Affymetrix Human Clariom D Assay TC1000011467.hg.1 ENST00000425267.3 Ensembl
Affymetrix Human Clariom D Assay TC1000011467.hg.1 ENST00000425267.3 Ensembl
Affymetrix Human Clariom D Assay TC1000011467.hg.1 ENST00000425267.3 Ensembl
Affymetrix Human Clariom D Assay TC1000011467.hg.1 ENST00000425267.3 Ensembl
Affymetrix Human Clariom D Assay TC1000011467.hg.1 ENST00000425267.3 Ensembl
Affymetrix Human Clariom D Assay TC1000011467.hg.1 ENST00000425267.3 Ensembl
Affymetrix Human Clariom D Assay TC1000011467.hg.1 ENST00000425267.3 Ensembl
Affymetrix Human Clariom D Assay TC1000011467.hg.1 ENST00000425267.3 Ensembl
Affymetrix Human Clariom D Assay TC1000011467.hg.1 ENST00000425267.3 Ensembl
Affymetrix Human Clariom D Assay TC1000011467.hg.1 ENST00000425267.3 Ensembl
Affymetrix Human Clariom D Assay TC1000011467.hg.1 ENST00000425267.3 Ensembl
Affymetrix Human Clariom D Assay TC1000011467.hg.1 ENST00000425267.3 Ensembl
Affymetrix Human Clariom D Assay TC1000011467.hg.1 ENST00000425267.3 Ensembl
Affymetrix Human Clariom D Assay TC1000011467.hg.1 ENST00000440198.1 Ensembl
Affymetrix Human Clariom D Assay TC1000011467.hg.1 ENST00000440198.1 Ensembl
Affymetrix Human Clariom D Assay TC1000011467.hg.1 ENST00000440198.1 Ensembl
Affymetrix Human Clariom D Assay TC1000011467.hg.1 ENST00000440198.1 Ensembl
Affymetrix Human Clariom D Assay TC1000011467.hg.1 ENST00000440198.1 Ensembl
Affymetrix Human Clariom D Assay TC1000011467.hg.1 ENST00000440198.1 Ensembl
Affymetrix Human Clariom D Assay TC1000011467.hg.1 ENST00000440198.1 Ensembl
Affymetrix Human Clariom D Assay TC1000011467.hg.1 ENST00000440198.1 Ensembl
Affymetrix Human Clariom D Assay TC1000011467.hg.1 ENST00000440198.1 Ensembl
Affymetrix Human Clariom D Assay TC1000011467.hg.1 ENST00000440198.1 Ensembl
Affymetrix Human Clariom D Assay TC1000011467.hg.1 ENST00000440198.1 Ensembl
Affymetrix Human Clariom D Assay TC1000011467.hg.1 ENST00000596633.1 Ensembl
Affymetrix Human Clariom D Assay TC1000011467.hg.1 ENST00000596633.1 Ensembl
Affymetrix Human Clariom D Assay TC1000011467.hg.1 ENST00000596633.1 Ensembl
Affymetrix Human Clariom D Assay TC1000011467.hg.1 ENST00000596633.1 Ensembl
Affymetrix Human Clariom D Assay TC1000011467.hg.1 ENST00000596633.1 Ensembl
Affymetrix Human Clariom D Assay TC1000011467.hg.1 ENST00000596633.1 Ensembl
Affymetrix Human Clariom D Assay TC1000011467.hg.1 ENST00000596633.1 Ensembl
Affymetrix Human Clariom D Assay TC1000011467.hg.1 ENST00000596633.1 Ensembl
Affymetrix Human Clariom D Assay TC1000011467.hg.1 ENST00000596633.1 Ensembl
Affymetrix Human Clariom D Assay TC1000011467.hg.1 ENST00000596633.1 Ensembl
Affymetrix Human Clariom D Assay TC1000011467.hg.1 ENST00000596633.1 Ensembl
Affymetrix Human Clariom D Assay TC1000011467.hg.1 ENST00000596633.1 Ensembl
Affymetrix Human Clariom D Assay TC1000011467.hg.1 ENST00000596633.1 Ensembl
Affymetrix Human Clariom D Assay TC1000011467.hg.1 ENST00000596633.1 Ensembl
Affymetrix Human Clariom D Assay TC1000011467.hg.1 ENST00000596633.1 Ensembl
Affymetrix Human Clariom D Assay TC1000011467.hg.1 ENST00000596633.1 Ensembl
Affymetrix Human Clariom D Assay TC1000011467.hg.1 ENST00000611247.1 Ensembl
Affymetrix Human Clariom D Assay TC1000011467.hg.1 ENST00000611247.1 Ensembl
Affymetrix Human Clariom D Assay TC1000011467.hg.1 ENST00000611247.1 Ensembl
Affymetrix Human Clariom D Assay TC1000011467.hg.1 ENST00000611247.1 Ensembl
Affymetrix Human Clariom D Assay TC1000011467.hg.1 NR_033969.1 Refseq
Affymetrix Human Clariom D Assay TC1000011467.hg.1 NR_033969.1 Refseq
Affymetrix Human Clariom D Assay TC1000011467.hg.1 NR_033969.1 Refseq
Affymetrix Human Clariom D Assay TC1000011467.hg.1 NR_033969.1 Refseq
Affymetrix Human Clariom D Assay TC1000011467.hg.1 NR_033969.1 Refseq
Affymetrix Human Clariom D Assay TC1000011467.hg.1 NR_033969.1 Refseq
Affymetrix Human Clariom D Assay TC1000011467.hg.1 NR_033969.1 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-c694fbbc8-74xkl:80/100.66.128.6:80.
git-commit: 2cf2d5f40d96ab2bfa499d940143ca22e465fdb8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0