Assay Details
Gene Symbol: GNA11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_002067.4 | NP_002058.2 | 7 | 1974 | 90 | |
GenBank mRNA | AL110227.1 | - | 1 | 323 | 90 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AL110227.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
skeletal system development
action potential
signal transduction
G-protein coupled acetylcholine receptor signaling pathway
heart development
phototransduction, visible light
entrainment of circadian clock
platelet activation
regulation of melanocyte differentiation
developmental pigmentation
phospholipase C-activating dopamine receptor signaling pathway
cellular response to pH
GTPase activity
signal transducer activity
GTP binding
G-protein beta/gamma-subunit complex binding
type 2A serotonin receptor binding
metal ion binding
skeletal system development
action potential
signal transduction
G-protein coupled acetylcholine receptor signaling pathway
heart development
phototransduction, visible light
entrainment of circadian clock
platelet activation
regulation of melanocyte differentiation
developmental pigmentation
phospholipase C-activating dopamine receptor signaling pathway
cellular response to pH
GTPase activity
signal transducer activity
GTP binding
G-protein beta/gamma-subunit complex binding
type 2A serotonin receptor binding
metal ion binding
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1900009276.hg.1 | ENST00000585980.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009276.hg.1 | ENST00000585980.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009276.hg.1 | ENST00000585980.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006616.hg.1 | ENST00000078429.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006616.hg.1 | ENST00000078429.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006616.hg.1 | ENST00000078429.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006616.hg.1 | ENST00000078429.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006616.hg.1 | ENST00000078429.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006616.hg.1 | ENST00000078429.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006616.hg.1 | ENST00000078429.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006616.hg.1 | ENST00000078429.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006616.hg.1 | ENST00000078429.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006616.hg.1 | ENST00000078429.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006616.hg.1 | ENST00000078429.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006616.hg.1 | ENST00000078429.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006616.hg.1 | ENST00000078429.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006616.hg.1 | ENST00000078429.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006616.hg.1 | ENST00000078429.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006616.hg.1 | ENST00000078429.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006616.hg.1 | ENST00000078429.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006616.hg.1 | ENST00000078429.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006616.hg.1 | ENST00000586180.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006616.hg.1 | ENST00000586180.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006616.hg.1 | ENST00000586180.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006616.hg.1 | ENST00000586180.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006616.hg.1 | ENST00000586180.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006616.hg.1 | ENST00000586763.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006616.hg.1 | ENST00000586763.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006616.hg.1 | ENST00000586763.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006616.hg.1 | ENST00000587636.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006616.hg.1 | ENST00000587636.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006616.hg.1 | ENST00000587636.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006616.hg.1 | ENST00000587636.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006616.hg.1 | ENST00000587636.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006616.hg.1 | ENST00000587636.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006616.hg.1 | ENST00000588401.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006616.hg.1 | ENST00000588401.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006616.hg.1 | ENST00000588401.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006616.hg.1 | ENST00000590534.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006616.hg.1 | ENST00000590534.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006616.hg.1 | ENST00000590534.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006616.hg.1 | ENST00000590534.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006616.hg.1 | ENST00000590534.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006616.hg.1 | ENST00000590534.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006616.hg.1 | ENST00000590534.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006616.hg.1 | ENST00000590534.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006616.hg.1 | ENST00000590534.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006616.hg.1 | ENST00000590534.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006616.hg.1 | ENST00000591301.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006616.hg.1 | ENST00000591301.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006616.hg.1 | ENST00000591301.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900006616.hg.1 | NM_002067.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900006616.hg.1 | NM_002067.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900006616.hg.1 | NM_002067.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900006616.hg.1 | NM_002067.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900006616.hg.1 | NM_002067.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900006616.hg.1 | NM_002067.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900006616.hg.1 | NM_002067.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900006616.hg.1 | NM_002067.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900006616.hg.1 | NM_002067.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900006616.hg.1 | NM_002067.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900006616.hg.1 | NM_002067.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900006616.hg.1 | NM_002067.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900006616.hg.1 | NM_002067.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900006616.hg.1 | NM_002067.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900006616.hg.1 | NM_002067.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900006616.hg.1 | NM_002067.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900006616.hg.1 | NM_002067.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900006616.hg.1 | NM_002067.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1900009276.hg.1 | ENST00000585980.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009276.hg.1 | ENST00000585980.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009276.hg.1 | ENST00000585980.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006616.hg.1 | ENST00000078429.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006616.hg.1 | ENST00000078429.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006616.hg.1 | ENST00000078429.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006616.hg.1 | ENST00000078429.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006616.hg.1 | ENST00000078429.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006616.hg.1 | ENST00000078429.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006616.hg.1 | ENST00000078429.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006616.hg.1 | ENST00000078429.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006616.hg.1 | ENST00000078429.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006616.hg.1 | ENST00000078429.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006616.hg.1 | ENST00000078429.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006616.hg.1 | ENST00000078429.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006616.hg.1 | ENST00000078429.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006616.hg.1 | ENST00000078429.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006616.hg.1 | ENST00000078429.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006616.hg.1 | ENST00000078429.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006616.hg.1 | ENST00000078429.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006616.hg.1 | ENST00000078429.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006616.hg.1 | ENST00000586180.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006616.hg.1 | ENST00000586180.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006616.hg.1 | ENST00000586180.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006616.hg.1 | ENST00000586180.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006616.hg.1 | ENST00000586180.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006616.hg.1 | ENST00000586763.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006616.hg.1 | ENST00000586763.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006616.hg.1 | ENST00000586763.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006616.hg.1 | ENST00000587636.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006616.hg.1 | ENST00000587636.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006616.hg.1 | ENST00000587636.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006616.hg.1 | ENST00000587636.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006616.hg.1 | ENST00000587636.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006616.hg.1 | ENST00000587636.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006616.hg.1 | ENST00000588401.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006616.hg.1 | ENST00000588401.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006616.hg.1 | ENST00000588401.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006616.hg.1 | ENST00000590534.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006616.hg.1 | ENST00000590534.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006616.hg.1 | ENST00000590534.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006616.hg.1 | ENST00000590534.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006616.hg.1 | ENST00000590534.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006616.hg.1 | ENST00000590534.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006616.hg.1 | ENST00000590534.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006616.hg.1 | ENST00000590534.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006616.hg.1 | ENST00000590534.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006616.hg.1 | ENST00000590534.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006616.hg.1 | ENST00000591301.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006616.hg.1 | ENST00000591301.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006616.hg.1 | ENST00000591301.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900006616.hg.1 | NM_002067.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1900006616.hg.1 | NM_002067.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1900006616.hg.1 | NM_002067.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1900006616.hg.1 | NM_002067.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1900006616.hg.1 | NM_002067.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1900006616.hg.1 | NM_002067.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1900006616.hg.1 | NM_002067.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1900006616.hg.1 | NM_002067.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1900006616.hg.1 | NM_002067.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1900006616.hg.1 | NM_002067.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1900006616.hg.1 | NM_002067.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1900006616.hg.1 | NM_002067.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1900006616.hg.1 | NM_002067.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1900006616.hg.1 | NM_002067.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1900006616.hg.1 | NM_002067.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1900006616.hg.1 | NM_002067.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1900006616.hg.1 | NM_002067.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1900006616.hg.1 | NM_002067.4 | Refseq |
