This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05004908_m1
See other REXO1 GE Assays ›
Gene Symbol:
REXO1
Gene Name:
RNA exonuclease 1 homolog
Gene Aliases:
ELOABP1, EloA-BP1, REX1, TCEB3BP1
Chromosome Location:
Chr.19: 1815246 - 1848684 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05004908_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: REXO1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA BC016923.1 - 1 - 2 243 69

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the BC016923.1 transcripts.

Set Membership:

Amplicon length less than or equal to 70 Mammalian Gene Collection Homo sapiens Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1900009188.hg.1 ENST00000170168.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009188.hg.1 ENST00000170168.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009188.hg.1 ENST00000170168.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009188.hg.1 ENST00000170168.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009188.hg.1 ENST00000170168.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009188.hg.1 ENST00000170168.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009188.hg.1 ENST00000170168.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009188.hg.1 ENST00000170168.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009188.hg.1 ENST00000170168.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009188.hg.1 ENST00000170168.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009188.hg.1 ENST00000170168.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009188.hg.1 ENST00000170168.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009188.hg.1 ENST00000170168.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009188.hg.1 ENST00000170168.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009188.hg.1 ENST00000170168.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009188.hg.1 ENST00000170168.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009188.hg.1 ENST00000170168.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009188.hg.1 ENST00000170168.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009188.hg.1 ENST00000170168.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009188.hg.1 ENST00000170168.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009188.hg.1 ENST00000170168.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009188.hg.1 ENST00000170168.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009188.hg.1 ENST00000170168.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009188.hg.1 ENST00000170168.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009188.hg.1 ENST00000170168.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009188.hg.1 ENST00000170168.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009188.hg.1 ENST00000170168.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009188.hg.1 ENST00000170168.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009188.hg.1 ENST00000170168.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009188.hg.1 ENST00000411312.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009188.hg.1 ENST00000586291.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009188.hg.1 ENST00000586291.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009188.hg.1 ENST00000586291.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009188.hg.1 ENST00000586291.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009188.hg.1 ENST00000586291.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009188.hg.1 ENST00000586291.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009188.hg.1 ENST00000586291.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009188.hg.1 ENST00000586343.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009188.hg.1 ENST00000586343.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009188.hg.1 ENST00000586343.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009188.hg.1 ENST00000586343.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009188.hg.1 ENST00000586343.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009188.hg.1 ENST00000586343.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009188.hg.1 ENST00000586343.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009188.hg.1 ENST00000586343.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009188.hg.1 ENST00000586343.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009188.hg.1 ENST00000587404.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009188.hg.1 ENST00000587404.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009188.hg.1 ENST00000587404.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009188.hg.1 ENST00000587404.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009188.hg.1 ENST00000587404.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009188.hg.1 ENST00000587524.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009188.hg.1 ENST00000587524.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009188.hg.1 ENST00000587524.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009188.hg.1 ENST00000587524.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009188.hg.1 ENST00000588743.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009188.hg.1 ENST00000588743.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009188.hg.1 ENST00000588743.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009188.hg.1 ENST00000588743.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009188.hg.1 ENST00000588743.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009188.hg.1 ENST00000588743.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009188.hg.1 ENST00000590936.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009188.hg.1 ENST00000590936.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009188.hg.1 ENST00000590936.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009188.hg.1 ENST00000590936.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009188.hg.1 ENST00000590936.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009188.hg.1 ENST00000590936.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009188.hg.1 ENST00000590936.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009188.hg.1 ENST00000590936.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009188.hg.1 ENST00000590936.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009188.hg.1 ENST00000590936.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009188.hg.1 ENST00000590936.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009188.hg.1 ENST00000590936.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009188.hg.1 ENST00000590936.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009188.hg.1 ENST00000590936.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009188.hg.1 ENST00000590936.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009188.hg.1 ENST00000590936.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009188.hg.1 ENST00000590936.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009188.hg.1 NM_020695.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900009188.hg.1 NM_020695.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900009188.hg.1 NM_020695.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900009188.hg.1 NM_020695.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900009188.hg.1 NM_020695.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900009188.hg.1 NM_020695.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900009188.hg.1 NM_020695.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900009188.hg.1 NM_020695.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900009188.hg.1 NM_020695.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900009188.hg.1 NM_020695.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900009188.hg.1 NM_020695.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900009188.hg.1 NM_020695.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900009188.hg.1 NM_020695.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900009188.hg.1 NM_020695.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900009188.hg.1 NM_020695.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900009188.hg.1 NM_020695.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900009188.hg.1 NM_020695.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900009188.hg.1 NM_020695.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900009188.hg.1 NM_020695.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900009188.hg.1 NM_020695.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900009188.hg.1 NM_020695.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900009188.hg.1 NM_020695.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900009188.hg.1 NM_020695.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900009188.hg.1 NM_020695.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900009188.hg.1 NM_020695.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900009188.hg.1 NM_020695.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900009188.hg.1 NM_020695.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900009188.hg.1 NM_020695.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900009188.hg.1 NM_020695.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900009188.hg.1 NR_031730.1 Refseq
Affymetrix Human Clariom D Assay TC1900009188.hg.1 ENST00000170168.8 Ensembl
Affymetrix Human Clariom D Assay TC1900009188.hg.1 ENST00000170168.8 Ensembl
Affymetrix Human Clariom D Assay TC1900009188.hg.1 ENST00000170168.8 Ensembl
Affymetrix Human Clariom D Assay TC1900009188.hg.1 ENST00000170168.8 Ensembl
Affymetrix Human Clariom D Assay TC1900009188.hg.1 ENST00000170168.8 Ensembl
Affymetrix Human Clariom D Assay TC1900009188.hg.1 ENST00000170168.8 Ensembl
Affymetrix Human Clariom D Assay TC1900009188.hg.1 ENST00000170168.8 Ensembl
Affymetrix Human Clariom D Assay TC1900009188.hg.1 ENST00000170168.8 Ensembl
Affymetrix Human Clariom D Assay TC1900009188.hg.1 ENST00000170168.8 Ensembl
Affymetrix Human Clariom D Assay TC1900009188.hg.1 ENST00000170168.8 Ensembl
Affymetrix Human Clariom D Assay TC1900009188.hg.1 ENST00000170168.8 Ensembl
Affymetrix Human Clariom D Assay TC1900009188.hg.1 ENST00000170168.8 Ensembl
Affymetrix Human Clariom D Assay TC1900009188.hg.1 ENST00000170168.8 Ensembl
Affymetrix Human Clariom D Assay TC1900009188.hg.1 ENST00000170168.8 Ensembl
Affymetrix Human Clariom D Assay TC1900009188.hg.1 ENST00000170168.8 Ensembl
Affymetrix Human Clariom D Assay TC1900009188.hg.1 ENST00000170168.8 Ensembl
Affymetrix Human Clariom D Assay TC1900009188.hg.1 ENST00000170168.8 Ensembl
Affymetrix Human Clariom D Assay TC1900009188.hg.1 ENST00000170168.8 Ensembl
Affymetrix Human Clariom D Assay TC1900009188.hg.1 ENST00000170168.8 Ensembl
Affymetrix Human Clariom D Assay TC1900009188.hg.1 ENST00000170168.8 Ensembl
Affymetrix Human Clariom D Assay TC1900009188.hg.1 ENST00000170168.8 Ensembl
Affymetrix Human Clariom D Assay TC1900009188.hg.1 ENST00000170168.8 Ensembl
Affymetrix Human Clariom D Assay TC1900009188.hg.1 ENST00000170168.8 Ensembl
Affymetrix Human Clariom D Assay TC1900009188.hg.1 ENST00000170168.8 Ensembl
Affymetrix Human Clariom D Assay TC1900009188.hg.1 ENST00000170168.8 Ensembl
Affymetrix Human Clariom D Assay TC1900009188.hg.1 ENST00000170168.8 Ensembl
Affymetrix Human Clariom D Assay TC1900009188.hg.1 ENST00000170168.8 Ensembl
Affymetrix Human Clariom D Assay TC1900009188.hg.1 ENST00000170168.8 Ensembl
Affymetrix Human Clariom D Assay TC1900009188.hg.1 ENST00000170168.8 Ensembl
Affymetrix Human Clariom D Assay TC1900009188.hg.1 ENST00000411312.1 Ensembl
Affymetrix Human Clariom D Assay TC1900009188.hg.1 ENST00000586291.1 Ensembl
Affymetrix Human Clariom D Assay TC1900009188.hg.1 ENST00000586291.1 Ensembl
Affymetrix Human Clariom D Assay TC1900009188.hg.1 ENST00000586291.1 Ensembl
Affymetrix Human Clariom D Assay TC1900009188.hg.1 ENST00000586291.1 Ensembl
Affymetrix Human Clariom D Assay TC1900009188.hg.1 ENST00000586291.1 Ensembl
Affymetrix Human Clariom D Assay TC1900009188.hg.1 ENST00000586291.1 Ensembl
Affymetrix Human Clariom D Assay TC1900009188.hg.1 ENST00000586291.1 Ensembl
Affymetrix Human Clariom D Assay TC1900009188.hg.1 ENST00000586343.2 Ensembl
Affymetrix Human Clariom D Assay TC1900009188.hg.1 ENST00000586343.2 Ensembl
Affymetrix Human Clariom D Assay TC1900009188.hg.1 ENST00000586343.2 Ensembl
Affymetrix Human Clariom D Assay TC1900009188.hg.1 ENST00000586343.2 Ensembl
Affymetrix Human Clariom D Assay TC1900009188.hg.1 ENST00000586343.2 Ensembl
Affymetrix Human Clariom D Assay TC1900009188.hg.1 ENST00000586343.2 Ensembl
Affymetrix Human Clariom D Assay TC1900009188.hg.1 ENST00000586343.2 Ensembl
Affymetrix Human Clariom D Assay TC1900009188.hg.1 ENST00000586343.2 Ensembl
Affymetrix Human Clariom D Assay TC1900009188.hg.1 ENST00000586343.2 Ensembl
Affymetrix Human Clariom D Assay TC1900009188.hg.1 ENST00000587404.1 Ensembl
Affymetrix Human Clariom D Assay TC1900009188.hg.1 ENST00000587404.1 Ensembl
Affymetrix Human Clariom D Assay TC1900009188.hg.1 ENST00000587404.1 Ensembl
Affymetrix Human Clariom D Assay TC1900009188.hg.1 ENST00000587404.1 Ensembl
Affymetrix Human Clariom D Assay TC1900009188.hg.1 ENST00000587404.1 Ensembl
Affymetrix Human Clariom D Assay TC1900009188.hg.1 ENST00000587524.1 Ensembl
Affymetrix Human Clariom D Assay TC1900009188.hg.1 ENST00000587524.1 Ensembl
Affymetrix Human Clariom D Assay TC1900009188.hg.1 ENST00000587524.1 Ensembl
Affymetrix Human Clariom D Assay TC1900009188.hg.1 ENST00000587524.1 Ensembl
Affymetrix Human Clariom D Assay TC1900009188.hg.1 ENST00000588743.2 Ensembl
Affymetrix Human Clariom D Assay TC1900009188.hg.1 ENST00000588743.2 Ensembl
Affymetrix Human Clariom D Assay TC1900009188.hg.1 ENST00000588743.2 Ensembl
Affymetrix Human Clariom D Assay TC1900009188.hg.1 ENST00000588743.2 Ensembl
Affymetrix Human Clariom D Assay TC1900009188.hg.1 ENST00000588743.2 Ensembl
Affymetrix Human Clariom D Assay TC1900009188.hg.1 ENST00000588743.2 Ensembl
Affymetrix Human Clariom D Assay TC1900009188.hg.1 ENST00000590936.5 Ensembl
Affymetrix Human Clariom D Assay TC1900009188.hg.1 ENST00000590936.5 Ensembl
Affymetrix Human Clariom D Assay TC1900009188.hg.1 ENST00000590936.5 Ensembl
Affymetrix Human Clariom D Assay TC1900009188.hg.1 ENST00000590936.5 Ensembl
Affymetrix Human Clariom D Assay TC1900009188.hg.1 ENST00000590936.5 Ensembl
Affymetrix Human Clariom D Assay TC1900009188.hg.1 ENST00000590936.5 Ensembl
Affymetrix Human Clariom D Assay TC1900009188.hg.1 ENST00000590936.5 Ensembl
Affymetrix Human Clariom D Assay TC1900009188.hg.1 ENST00000590936.5 Ensembl
Affymetrix Human Clariom D Assay TC1900009188.hg.1 ENST00000590936.5 Ensembl
Affymetrix Human Clariom D Assay TC1900009188.hg.1 ENST00000590936.5 Ensembl
Affymetrix Human Clariom D Assay TC1900009188.hg.1 ENST00000590936.5 Ensembl
Affymetrix Human Clariom D Assay TC1900009188.hg.1 ENST00000590936.5 Ensembl
Affymetrix Human Clariom D Assay TC1900009188.hg.1 ENST00000590936.5 Ensembl
Affymetrix Human Clariom D Assay TC1900009188.hg.1 ENST00000590936.5 Ensembl
Affymetrix Human Clariom D Assay TC1900009188.hg.1 ENST00000590936.5 Ensembl
Affymetrix Human Clariom D Assay TC1900009188.hg.1 ENST00000590936.5 Ensembl
Affymetrix Human Clariom D Assay TC1900009188.hg.1 ENST00000590936.5 Ensembl
Affymetrix Human Clariom D Assay TC1900009188.hg.1 NM_020695.3 Refseq
Affymetrix Human Clariom D Assay TC1900009188.hg.1 NM_020695.3 Refseq
Affymetrix Human Clariom D Assay TC1900009188.hg.1 NM_020695.3 Refseq
Affymetrix Human Clariom D Assay TC1900009188.hg.1 NM_020695.3 Refseq
Affymetrix Human Clariom D Assay TC1900009188.hg.1 NM_020695.3 Refseq
Affymetrix Human Clariom D Assay TC1900009188.hg.1 NM_020695.3 Refseq
Affymetrix Human Clariom D Assay TC1900009188.hg.1 NM_020695.3 Refseq
Affymetrix Human Clariom D Assay TC1900009188.hg.1 NM_020695.3 Refseq
Affymetrix Human Clariom D Assay TC1900009188.hg.1 NM_020695.3 Refseq
Affymetrix Human Clariom D Assay TC1900009188.hg.1 NM_020695.3 Refseq
Affymetrix Human Clariom D Assay TC1900009188.hg.1 NM_020695.3 Refseq
Affymetrix Human Clariom D Assay TC1900009188.hg.1 NM_020695.3 Refseq
Affymetrix Human Clariom D Assay TC1900009188.hg.1 NM_020695.3 Refseq
Affymetrix Human Clariom D Assay TC1900009188.hg.1 NM_020695.3 Refseq
Affymetrix Human Clariom D Assay TC1900009188.hg.1 NM_020695.3 Refseq
Affymetrix Human Clariom D Assay TC1900009188.hg.1 NM_020695.3 Refseq
Affymetrix Human Clariom D Assay TC1900009188.hg.1 NM_020695.3 Refseq
Affymetrix Human Clariom D Assay TC1900009188.hg.1 NM_020695.3 Refseq
Affymetrix Human Clariom D Assay TC1900009188.hg.1 NM_020695.3 Refseq
Affymetrix Human Clariom D Assay TC1900009188.hg.1 NM_020695.3 Refseq
Affymetrix Human Clariom D Assay TC1900009188.hg.1 NM_020695.3 Refseq
Affymetrix Human Clariom D Assay TC1900009188.hg.1 NM_020695.3 Refseq
Affymetrix Human Clariom D Assay TC1900009188.hg.1 NM_020695.3 Refseq
Affymetrix Human Clariom D Assay TC1900009188.hg.1 NM_020695.3 Refseq
Affymetrix Human Clariom D Assay TC1900009188.hg.1 NM_020695.3 Refseq
Affymetrix Human Clariom D Assay TC1900009188.hg.1 NM_020695.3 Refseq
Affymetrix Human Clariom D Assay TC1900009188.hg.1 NM_020695.3 Refseq
Affymetrix Human Clariom D Assay TC1900009188.hg.1 NM_020695.3 Refseq
Affymetrix Human Clariom D Assay TC1900009188.hg.1 NM_020695.3 Refseq
Affymetrix Human Clariom D Assay TC1900009188.hg.1 NR_031730.1 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0