This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05002795_s1
See other KMT2D GE Assays ›
Gene Symbol:
KMT2D
Gene Name:
lysine methyltransferase 2D
Gene Aliases:
AAD10, ALR, CAGL114, KABUK1, KMS, MLL2, MLL4, TNRC21
Chromosome Location:
Chr.12: 49018975 - 49060884 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05002795_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: KMT2D
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_003482.3 NP_003473.3 39 11549 106
XM_005269162.4 XP_005269219.1 40 12828 106
XM_006719614.3 XP_006719677.1 40 12837 106
XM_006719616.3 XP_006719679.1 40 12825 106
XM_011538770.2 XP_011537072.1 40 12840 106
XM_011538771.2 XP_011537073.1 40 12837 106
XM_011538772.2 XP_011537074.1 40 12831 106
XM_011538773.2 XP_011537075.1 40 12828 106
XM_011538774.2 XP_011537076.1 40 12820 106
XM_011538776.2 XP_011537078.1 40 12747 106
GenBank mRNA AB209494.1 - 6 3051 106
AF010403.1 - 39 10724 106
AF010404.1 - 30 9809 106
Y08267.1 - 1 487 106

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene KMT2D

Gene Ontology Categories:

Function(s) Process(es)

oocyte growth
chromatin silencing
transcription, DNA-templated
regulation of transcription, DNA-templated
positive regulation of cell proliferation
positive regulation of intracellular estrogen receptor signaling pathway
response to estrogen
positive regulation of transcription from RNA polymerase II promoter
oogenesis
histone H3-K4 methylation
beta-catenin-TCF complex assembly
DNA binding
protein binding
zinc ion binding
histone-lysine N-methyltransferase activity
histone methyltransferase activity (H3-K4 specific)
transcription regulatory region DNA binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0