This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05002206_sH
See other ZNF239 GE Assays ›
Gene Symbol:
ZNF239
Gene Name:
zinc finger protein 239
Gene Aliases:
HOK-2, MOK2
Chromosome Location:
Chr.10: 43554516 - 43574619 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05002206_sH
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ZNF239
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001099282.1 NP_001092752.1 4 1153 100
NM_001099283.1 NP_001092753.1 2 990 100
NM_001099284.1 NP_001092754.1 3 1113 100
NM_001324347.1 NP_001311276.1 3 1031 100
NM_001324348.1 NP_001311277.1 2 1059 100
NM_001324349.1 NP_001311278.1 4 1223 100
NM_001324350.1 NP_001311279.1 3 1100 100
NM_001324351.1 NP_001311280.1 3 1182 100
NM_001324352.1 NP_001311281.1 4 1158 100
NM_001324353.1 NP_001311282.1 5 1263 100
NM_005674.2 NP_005665.2 2 1472 100
XM_005271832.2 XP_005271889.1 2 985 100
XM_006718001.2 XP_006718064.1 2 1084 100
XM_006718003.3 XP_006718066.1 2 984 100
XM_011540232.2 XP_011538534.1 3 4071 100
XM_011540234.2 XP_011538536.1 3 1118 100
XM_011540235.2 XP_011538537.1 3 1117 100
XM_011540236.2 XP_011538538.1 3 1126 100
XM_011540237.2 XP_011538539.1 3 1076 100
XM_011540238.2 XP_011538540.1 2 928 100
XM_017016740.1 XP_016872229.1 4 1155 100
GenBank mRNA AK292013.1 - 3 1109 100
BC026030.1 - 4 1097 5460088 100
X82125.1 - 2 1472 100

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene ZNF239

Panther Classification:

Molecular Function -

C2H2 zinc finger transcription factor

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
DNA binding
transcription factor activity, sequence-specific DNA binding
RNA binding
protein binding
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1000010446.hg.1 ENST00000306006.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010446.hg.1 ENST00000306006.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010446.hg.1 ENST00000306006.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010446.hg.1 ENST00000306006.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010446.hg.1 ENST00000306006.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010446.hg.1 ENST00000306006.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010446.hg.1 ENST00000374446.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010446.hg.1 ENST00000374446.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010446.hg.1 ENST00000374446.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010446.hg.1 ENST00000374446.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010446.hg.1 ENST00000374446.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010446.hg.1 ENST00000374446.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010446.hg.1 ENST00000426961.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010446.hg.1 ENST00000426961.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010446.hg.1 ENST00000426961.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010446.hg.1 ENST00000426961.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010446.hg.1 ENST00000426961.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010446.hg.1 ENST00000491188.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010446.hg.1 ENST00000491188.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010446.hg.1 ENST00000535642.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010446.hg.1 ENST00000535642.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010446.hg.1 ENST00000535642.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010446.hg.1 ENST00000535642.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010446.hg.1 NM_001099282.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010446.hg.1 NM_001099282.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010446.hg.1 NM_001099282.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010446.hg.1 NM_001099282.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010446.hg.1 NM_001099282.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010446.hg.1 NM_001099282.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010446.hg.1 NM_001099283.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010446.hg.1 NM_001099283.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010446.hg.1 NM_001099283.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010446.hg.1 NM_001099283.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010446.hg.1 NM_001099284.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010446.hg.1 NM_001099284.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010446.hg.1 NM_001099284.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010446.hg.1 NM_001099284.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010446.hg.1 NM_001099284.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010446.hg.1 NM_005674.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010446.hg.1 NM_005674.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010446.hg.1 NM_005674.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010446.hg.1 NM_005674.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010446.hg.1 NM_005674.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010446.hg.1 NM_005674.2 Refseq
Affymetrix Human Clariom D Assay TC1000010446.hg.1 ENST00000306006.10 Ensembl
Affymetrix Human Clariom D Assay TC1000010446.hg.1 ENST00000306006.10 Ensembl
Affymetrix Human Clariom D Assay TC1000010446.hg.1 ENST00000306006.10 Ensembl
Affymetrix Human Clariom D Assay TC1000010446.hg.1 ENST00000306006.10 Ensembl
Affymetrix Human Clariom D Assay TC1000010446.hg.1 ENST00000306006.10 Ensembl
Affymetrix Human Clariom D Assay TC1000010446.hg.1 ENST00000306006.10 Ensembl
Affymetrix Human Clariom D Assay TC1000010446.hg.1 ENST00000374446.6 Ensembl
Affymetrix Human Clariom D Assay TC1000010446.hg.1 ENST00000374446.6 Ensembl
Affymetrix Human Clariom D Assay TC1000010446.hg.1 ENST00000374446.6 Ensembl
Affymetrix Human Clariom D Assay TC1000010446.hg.1 ENST00000374446.6 Ensembl
Affymetrix Human Clariom D Assay TC1000010446.hg.1 ENST00000374446.6 Ensembl
Affymetrix Human Clariom D Assay TC1000010446.hg.1 ENST00000374446.6 Ensembl
Affymetrix Human Clariom D Assay TC1000010446.hg.1 ENST00000426961.1 Ensembl
Affymetrix Human Clariom D Assay TC1000010446.hg.1 ENST00000426961.1 Ensembl
Affymetrix Human Clariom D Assay TC1000010446.hg.1 ENST00000426961.1 Ensembl
Affymetrix Human Clariom D Assay TC1000010446.hg.1 ENST00000426961.1 Ensembl
Affymetrix Human Clariom D Assay TC1000010446.hg.1 ENST00000426961.1 Ensembl
Affymetrix Human Clariom D Assay TC1000010446.hg.1 ENST00000491188.1 Ensembl
Affymetrix Human Clariom D Assay TC1000010446.hg.1 ENST00000491188.1 Ensembl
Affymetrix Human Clariom D Assay TC1000010446.hg.1 ENST00000535642.5 Ensembl
Affymetrix Human Clariom D Assay TC1000010446.hg.1 ENST00000535642.5 Ensembl
Affymetrix Human Clariom D Assay TC1000010446.hg.1 ENST00000535642.5 Ensembl
Affymetrix Human Clariom D Assay TC1000010446.hg.1 ENST00000535642.5 Ensembl
Affymetrix Human Clariom D Assay TC1000010446.hg.1 NM_001099282.1 Refseq
Affymetrix Human Clariom D Assay TC1000010446.hg.1 NM_001099282.1 Refseq
Affymetrix Human Clariom D Assay TC1000010446.hg.1 NM_001099282.1 Refseq
Affymetrix Human Clariom D Assay TC1000010446.hg.1 NM_001099282.1 Refseq
Affymetrix Human Clariom D Assay TC1000010446.hg.1 NM_001099282.1 Refseq
Affymetrix Human Clariom D Assay TC1000010446.hg.1 NM_001099282.1 Refseq
Affymetrix Human Clariom D Assay TC1000010446.hg.1 NM_001099283.1 Refseq
Affymetrix Human Clariom D Assay TC1000010446.hg.1 NM_001099283.1 Refseq
Affymetrix Human Clariom D Assay TC1000010446.hg.1 NM_001099283.1 Refseq
Affymetrix Human Clariom D Assay TC1000010446.hg.1 NM_001099283.1 Refseq
Affymetrix Human Clariom D Assay TC1000010446.hg.1 NM_001099284.1 Refseq
Affymetrix Human Clariom D Assay TC1000010446.hg.1 NM_001099284.1 Refseq
Affymetrix Human Clariom D Assay TC1000010446.hg.1 NM_001099284.1 Refseq
Affymetrix Human Clariom D Assay TC1000010446.hg.1 NM_001099284.1 Refseq
Affymetrix Human Clariom D Assay TC1000010446.hg.1 NM_001099284.1 Refseq
Affymetrix Human Clariom D Assay TC1000010446.hg.1 NM_005674.2 Refseq
Affymetrix Human Clariom D Assay TC1000010446.hg.1 NM_005674.2 Refseq
Affymetrix Human Clariom D Assay TC1000010446.hg.1 NM_005674.2 Refseq
Affymetrix Human Clariom D Assay TC1000010446.hg.1 NM_005674.2 Refseq
Affymetrix Human Clariom D Assay TC1000010446.hg.1 NM_005674.2 Refseq
Affymetrix Human Clariom D Assay TC1000010446.hg.1 NM_005674.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0