Assay Details
Gene Symbol: UPF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK124930.1 | - | 1 | 515 | 89 | |
BX647325.1 | - | 1 | 446 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |

Gene Symbol: UPF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK124930.1 | - | 1 | 515 | 89 | |
BX647325.1 | - | 1 | 446 | 89 |
Set Membership: |
![]() ![]() ![]() ![]() |