This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05001981_m1
See other RBM20 GE Assays ›
Gene Symbol:
RBM20
Gene Name:
RNA binding motif protein 20
Gene Aliases:
-
Chromosome Location:
Chr.10: 110641934 - 110839471 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05001981_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: RBM20
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001134363.2 NP_001127835.2 1 - 2 255 79
GenBank mRNA EU822950.1 - 1 - 2 255 79

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Homo sapiens Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

mRNA processing
heart development
RNA splicing
positive regulation of RNA splicing
nucleotide binding
RNA binding
zinc ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1000008898.hg.1 ENST00000369519.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008898.hg.1 ENST00000369519.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008898.hg.1 ENST00000369519.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008898.hg.1 ENST00000369519.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008898.hg.1 ENST00000369519.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008898.hg.1 ENST00000369519.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008898.hg.1 ENST00000369519.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008898.hg.1 ENST00000369519.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008898.hg.1 ENST00000369519.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008898.hg.1 ENST00000369519.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008898.hg.1 ENST00000369519.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008898.hg.1 ENST00000369519.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008898.hg.1 ENST00000369519.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008898.hg.1 ENST00000369519.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008898.hg.1 ENST00000369519.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008898.hg.1 ENST00000369519.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008898.hg.1 ENST00000369519.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008898.hg.1 ENST00000369519.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008898.hg.1 ENST00000465774.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008898.hg.1 ENST00000465774.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008898.hg.1 ENST00000465774.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008898.hg.1 ENST00000471172.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008898.hg.1 ENST00000471172.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008898.hg.1 ENST00000471172.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008898.hg.1 ENST00000480343.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008898.hg.1 ENST00000480343.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008898.hg.1 ENST00000480343.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008898.hg.1 ENST00000480343.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008898.hg.1 NM_001134363.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008898.hg.1 NM_001134363.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008898.hg.1 NM_001134363.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008898.hg.1 NM_001134363.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008898.hg.1 NM_001134363.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008898.hg.1 NM_001134363.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008898.hg.1 NM_001134363.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008898.hg.1 NM_001134363.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008898.hg.1 NM_001134363.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008898.hg.1 NM_001134363.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008898.hg.1 NM_001134363.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008898.hg.1 NM_001134363.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008898.hg.1 NM_001134363.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008898.hg.1 NM_001134363.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008898.hg.1 NM_001134363.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008898.hg.1 NM_001134363.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008898.hg.1 NM_001134363.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008898.hg.1 NM_001134363.2 Refseq
Affymetrix Human Clariom D Assay TC1000008898.hg.1 ENST00000369519.3 Ensembl
Affymetrix Human Clariom D Assay TC1000008898.hg.1 ENST00000369519.3 Ensembl
Affymetrix Human Clariom D Assay TC1000008898.hg.1 ENST00000369519.3 Ensembl
Affymetrix Human Clariom D Assay TC1000008898.hg.1 ENST00000369519.3 Ensembl
Affymetrix Human Clariom D Assay TC1000008898.hg.1 ENST00000369519.3 Ensembl
Affymetrix Human Clariom D Assay TC1000008898.hg.1 ENST00000369519.3 Ensembl
Affymetrix Human Clariom D Assay TC1000008898.hg.1 ENST00000369519.3 Ensembl
Affymetrix Human Clariom D Assay TC1000008898.hg.1 ENST00000369519.3 Ensembl
Affymetrix Human Clariom D Assay TC1000008898.hg.1 ENST00000369519.3 Ensembl
Affymetrix Human Clariom D Assay TC1000008898.hg.1 ENST00000369519.3 Ensembl
Affymetrix Human Clariom D Assay TC1000008898.hg.1 ENST00000369519.3 Ensembl
Affymetrix Human Clariom D Assay TC1000008898.hg.1 ENST00000369519.3 Ensembl
Affymetrix Human Clariom D Assay TC1000008898.hg.1 ENST00000369519.3 Ensembl
Affymetrix Human Clariom D Assay TC1000008898.hg.1 ENST00000369519.3 Ensembl
Affymetrix Human Clariom D Assay TC1000008898.hg.1 ENST00000369519.3 Ensembl
Affymetrix Human Clariom D Assay TC1000008898.hg.1 ENST00000369519.3 Ensembl
Affymetrix Human Clariom D Assay TC1000008898.hg.1 ENST00000369519.3 Ensembl
Affymetrix Human Clariom D Assay TC1000008898.hg.1 ENST00000369519.3 Ensembl
Affymetrix Human Clariom D Assay TC1000008898.hg.1 ENST00000465774.2 Ensembl
Affymetrix Human Clariom D Assay TC1000008898.hg.1 ENST00000465774.2 Ensembl
Affymetrix Human Clariom D Assay TC1000008898.hg.1 ENST00000465774.2 Ensembl
Affymetrix Human Clariom D Assay TC1000008898.hg.1 ENST00000471172.1 Ensembl
Affymetrix Human Clariom D Assay TC1000008898.hg.1 ENST00000471172.1 Ensembl
Affymetrix Human Clariom D Assay TC1000008898.hg.1 ENST00000471172.1 Ensembl
Affymetrix Human Clariom D Assay TC1000008898.hg.1 ENST00000480343.2 Ensembl
Affymetrix Human Clariom D Assay TC1000008898.hg.1 ENST00000480343.2 Ensembl
Affymetrix Human Clariom D Assay TC1000008898.hg.1 ENST00000480343.2 Ensembl
Affymetrix Human Clariom D Assay TC1000008898.hg.1 ENST00000480343.2 Ensembl
Affymetrix Human Clariom D Assay TC1000008898.hg.1 NM_001134363.2 Refseq
Affymetrix Human Clariom D Assay TC1000008898.hg.1 NM_001134363.2 Refseq
Affymetrix Human Clariom D Assay TC1000008898.hg.1 NM_001134363.2 Refseq
Affymetrix Human Clariom D Assay TC1000008898.hg.1 NM_001134363.2 Refseq
Affymetrix Human Clariom D Assay TC1000008898.hg.1 NM_001134363.2 Refseq
Affymetrix Human Clariom D Assay TC1000008898.hg.1 NM_001134363.2 Refseq
Affymetrix Human Clariom D Assay TC1000008898.hg.1 NM_001134363.2 Refseq
Affymetrix Human Clariom D Assay TC1000008898.hg.1 NM_001134363.2 Refseq
Affymetrix Human Clariom D Assay TC1000008898.hg.1 NM_001134363.2 Refseq
Affymetrix Human Clariom D Assay TC1000008898.hg.1 NM_001134363.2 Refseq
Affymetrix Human Clariom D Assay TC1000008898.hg.1 NM_001134363.2 Refseq
Affymetrix Human Clariom D Assay TC1000008898.hg.1 NM_001134363.2 Refseq
Affymetrix Human Clariom D Assay TC1000008898.hg.1 NM_001134363.2 Refseq
Affymetrix Human Clariom D Assay TC1000008898.hg.1 NM_001134363.2 Refseq
Affymetrix Human Clariom D Assay TC1000008898.hg.1 NM_001134363.2 Refseq
Affymetrix Human Clariom D Assay TC1000008898.hg.1 NM_001134363.2 Refseq
Affymetrix Human Clariom D Assay TC1000008898.hg.1 NM_001134363.2 Refseq
Affymetrix Human Clariom D Assay TC1000008898.hg.1 NM_001134363.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0