This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05000851_s1
See other GPRC5D GE Assays ›
Gene Symbol:
GPRC5D
Gene Name:
G protein-coupled receptor class C group 5 member D
Gene Aliases:
-
Chromosome Location:
Chr.12: 12940518 - 12955908 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05000851_s1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GPRC5D
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_018654.1 NP_061124.1 1 519 69
XM_005253421.3 XP_005253478.1 3 2959 69
XM_017019582.1 XP_016875071.1 2 1052 69
XM_017019583.1 XP_016875072.1 2 1051 69
GenBank mRNA AB099817.1 - 1 519 69
AF209923.1 - 1 519 69
BC069341.1 - 1 519 7262221 69
BC107077.1 - 1 519 40027061 69
BC107078.1 - 1 519 40027066 69

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 G-protein coupled receptor 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene GPRC5D

Panther Classification:

Molecular Function -

G-protein coupled receptor

Gene Ontology Categories:

Function(s) Process(es)

G-protein coupled receptor signaling pathway
G-protein coupled receptor activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1200009980.hg.1 ENST00000228887.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200009980.hg.1 ENST00000228887.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200009980.hg.1 ENST00000228887.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200009980.hg.1 ENST00000228887.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200009980.hg.1 ENST00000396333.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200009980.hg.1 ENST00000396333.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200009980.hg.1 ENST00000541128.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200009980.hg.1 ENST00000541128.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200009980.hg.1 ENST00000541128.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200009980.hg.1 NM_018654.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200009980.hg.1 NM_018654.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200009980.hg.1 NM_018654.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200009980.hg.1 NM_018654.1 Refseq
Affymetrix Human Clariom D Assay TC1200009980.hg.1 ENST00000228887.5 Ensembl
Affymetrix Human Clariom D Assay TC1200009980.hg.1 ENST00000228887.5 Ensembl
Affymetrix Human Clariom D Assay TC1200009980.hg.1 ENST00000228887.5 Ensembl
Affymetrix Human Clariom D Assay TC1200009980.hg.1 ENST00000228887.5 Ensembl
Affymetrix Human Clariom D Assay TC1200009980.hg.1 ENST00000396333.3 Ensembl
Affymetrix Human Clariom D Assay TC1200009980.hg.1 ENST00000396333.3 Ensembl
Affymetrix Human Clariom D Assay TC1200009980.hg.1 ENST00000541128.1 Ensembl
Affymetrix Human Clariom D Assay TC1200009980.hg.1 ENST00000541128.1 Ensembl
Affymetrix Human Clariom D Assay TC1200009980.hg.1 ENST00000541128.1 Ensembl
Affymetrix Human Clariom D Assay TC1200009980.hg.1 NM_018654.1 Refseq
Affymetrix Human Clariom D Assay TC1200009980.hg.1 NM_018654.1 Refseq
Affymetrix Human Clariom D Assay TC1200009980.hg.1 NM_018654.1 Refseq
Affymetrix Human Clariom D Assay TC1200009980.hg.1 NM_018654.1 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0